Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  110  120  130  140  150  160  170  180  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1200  1605 
6211 - 6240 of 48136 products displayed
  • Jagged1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546970
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
protein jagged-1, JAG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546970 375.00
Check your price
  • JAB1 Antibody (Monoclonal, 2A10)
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA123244
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Immunoprecipitation (0.5-1 µg/ml), Western Blot (1:1000)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123244 375.00
Check your price
  • ITGA3 Antibody (Monoclonal, IA3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127165
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127165 375.00
Check your price
  • IRF7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112811
  • Size 100 µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
IRF-7H, IRF7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112811 375.00
Check your price
  • IRF7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112810
  • Size 100 µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
IRF-7H, IRF7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112810 375.00
Check your price
  • IRAK2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130549
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130549 375.00
Check your price
  • Insulin/Proinsulin Antibody, Biotin (Monoclonal, D3E7)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA127288
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Ins1, Ins2, ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127288 375.00
Check your price
  • Insulin Antibody (Monoclonal, 2D11-H5)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA135525
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135525 375.00
Check your price
  • Influenza B NP Antibody, FITC (Monoclonal, H89B)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17306
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:5-1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17306 375.00
Check your price
  • Influenza A NP Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141071
  • Size 100 µg
Applications
ELISA (1:100-1:2000), Western Blot (1-3 µg/ml)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141071 375.00
Check your price
  • Influenza A NP Antibody (Monoclonal, IV F8)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA126958
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126958 375.00
Check your price
  • Influenza A NP Antibody (Monoclonal, H72G)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17323
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17323 375.00
Check your price
  • Influenza A M2 Antibody (Monoclonal, 14C2)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA1082
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Immunoprecipitation (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Influenza Matrix protein M2, Influenza M2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1082 375.00
Check your price
  • Influenza A H6N1 MP1 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141124
  • Size 100 µg
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (0.5-2 µg/ml)
Alternate names:
Influenza H6N1, H6N1 MP1, H6N1 Major Protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141124 375.00
Check your price
  • Importin beta-1 Antibody (Monoclonal, 3E9)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3070
  • Size 100 µg
Applications
Flow Cytometry (1 ug/test), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Immunoprecipitation (Assay dependent), Inhibition Assays (Assay Dependent), Western Blot (1:5,000)
Alternate names:
Pore targeting complex 97 kDa subunit; Nuclear factor p97; Karyopherin subunit beta-1; Importin-90; SCG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3070 375.00
Check your price
  • IL20RA Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141167
  • Size 100 µg
Applications
Flow Cytometry (2-5 µg/10^6 cells), Western Blot (1-3 µg/ml)
Alternate names:
ZcytoR7; IL-20R1; Cytokine receptor family 2 member 8; Cytokine receptor class-II member 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141167 375.00
Check your price
  • IL1RA Antibody
  • Host Goat
  • Target Species Horse
  • Conjugate Unconjugated
  • Catalog # PA546924
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL1 inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546924 375.00
Check your price
  • IL1R1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546930
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.5-2.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p80; Interleukin-1 receptor type I; Interleukin-1 receptor alpha; CD121 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546930 375.00
Check your price
  • IL1F9 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546988
  • Size 100 µg
Applications
Blocking Assay (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Interleukin-1 homolog 1; Interleukin-1 family member 9; Interleukin-1 epsilon; IL-1-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546988 375.00
Check your price
  • IL17RB Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547093
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-17B receptor; IL-17 receptor homolog 1; Cytokine receptor-like 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547093 375.00
Check your price
  • IL13RA2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546976
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.2-10 µg/ml)
Alternate names:
Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546976 375.00
Check your price
  • IL10RB Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547727
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2, IL-10 receptor subunit beta, IL-10R subunit 2, IL-10R subunit beta, IL-10RB, cytokine receptor class-II CRF2-4, cytokine receptor class-II member 4, cytokine receptor family 2 member 4, cytokine receptor family II, member 4, interleukin-10 receptor subunit 2, inte
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547727 375.00
Check your price
  • IL-7 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546944
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.2- 0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-7, IL7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546944 375.00
Check your price
  • IL-27 p28 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546981
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (1-5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546981 375.00
Check your price
  • IL-27 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546993
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546993 375.00
Check your price
  • IL-26 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546973
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Protein AK155
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546973 375.00
Check your price
  • IL-21 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546962
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-21, Za11, interleukin-21, interleukin-21 isoform, CX2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546962 375.00
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA546923
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546923 375.00
Check your price
  • IL-2 Antibody
  • Host Goat
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA546915
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.75-3.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546915 375.00
Check your price
  • IL-19 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546903
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
NG.1; Melanoma differentiation-associated protein-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546903 375.00
Check your price