Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6301 - 6330 of 48882 products displayed
  • PACSIN2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA141564
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.02-1 µg/ml)
Alternate names:
Syndapin-II; Syndapin-2; Synaptic dynamin-associated protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141564 375.00
Check your price
  • p53 Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA112648
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112648 375.00
Check your price
  • P2Y2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14517
  • Size 100µg
Applications
Immunohistochemistry (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Purinergic receptor; P2U purinoceptor 1; ATP receptor; P2U receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14517 375.00
Check your price
  • P2Y2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146150
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (Assay-dependent), Immunohistochemistry (1:100-1:200), Western Blot (1:500-1:1000)
Alternate names:
Purinergic receptor; P2U purinoceptor 1; ATP receptor; P2U receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146150 375.00
Check your price
  • p21 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA133926
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
p21; Melanoma differentiation-associated protein 6; CDK-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133926 375.00
Check your price
  • p130 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191092
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
pRb2; Retinoblastoma-related protein 2; 130 kDa retinoblastoma-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191092 375.00
Check your price
  • ORC1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Xenopus
  • Conjugate Unconjugated
  • Catalog # MA190545
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190545 375.00
Check your price
  • ORC1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126189
  • Size 100µg
Applications
Western Blot (1:1250-1:2500)
Alternate names:
Replication control protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126189 375.00
Check your price
  • ORAI1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA174181
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (0.5-1 µg/ml)
Alternate names:
Transmembrane protein 142A; Protein orai-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA174181 375.00
Check your price
  • Opsin 4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA1781
  • Size 100µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (Frozen) (1:1000), Western Blot (0.1-0.2 ug/ml)
Alternate names:
Opsin-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1781 375.00
Check your price
  • OPRM1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14561
  • Size 100µg
Applications
Western Blot (1:500-1:1,000)
Alternate names:
Opioid receptor B; Mu opioid receptor; Mu opiate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14561 375.00
Check your price
  • Oncostatin M Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547002
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.02-0.04 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
MGC20461
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547002 375.00
Check your price
  • Oncostatin M Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547022
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.6-3.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
MGC20461
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547022 375.00
Check your price
  • NUP85 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547799
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Pericentrin-1; Nucleoporin Nup85; Nucleoporin Nup75; FROUNT; 85 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547799 375.00
Check your price
  • NSE Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116696
  • Size 100µg
Applications
ELISA (1:100-1:2000), Immunohistochemistry (Paraffin) (1:100-1:300), Western Blot (1:100-1:2000)
Alternate names:
Neuron-specific enolase; Neural enolase; Enolase 2; 2-phospho-D-glycerate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116696 375.00
Check your price
  • NRXN3 Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547714
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Neurexin III-beta; Neurexin-3-alpha; Neurexin III-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547714 375.00
Check your price
  • NPTXR Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546949
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
NPR, NPTXR, neuronal pentraxin receptor, NPR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546949 375.00
Check your price
  • NPM1 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1029
  • Size 100µg
Applications
Immunofluorescence (1:100), Immunohistochemistry (1:200), Western Blot (1:500-1:1000)
Alternate names:
Numatrin; Nucleolar protein NO38; Nucleolar phosphoprotein B23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1029 375.00
Check your price
  • NOXA Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141010
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1-2 µg/ml)
Alternate names:
Protein Noxa; Immediate-early-response protein APR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141010 375.00
Check your price
  • NOTUM Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112858
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1:1,000)
Alternate names:
hNOTUM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112858 375.00
Check your price
  • Nope Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546908
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Western Blot (0.1 µg/ml)
Alternate names:
Protein DDM36; Neighbor of punc e11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546908 375.00
Check your price
  • Nogo Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141202
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Nogo-66 receptor; Nogo receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141202 375.00
Check your price
  • nNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1033
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:100-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS1; Neuronal NOS; NOS type I; NC-NOS; Constitutive NOS; BNOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1033 375.00
Check your price
  • Nidogen Monoclonal Antibody for Western Blot, ICC and IHC (F)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA106501
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunohistochemistry (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Entactin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106501 375.00
Check your price
  • NGFR Monoclonal Antibody for Western Blot, IF, ICC, IHC (PFA) and ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # OSM00019A
  • Size 100µg
Applications
ELISA (5-15 µg/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (PFA fixed) (5-15 µg/ml), Western Blot (5-15 µg/ml)
Alternate names:
p75 ICD; Low-affinity nerve growth factor receptor; Low affinity neurotrophin receptor p75NTR; Gp80-LNGFR
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00019A 375.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546916
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.25-5 µg/ml)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546916 375.00
Check your price
  • NGAL Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546938
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546938 375.00
Check your price
  • NG2 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190928
  • Size 100µg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Melanoma-associated chondroitin sulfate proteoglycan; Melanoma chondroitin sulfate proteoglycan; Chondroitin sulfate proteoglycan NG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190928 375.00
Check your price
  • NFYA Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124431
  • Size 100µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (5-8 µg/ml)
Alternate names:
Nuclear transcription factor Y subunit A; CAAT box DNA-binding protein subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124431 375.00
Check your price
  • NFkB p65 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141573
  • Size 100µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1:100-1:2000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141573 375.00
Check your price