Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1605 
6361 - 6390 of 48148 products displayed
  • LGR7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141532
  • Size 100µg
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Relaxin family peptide receptor 1; Leucine-rich repeat-containing G-protein coupled receptor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141532 375.00
Check your price
  • Leptin Polyclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547023
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.05-0.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Obesity factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547023 375.00
Check your price
  • Langerin Polyclonal Antibody for Western Blot, IHC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA141053
  • Size 100µg
Applications
Flow Cytometry (1:10-1:1000), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:500), Western Blot (1-3 µg/ml)
Alternate names:
Langerin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141053 375.00
Check your price
  • LAMP1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1654A
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (1:250-1:1000)
Alternate names:
CD107 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1654A 375.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA106102
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, PRO1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106102 375.00
Check your price
  • LALBA Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191042
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Lysozyme-like protein 7; Lactose synthase B protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191042 375.00
Check your price
  • L-Myc Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547567
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
V-myc myelocytomatosis viral oncogene homolog; Protein L-Myc-1; Class E basic helix-loop-helix protein 38
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547567 375.00
Check your price
  • KSV Cyclin D Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA191431
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Kaposi's Sarcoma Virus Type D Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191431 375.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1643
  • Size 100µg
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunoprecipitation (5 µg), Western Blot (1:4000-1:6000 )
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1643 375.00
Check your price
  • KCNMB1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1924
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (2.0 µg/ml)
Alternate names:
Slo-beta-1; Maxi K channel subunit beta-1; K(VCA)beta-1; Charybdotoxin receptor subunit beta-1; Calcium-activated potassium channel, subfamily M subunit beta-1; BK channel subunit beta-1; Slowpoke-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1924 375.00
Check your price
  • Kallikrein 7 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546998
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Stratum corneum chymotryptic enzyme; Serine protease 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546998 375.00
Check your price
  • Jagged1 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546970
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
protein jagged-1, JAG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546970 375.00
Check your price
  • JAB1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA123244
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Immunoprecipitation (0.5-1 µg/ml), Western Blot (1:1000)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123244 375.00
Check your price
  • ITGA3 Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127165
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127165 375.00
Check your price
  • IRF7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112811
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
IRF-7H, IRF7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112811 375.00
Check your price
  • IRF7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112810
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
IRF-7H, IRF7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112810 375.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130549
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130549 375.00
Check your price
  • Insulin Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA135525
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135525 375.00
Check your price
  • INHBA Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547004
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (2-6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Activin beta-A chain; Erythroid differentiation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547004 375.00
Check your price
  • Influenza B NP Monoclonal Antibody, FITC conjugate for IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17306
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:5-1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17306 375.00
Check your price
  • Influenza A NP Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141071
  • Size 100µg
Applications
ELISA (1:100-1:2000), Western Blot (1-3 µg/ml)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141071 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA126958
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126958 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17323
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17323 375.00
Check your price
  • Influenza A HA Monoclonal Antibody for Western Blot, IF, ICC, IHC, ELISA, HA and IA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122684
  • Size 100µg
Applications
ELISA (Assay dependent), Hemagglutination Assay (Assay Dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay-Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H1N1, H1N1 HA, H1N1 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122684 375.00
Check your price
  • Influenza A H6N1 MP1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141124
  • Size 100µg
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (0.5-2 µg/ml)
Alternate names:
Influenza H6N1, H6N1 MP1, H6N1 Major Protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141124 375.00
Check your price
  • Importin beta-1 Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and IA
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3070
  • Size 100µg
Applications
Flow Cytometry (1 ug/test), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Immunoprecipitation (Assay dependent), Inhibition Assays (Assay Dependent), Western Blot (1:5,000)
Alternate names:
Pore targeting complex 97 kDa subunit; Nuclear factor p97; Karyopherin subunit beta-1; Importin-90; SCG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3070 375.00
Check your price
  • IL1RA Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Horse
  • Conjugate Unconjugated
  • Catalog # PA546924
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL1 inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546924 375.00
Check your price
  • IL1R1 Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546930
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.5-2.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p80; Interleukin-1 receptor type I; Interleukin-1 receptor alpha; CD121 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546930 375.00
Check your price
  • IL1F9 Polyclonal Antibody for Western Blot and BLOCK
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546988
  • Size 100µg
Applications
Blocking Assay (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Interleukin-1 homolog 1; Interleukin-1 family member 9; Interleukin-1 epsilon; IL-1-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546988 375.00
Check your price
  • IL13RA2 Polyclonal Antibody for Western Blot, IHC (P), Flow and BLOCK
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546976
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.2-10 µg/ml)
Alternate names:
Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546976 375.00
Check your price