Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1611 
6391 - 6420 of 48310 products displayed
  • Clostridium difficile Toxin A Antibody (Monoclonal, E64E)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17414
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
C. difficile Toxin A, C. difficile Toxoid A, TcdA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17414 375.00
Check your price
  • CLEC12A Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547425
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Myeloid inhibitory C-type lectin-like receptor; C-type lectin-like molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547425 375.00
Check your price
  • CLEC12A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547423
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Myeloid inhibitory C-type lectin-like receptor; Dendritic cell-associated lectin 2; C-type lectin-like molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547423 375.00
Check your price
  • cIAP1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547036
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
TNFR2-TRAF-signaling complex protein 2; RING finger protein 48; Inhibitor of apoptosis protein 2; IAP homolog B; C-IAP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547036 375.00
Check your price
  • CHRNE Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124273
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
ACHRG, MGC133376
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124273 375.00
Check your price
  • CHRNE Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124272
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
ACHRE, CMS1D, CMS1E, CMS2A, FCCMS, SCCMS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124272 375.00
Check your price
  • CHRND Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124271
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
ACHRD, CMS2A, FCCMS, SCCMS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124271 375.00
Check your price
  • CHOP Antibody (Monoclonal, 9C8)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1250
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (1/100), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1 ug/ml)
Alternate names:
Growth arrest and DNA-damage-inducible protein GADD153; CCAAT/enhancer-binding protein homologous protein; C/EBP-homologous protein 10; C/EBP-homologous protein; C/EBP zeta; Growth arrest and DNA damage-inducible protein GADD153
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1250 375.00
Check your price
  • Cholera Toxin beta Antibody (Monoclonal)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA121550
  • Size 100 µg
Applications
ELISA (1:1000000), Immunohistochemistry (Assay dependent)
Alternate names:
beta subunit cholera toxin, CTB, VC1456
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121550 375.00
Check your price
  • Chloramphenicol Acetyltransferase Antibody (Monoclonal, CAT-1)
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA191137
  • Size 100 µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191137 375.00
Check your price
  • Chlamydia LPS Antibody (Monoclonal, F70G)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17340
  • Size 100 µg
Applications
ELISA (1:20), Immunofluorescence (1:10), Immunohistochemistry (1:10)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17340 375.00
Check your price
  • CHI3L1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546996
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
YKL-40; Cartilage glycoprotein 39; 39 kDa synovial protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546996 375.00
Check your price
  • CHD7 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547918
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
ATP-dependent helicase CHD7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547918 375.00
Check your price
  • CEACAM8 Antibody (Monoclonal, 80H3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126144
  • Size 100 µg
Applications
Flow Cytometry (10 µl for 5x10^5 cells), Immunohistochemistry (Frozen) (1:25-1:50)
Alternate names:
Non-specific cross-reacting antigen NCA-95; Carcinoembryonic antigen CGM6; CD67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126144 375.00
Check your price
  • CEA Antibody, FITC (Monoclonal, C365D3(NCRC23))
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA180578
  • Size 100 µg
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180578 375.00
Check your price
  • CDK7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124252
  • Size 100 µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (1 µg/ml)
Alternate names:
TFIIH basal transcription factor complex kinase subunit; Serine/threonine-protein kinase 1; Cell division protein kinase 7; CDK-activating kinase 1; 39 kDa protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124252 375.00
Check your price
  • CDK1/CDK2 Antibody (Monoclonal, AN21.2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191380
  • Size 100 µg
Applications
ELISA (Assay-dependent), Western Blot (2 µg/ml)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog; p33 protein kinase; Cell division protein kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191380 375.00
Check your price
  • Cdc25C Antibody (Monoclonal, TC-19)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190555
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (2.5 µg/ml), Immunoprecipitation (1 µg), Western Blot (1 µg/ml)
Alternate names:
Dual specificity phosphatase Cdc25C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190555 375.00
Check your price
  • CD93 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546921
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Matrix-remodeling-associated protein 4; Complement component 1 q subcomponent receptor 1; CDw93; C1q/MBL/SPA receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546921 375.00
Check your price
  • CD90 Antibody (Monoclonal, AF-9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135307
  • Size 100 µg
Applications
ELISA (Assay dependent.), Flow Cytometry (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Thy-1 antigen; CDw90
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135307 375.00
Check your price
  • CD85 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547240
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.3-1.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Monocyte/macrophage immunoglobulin-like receptor 7; Immunoglobulin-like transcript 2; CD85 antigen-like family member J
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547240 375.00
Check your price
  • CD81 Antibody, FITC (Monoclonal, 1D6)
  • Host Mouse
  • Target Species Goat, Human, Non-human primate, Sheep
  • Conjugate FITC
  • Catalog # MA181261
  • Size 100 µg
Applications
Flow Cytometry (Neat)
Alternate names:
Tetraspanin-28; Target of the antiproliferative antibody 1; 26 kDa cell surface protein TAPA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181261 375.00
Check your price
  • CD80 Antibody (Monoclonal, AS29)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112159
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
CTLA-4 counter-receptor B7.1; BB1; Activation B7-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112159 375.00
Check your price
  • CD73 Antibody
  • Host Sheep
  • Target Species Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA547628
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Ecto-5'-nucleotidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547628 375.00
Check your price
  • CD47 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547215
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Integrin-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547215 375.00
Check your price
  • CD44v7 Antibody (Monoclonal, VFF-9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182394
  • Size 100 µg
Applications
Flow Cytometry (1/50), Immunohistochemistry (Paraffin, Frozen) (1:50), Western Blot (Assay Dependent)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Hyaluronate receptor; Hermes antigen; Heparan sulfate proteoglycan; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182394 375.00
Check your price
  • CD3e Antibody (Monoclonal, UCH-T1)
  • Host Mouse
  • Target Species Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA121454
  • Size 100 µg
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121454 375.00
Check your price
  • CD3e Antibody (Monoclonal, CA17.2A12)
  • Host Mouse
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # MA174203
  • Size 100 µg
Applications
Flow Cytometry (1/25-1/50), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174203 375.00
Check your price
  • CD34 Antibody (Monoclonal, MEC14.7)
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA122646
  • Size 100 µg
Applications
ELISA (Assay dependent), Flow Cytometry (1:500), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:50-1:500), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122646 375.00
Check your price
  • CD301 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547665
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Macrophage lectin 2; C-type lectin superfamily member 14; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage asialoglycoprotein-binding protein 1; MMGL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547665 375.00
Check your price