Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6481 - 6510 of 48881 products displayed
  • Cytokeratin 14 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124324
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Keratin-14; Cytokeratin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124324 375.00
Check your price
  • Cystatin C Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Sheep
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546963
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Cystatin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546963 375.00
Check your price
  • Cystatin C Polyclonal Antibody for Western Blot, IHC (F), IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546906
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cystatin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546906 375.00
Check your price
  • Cystatin C Polyclonal Antibody for Western Blot, IHC (P), IP and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546905
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (7.8 µg/ml), Western Blot (0.25-2 µg/ml)
Alternate names:
Post-gamma-globulin; Neuroendocrine basic polypeptide; Gamma-trace; Cystatin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546905 375.00
Check your price
  • Cystatin B Polyclonal Antibody for Western Blot, IHC and IP
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA546909
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Stefin-B; Liver thiol proteinase inhibitor; CPI-B; Cystatin-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546909 375.00
Check your price
  • CYLD Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141097
  • Size 100µg
Applications
Immunocytochemistry (5-10 µg/ml), Immunofluorescence (5-10 µg/ml), Immunohistochemistry (Assay-Dependent), Western Blot (2-4 µg/ml)
Alternate names:
Ubiquitin-specific-processing protease CYLD; Ubiquitin thioesterase CYLD; Deubiquitinating enzyme CYLD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141097 375.00
Check your price
  • Cyclophilin 40 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1028
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Rotamase D; Cyclophilin-related protein; Cyclophilin-40; 40 kDa peptidyl-prolyl cis-trans isomerase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1028 375.00
Check your price
  • Cyclin A2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Xenopus
  • Conjugate Unconjugated
  • Catalog # MA124921
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Cyclin-A2, ccna2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124921 375.00
Check your price
  • CXCR1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141096
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141096 375.00
Check your price
  • CXCR1 Monoclonal Antibody for Flow and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124668
  • Size 100µg
Applications
Flow Cytometry (10 µl for 10^6 PBMC), Inhibition Assays (0.1-0.3 µg/ml)
Alternate names:
IL-8 receptor type 1; High affinity interleukin-8 receptor A; CDw128a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124668 375.00
Check your price
  • CXCL9 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547020
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (6-20 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B9; Protein m119; Monokine induced by interferon-gamma; Gamma-interferon-induced monokine
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547020 375.00
Check your price
  • CXCL6 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546937
  • Size 100µg
Applications
Neutralization (0.8-4.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B6; Granulocyte chemotactic protein 2; Chemokine alpha 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546937 375.00
Check your price
  • CXCL3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124262
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Macrophage inflammatory protein 2-beta; Growth-regulated protein gamma; GRO-gamma(1-73)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124262 375.00
Check your price
  • CXCL2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547015
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Neutralization (0.015-0.075 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Macrophage inflammatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547015 375.00
Check your price
  • CXCL13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA129046
  • Size 100µg
Applications
Western Blot (1:5000)
Alternate names:
Small-inducible cytokine B13; CXC chemokine BLC; B lymphocyte chemoattractant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129046 375.00
Check your price
  • CXCL13 Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547018
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (20-50 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B13; CXC chemokine BLC; B lymphocyte chemoattractant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547018 375.00
Check your price
  • CXCL13 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547035
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (1-4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B13; CXC chemokine BLC; B lymphocyte chemoattractant; B cell-attracting chemokine 1; Angie
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547035 375.00
Check your price
  • CXCL10 Polyclonal Antibody for Western Blot, IF, ICC, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546999
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (1-4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine B10; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546999 375.00
Check your price
  • CXCL1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546932
  • Size 100µg
Applications
Neutralization (0.03-0.15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73); C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546932 375.00
Check your price
  • CX3CR1 Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA129025
  • Size 100µg
Applications
Immunohistochemistry (Paraffin, Frozen) (1:100-1:500), Immunoprecipitation (1:300-1:800), Western Blot (1:1000-1:2000)
Alternate names:
RBS11; Fractalkine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129025 375.00
Check your price
  • CTNNA3 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106311
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:25-1:50), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Cadherin-associated protein; Alpha T-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106311 375.00
Check your price
  • CTLA-4 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547547
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547547 375.00
Check your price
  • CSN2 Monoclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146056
  • Size 100µg
Applications
ELISA (1:100-1:2000), Immunohistochemistry (Frozen) (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
CASB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146056 375.00
Check your price
  • CSK Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA134816
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (1:250-1:300)
Alternate names:
p50CSK; Protein-tyrosine kinase MPK-2; C-Src kinase; Protein-tyrosine kinase CYL
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134816 375.00
Check your price
  • CRM1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1083
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Chromosome region maintenance 1 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1083 375.00
Check your price
  • CREB Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1850
  • Size 100µg
Applications
ChIP assay (1-3 ul), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (2 µg/ml)
Alternate names:
Cyclic AMP Response Element Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1850 375.00
Check your price
  • CRALBP Monoclonal Antibody for Western Blot, IF, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1813
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (1 ug/ml), Immunohistochemistry (Frozen) (1 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (0.02 ug/ml)
Alternate names:
Cellular retinaldehyde-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1813 375.00
Check your price
  • CPEB1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11100
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:100-1:1000)
Alternate names:
Cytoplasmic Polyadenylation Element Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11100 375.00
Check your price
  • COX2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA112236
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112236 375.00
Check your price
  • Collagen IV Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mammal
  • Conjugate Unconjugated
  • Catalog # PA128534
  • Size 100µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:5000-1:10,000), Western Blot (1:5000-1:10,000)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128534 375.00
Check your price