Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6571 - 6600 of 48877 products displayed
  • Eotaxin 3 Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547851
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (10-25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Thymic stroma chemokine-1; Small-inducible cytokine A26; Macrophage inflammatory protein 4-alpha; Eotaxin-3; CC chemokine IMAC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547851 375.00
Check your price
  • EOMES Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547818
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
T-box brain protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547818 375.00
Check your price
  • EGFR Monoclonal Antibody for IHC (F), IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124226
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124226 375.00
Check your price
  • EEA1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1063A
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
Zinc finger FYVE domain-containing protein 2; Endosome-associated protein p162
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1063A 375.00
Check your price
  • EBP50 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1090
  • Size 100µg
Applications
Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Solute carrier family 9 isoform A3 regulatory factor 1; Sodium-hydrogen exchanger regulatory factor 1; Regulatory cofactor of Na(+)/H(+) exchanger; Ezrin-radixin-moesin-binding phosphoprotein 50
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1090 375.00
Check your price
  • E. coli serotype O157 Monoclonal Antibody, FITC conjugate for IF and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # MA17304
  • Size 100µg
Applications
ELISA (1:5-1:20), Immunofluorescence (1:5-1:20)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17304 375.00
Check your price
  • E. coli serotype O157 Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17303
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17303 375.00
Check your price
  • E. coli J5 LPS Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183152
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183152 375.00
Check your price
  • E-selectin Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA106505
  • Size 100µg
Applications
Flow Cytometry (1:200), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106505 375.00
Check your price
  • DR4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130534
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
TNF-related apoptosis-inducing ligand receptor 1; Death receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130534 375.00
Check your price
  • Double Stranded DNA Monoclonal Antibody for IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135346
  • Size 100µg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135346 375.00
Check your price
  • DNMT3B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1884
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (2.0 µg/ml)
Alternate names:
DNA methyltransferase MmuIIIB; DNA methyltransferase HsaIIIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1884 375.00
Check your price
  • DNMT3A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1882
  • Size 100µg
Applications
Western Blot (4 µg/ml)
Alternate names:
DNA methyltransferase MmuIIIA; DNA methyltransferase HsaIIIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1882 375.00
Check your price
  • DNA Ligase III Monoclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Mouse
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123191
  • Size 100µg
Applications
Blocking Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay dependent)
Alternate names:
Polydeoxyribonucleotide synthase [ATP] 3; DNA ligase III
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123191 375.00
Check your price
  • DNA Ligase I Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126060
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Polydeoxyribonucleotide synthase [ATP] 1; DNA ligase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126060 375.00
Check your price
  • DMC1 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA120220
  • Size 100µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
RP1-199H16.4, DMC1H, HsLim15, LIM15, MGC150472, MGC150473, dJ199H16.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120220 375.00
Check your price
  • DLL4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546974
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Drosophila Delta homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546974 375.00
Check your price
  • DKK1 Polyclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547198
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
DKK-1, SK, dickkopf related protein-1, dickkopf-1 like, dickkopf-like protein 1, dickkopf-related protein 1, hDkk-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547198 375.00
Check your price
  • DFFB Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA130545
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500)
Alternate names:
DNA fragmentation factor 40 kDa subunit; Caspase-activated deoxyribonuclease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130545 375.00
Check your price
  • Desmocollin 3 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547909
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ARVC10, ARVD10, CDHF5, CMD1BB, HDGC, HDGC, cadherin family member 5, desmoglein-2, CDHF6, PVA, 130 kDa pemphigus vulgaris antigen, 130-kD pemphigus vulgaris antigen, cadherin family member 6, desmoglein-3, pemphigus vulgaris antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547909 375.00
Check your price
  • DCBLD2 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123482
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
Endothelial and smooth muscle cell-derived neuropilin-like protein; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123482 375.00
Check your price
  • DCAF6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141255
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Nuclear receptor interaction protein; IQ motif and WD repeat-containing protein 1; Androgen receptor complex-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141255 375.00
Check your price
  • Cytomegalovirus pp65 Monoclonal Antibody, FITC conjugate for Western Blot and IF
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17296
  • Size 100µg
Applications
Immunofluorescence (1:5-1:20), Western Blot (1:5-1:20)
Alternate names:
CMV, HHV5gp078
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17296 375.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112594
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p53-related protein; p53-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112594 375.00
Check your price
  • Cytokeratin 7 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Amphibian, Mammal
  • Conjugate Unconjugated
  • Catalog # MA190894
  • Size 100µg
Applications
Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Type-II keratin Kb7; Sarcolectin; Keratin-7; Cytokeratin-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190894 375.00
Check your price
  • Cytokeratin 5/14 Monoclonal Antibody for IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190930
  • Size 100µg
Applications
Immunohistochemistry (Assay dependent)
Alternate names:
Keratin-14; Cytokeratin-14; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190930 375.00
Check your price
  • Cytokeratin 5/14 Monoclonal Antibody for Western Blot and IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112590
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Keratin-14; Cytokeratin-14; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112590 375.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106327
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106327 375.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody for Western Blot, ICC, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA119039
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (10 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1-2 ug/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119039 375.00
Check your price
  • Cytokeratin 15/18 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190935
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-15; Cytokeratin-15
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190935 375.00
Check your price