Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  240  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1630 
6601 - 6630 of 48881 products displayed
  • ATP Synthase beta Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1930
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (2-5 ug), Western Blot (1-2 µg/ml)
Alternate names:
ATP5B, ATPMB, ATPSB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1930 375.00
Check your price
  • ASGR1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547385
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Hepatic lectin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547385 375.00
Check your price
  • ASAH2 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547509
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Non-lysosomal ceramidase; N-acylsphingosine amidohydrolase 2; LCDase; BCDase; Acylsphingosine deacylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547509 375.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126903
  • Size 100µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126903 375.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112800
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112800 375.00
Check your price
  • Apolipoprotein A2 Polyclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126900
  • Size 100µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126900 375.00
Check your price
  • Apolipoprotein A1 Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA122558
  • Size 100µg
Applications
ELISA (1:10,000-1:20,000), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122558 375.00
Check your price
  • Apolipoprotein A1 Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123058
  • Size 100µg
Applications
ELISA (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123058 375.00
Check your price
  • APH1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA12010
  • Size 100µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Aph-1alpha; Presenilin-stabilization factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA12010 375.00
Check your price
  • APC Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA126195
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Deleted in polyposis 2.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126195 375.00
Check your price
  • APC Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116631
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
Deleted in polyposis 2.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116631 375.00
Check your price
  • Annexin A4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547586
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Annexin-4; Annexin IV; Protein II; Placental anticoagulant protein II; PP4-X; P32.5; Lipocortin IV; Endonexin I; Chromobindin-4; Carbohydrate-binding protein p33/p41; 35-beta calcimedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547586 375.00
Check your price
  • ANGPTL4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11053
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Secreted protein Bk89; Hepatic fibrinogen/angiopoietin-related protein; Fasting-induced adipose factor; Angiopoietin-like protein 4; 425O18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11053 375.00
Check your price
  • Angiopoietin 1 Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA546968
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
AGP1, AGPT, ANG1, ANG-1, angiopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546968 375.00
Check your price
  • Androgen Receptor Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1111B
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (2 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1111B 375.00
Check your price
  • ALPL Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547419
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547419 375.00
Check your price
  • ALPL Monoclonal Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA135004
  • Size 100µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135004 375.00
Check your price
  • alpha-2c Adrenergic Receptor Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14518
  • Size 100µg
Applications
Immunohistochemistry (1:100-1:300), Western Blot (1:500-1:1000)
Alternate names:
Alpha-2C adrenoreceptor; Alpha-2 adrenergic receptor subtype C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14518 375.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118256
  • Size 100µg
Applications
Immunohistochemistry (Paraffin, Frozen) (3 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118256 375.00
Check your price
  • alpha COP Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1067
  • Size 100µg
Applications
Immunocytochemistry (3 µg/ml), Immunofluorescence (3 ug/ml), Western Blot (1 µg/ml)
Alternate names:
HEP-COP; Alpha-coat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1067 375.00
Check your price
  • alpha Actinin 4 Monoclonal Antibody for Western Blot, IF, IHC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1800
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1µg per 10^6 cells), Immunofluorescence (4 ug/mL), Immunohistochemistry (1:20), Immunohistochemistry (Paraffin) (4 µg/ml), Western Blot (2 ug/mL)
Alternate names:
Non-muscle alpha-actinin 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1800 375.00
Check your price
  • AgRP Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547831
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Agrt, Art
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547831 375.00
Check your price
  • Aggrecan Neo Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA11746
  • Size 100µg
Applications
Immunofluorescence (Assay dependent), Western Blot (1 ug/ml)
Alternate names:
Cartilage-specific proteoglycan core protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11746 375.00
Check your price
  • Adiponectin Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA1054
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (2-4 ug/ml), Immunohistochemistry (1:20-100), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1 ug/ml)
Alternate names:
Gelatin-binding protein; Adipose most abundant gene transcript 1 protein; Adipocyte, C1q and collagen domain-containing protein; Adipocyte complement-related 30 kDa protein; 30 kDa adipocyte complement-related protein; Adipocyte-specific protein AdipoQ
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1054 375.00
Check your price
  • ADAMTS8 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547031
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
METH-8; METH-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547031 375.00
Check your price
  • ADAM12 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546950
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Meltrin-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546950 375.00
Check your price
  • ACVRL1 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547010
  • Size 100µg
Applications
Immunoprecipitation (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
TGF-B superfamily receptor type I; Activin receptor-like kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547010 375.00
Check your price
  • ACVR2B Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547005
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Activin receptor type IIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547005 375.00
Check your price
  • ACVR2A Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA191751
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191751 375.00
Check your price
  • Acinus Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130552
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130552 375.00
Check your price