Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  250  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
6931 - 6960 of 48882 products displayed
  • Phospho-AMPK alpha-1,2 (Thr172, Thr183) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536764
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536764 349.00
Check your price
  • EpCAM Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529634
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529634 349.00
Check your price
  • ADCK4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522180
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
uncharacterized aarF domain-containing protein kinase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522180 349.00
Check your price
  • TMEM219 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547932
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Transmembrane protein 219
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547932 349.00
Check your price
  • LXR beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1333
  • Size 100µg
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1:200-1:2000)
Alternate names:
Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1333 349.00
Check your price
  • Galectin 9 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547503
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
HUAT, LGALS9A, ecalectin, gal-9, galectin 9, galectin-9, tumor antigen HOM-HD-21, urate transporter/channel protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547503 349.00
Check your price
  • CHI3L1 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547363
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cartilage glycoprotein 39; Breast regression protein 39; BRP39 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547363 349.00
Check your price
  • TRPC4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518987
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
TRPC4; transient receptor potential cation channel, subfamily C, member 4; HTRP4; MGC119570; MGC119571; MGC119572; MGC119573; TRP4; transient receptor potential 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518987 349.00
Check your price
  • RNaseH2A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520667
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (4 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Ribonuclease HI subunit A; Ribonuclease HI large subunit; RNase H(35); Aicardi-Goutieres syndrome 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520667 349.00
Check your price
  • GATA3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1028
  • Size 100ug
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (Paraffin) (1:200-1:2000), Immunoprecipitation (5ug), Western Blot (1:1000)
Alternate names:
GATA-binding factor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1028 349.00
Check your price
  • COPS2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522802
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
Alien protein; Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522802 349.00
Check your price
  • Mouse anti-Human IgG (Lambda light chain) Secondary Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA180937
  • Size 100Tests
Applications
Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180937 348.00
Check your price
  • CD56 Monoclonal Antibody, PE-Texas Red® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Texas Red®
  • Catalog # MHCD5617
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD5617 352.00
Check your price
  • CD3e Monoclonal Antibody, Alexa Fluor® 405 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 405
  • Catalog # CD0326
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD0326 352.00
Check your price
  • CD14 Monoclonal Antibody, Alexa Fluor® 700 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 700
  • Catalog # MHCD1429
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Myeloid cell-specific leucine-rich glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD1429 352.00
Check your price
  • CD11b Monoclonal Antibody, Alexa Fluor® 700 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 700
  • Catalog # CD11b29
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Neutrophil adherence receptor; Leukocyte adhesion receptor MO1; Cell surface glycoprotein MAC-1 subunit alpha; CR-3 alpha chain; CD11 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD11b29 353.00
Check your price
  • Phospho-VAV1 (Tyr160) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126651
  • Size 50µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
p95vav
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126651 349.00
Check your price
  • TBK1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120344
  • Size 50µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay-dependent), Western Blot (1-2 µg/ml)
Alternate names:
TANK-binding kinase 1; T2K; NF-kappa-B-activating kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120344 349.00
Check your price
  • PDE7A Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532975
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
TM22; HCP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532975 349.00
Check your price
  • Oxytocin Receptor Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532961
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (40 µg/ml)
Alternate names:
OT-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532961 349.00
Check your price
  • ASCT2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533028
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (15-20 µg/ml)
Alternate names:
Solute carrier family 1 member 5; Sodium-dependent neutral amino acid transporter type 2; RD114/simian type D retrovirus receptor; Baboon M7 virus receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533028 349.00
Check your price
  • TGM2 Monoclonal Antibody for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA512739
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Transglutaminase-2; Transglutaminase H; Transglutaminase C; Tissue transglutaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512739 349.00
Check your price
  • Tartrate Resistant Acid Phosphatase Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512387
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:20)
Alternate names:
Type 5 acid phosphatase; Tartrate-resistant acid ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512387 349.00
Check your price
  • Phospho-S6 (Ser240, Ser244) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532581
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532581 349.00
Check your price
  • MUC1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Armenian Hamster
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511202
  • Size 500µL
Applications
Flow Cytometry (1-2 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:10,000)
Alternate names:
Tumor-associated mucin; Tumor-associated epithelial membrane antigen; Polymorphic epithelial mucin; Peanut-reactive urinary mucin; PEMT; Krebs von den Lungen-6; H23AG; Episialin; Carcinoma-associated mucin; Cancer antigen 15-3; Breast carcinoma-associated antigen DF3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511202 349.00
Check your price
  • ErbB2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513105
  • Size 500µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513105 349.00
Check your price
  • CD45RA Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516341
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516341 349.00
Check your price
  • CD4 Monoclonal Antibody, Alexa Fluor® 405 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 405
  • Catalog # MHCD0426
  • Size 500µL
Applications
Flow Cytometry (5 ul per 1x10^6 cells)
Alternate names:
T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0426 352.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA516358
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10µg/mg protein lysate), Western Blot (2.5-5.0 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516358 349.00
Check your price
  • TNFR1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523706
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-6 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p60; p55; Tumor necrosis factor receptor type I; Tumor necrosis factor receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523706 349.00
Check your price