Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
7081 - 7110 of 48881 products displayed
  • STK33 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523081
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:20,000)
Alternate names:
serine/threonine-protein kinase 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523081 349.00
Check your price
  • STEP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517220
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Striatum-enriched protein-tyrosine phosphatase; Neural-specific protein-tyrosine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517220 349.00
Check your price
  • SSBP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530487
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Sequence-specific single-stranded-DNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530487 349.00
Check your price
  • SRF Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527307
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
MCM1, serum response factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527307 349.00
Check your price
  • SERTAD1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528460
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transcriptional regulator interacting with the PHD-bromodomain 1; CDK4-binding protein p34SEI1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528460 349.00
Check your price
  • SEMA5A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA530884
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Semaphorin-F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530884 349.00
Check your price
  • SCP1 Polyclonal Antibody for IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146254
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:750), Immunofluorescence (1:100-1:750), Immunohistochemistry (Frozen) (1:750), Immunohistochemistry (Paraffin) (1:750), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Cancer/testis antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146254 349.00
Check your price
  • RAB5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA3915
  • Size 100µL
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1:500-1:5000)
Alternate names:
RAB5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3915 349.00
Check your price
  • PTPN2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517249
  • Size 100µL
Applications
ELISA (Assay Dependent), Western Blot (1:2000)
Alternate names:
T-cell protein-tyrosine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517249 349.00
Check your price
  • PTP1B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528358
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein-tyrosine phosphatase 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528358 349.00
Check your price
  • PRG2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530130
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Proteoglycan 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530130 349.00
Check your price
  • PPP2R5A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528444
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
PP2A B subunit isoform R5-alpha; PP2A B subunit isoform PR61-alpha; PP2A B subunit isoform B56-alpha; PP2A B subunit isoform B'-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528444 349.00
Check your price
  • PLZF Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529213
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Zinc finger protein PLZF; Zinc finger protein 145; Promyelocytic leukemia zinc finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529213 349.00
Check your price
  • PLVAP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531402
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Plasmalemma vesicle protein 1; Fenestrated endothelial-linked structure protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531402 349.00
Check your price
  • PKN2 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515887
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Protein-kinase C-related kinase 2; Protein kinase C-like 2; PKN gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515887 349.00
Check your price
  • Phospho-PPAR gamma (Ser112) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536763
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536763 349.00
Check your price
  • Phospho-OLIG2 (Ser10, Ser13, Ser14) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA535405
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Protein kinase C-binding protein RACK17; Protein kinase C-binding protein 2; Class E basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535405 349.00
Check your price
  • Phospho-Caspase 9 (Tyr153) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536020
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536020 349.00
Check your price
  • Phospho-BRCA1 (Ser1423) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA536829
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536829 349.00
Check your price
  • PGP9.5 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA146205
  • Size 100µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:2000)
Alternate names:
Ubiquitin thioesterase L1; PGP 9.5; Neuron cytoplasmic protein 9.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146205 349.00
Check your price
  • PDK4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522983
  • Size 100µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Pyruvate dehydrogenase kinase isoform 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522983 349.00
Check your price
  • PABPN1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527433
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Polyadenylate-binding nuclear protein 1; Poly(A)-binding protein II; Nuclear poly(A)-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527433 349.00
Check your price
  • Oval Cell Marker Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516137
  • Size 100µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516137 349.00
Check your price
  • NR4A1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527274
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Orphan nuclear receptor HMR; Nuclear protein N10; Nuclear hormone receptor NUR/77; Testicular receptor 3; ST-59; Orphan nuclear receptor TR3; Early response protein NAK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527274 349.00
Check your price
  • NAMPT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523198
  • Size 100µL
Applications
Western Blot (1:1000-1:5000)
Alternate names:
Visfatin; Pre-B-cell colony-enhancing factor 1 homolog; Pre-B-cell colony-enhancing factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523198 349.00
Check your price
  • MUC2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521329
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Intestinal mucin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521329 349.00
Check your price
  • MTHFR Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515844
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
RP11-56N19.4, 5,10-methylenetetrahydrofolate reductase (NADPH); methylenetetrahydrofolate reductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515844 349.00
Check your price
  • mCherry Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534974
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:3000), Western Blot (1:1000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534974 349.00
Check your price
  • MAOA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527381
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Monoamine oxidase type A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527381 349.00
Check your price
  • LIN28A Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515453
  • Size 100µL
Applications
Immunocytochemistry (1/200-1/1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515453 349.00
Check your price