Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
7111 - 7140 of 48882 products displayed
  • Cytokeratin 1 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526699
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Type-II keratin Kb1; Keratin-1; Hair alpha protein; Cytokeratin-1; 67 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526699 349.00
Check your price
  • Connexin 43 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511632
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Gap junction 43 kDa heart protein; Connexin-43
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511632 349.00
Check your price
  • Chromogranin A Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535071
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Pituitary secretory protein I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535071 349.00
Check your price
  • beta Amyloid Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513327
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513327 349.00
Check your price
  • ATF4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513293
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Tax-responsive enhancer element-binding protein 67; DNA-binding protein TAXREB67; Cyclic AMP-responsive element-binding protein 2; Activating transcription factor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513293 349.00
Check your price
  • ARPC1A Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513448
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
SOP2-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513448 349.00
Check your price
  • Apolipoprotein F Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523616
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Lipid transfer inhibitor protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523616 349.00
Check your price
  • Apolipoprotein A4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514544
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
APOA4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514544 349.00
Check your price
  • Adiponectin Receptor 1 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535347
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Progestin and adipoQ receptor family member I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535347 349.00
Check your price
  • BrdU Monoclonal Antibody, Alexa Fluor® 647 conjugate for IF and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Alexa Fluor® 647
  • Catalog # B35133
  • Size 350µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
B35133 351.00
Check your price
  • MVP Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA124690
  • Size 25µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Western Blot (2-10 µg/ml)
Alternate names:
Lung resistance-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124690 349.00
Check your price
  • IFN beta Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124778
  • Size 25µg
Applications
ELISA (0.5-1 µg/ml), Neutralization (0.05-0.20 ug/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124778 349.00
Check your price
  • P-Glycoprotein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126529
  • Size 250µL
Applications
Immunocytochemistry (1:20), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
P-glycoprotein 1; ATP-binding cassette sub-family B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126529 349.00
Check your price
  • MRP2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126536
  • Size 250µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Multidrug resistance-associated protein 2; Canalicular multidrug resistance protein; ATP-binding cassette sub-family C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126536 349.00
Check your price
  • V-ATPase 60 kDa subunit Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # A6427
  • Size 250µg
Applications
Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A6427 351.00
Check your price
  • DPM1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # A6429
  • Size 250µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Mannose-P-dolichol synthase; Dolichyl-phosphate beta-D-mannosyltransferase; Dolichol-phosphate mannose synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A6429 351.00
Check your price
  • Carboxypeptidase Y Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # A6428
  • Size 250µg
Applications
Immunocytochemistry (20 µg/ml), Immunofluorescence (20 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Carboxypeptidase Y, pcy1, cpy1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A6428 351.00
Check your price
  • Lysozyme Monoclonal Antibody for IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182873
  • Size 200µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/10), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
1,4-beta-N-acetylmuramidase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182873 349.00
Check your price
  • Mouse anti-Human IgG (CH3) Secondary Antibody for Western Blot, ELISA and HA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516557
  • Size 200ug
Applications
ELISA (Assay Dependent), Hemagglutination Assay (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516557 349.00
Check your price
  • ITGB1 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA2910
  • Size 200ug
Applications
Flow Cytometry (0.5 ug/test)
Alternate names:
VLA-4 subunit beta; Glycoprotein IIa; Fibronectin receptor subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA2910 349.00
Check your price
  • Vaccinia Virus Polyclonal Antibody, HRP conjugate for IHC and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173192
  • Size 1mL
Applications
ELISA (1:200-1000), Immunohistochemistry (1:20-200), Western Blot (1:20-200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173192 349.00
Check your price
  • Staphylococcus epidermidis Monoclonal Antibody for IF, ELISA and AGG
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA135788
  • Size 1mL
Applications
Agglutination (Assay Dependent), ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
S. epidermidis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135788 349.00
Check your price
  • Mycobacterium tuberculosis Polyclonal Antibody, FITC conjugate for IHC
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # PA173134
  • Size 1mL
Applications
Immunohistochemistry (1:10-50)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173134 349.00
Check your price
  • IL-1 beta Polyclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # P420B
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
P420B 349.00
Check your price
  • ZO-1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528858
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528858 349.00
Check your price
  • ZNF140 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521519
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
pHZ-39
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521519 349.00
Check your price
  • ZAK Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527881
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Sterile alpha motif- and leucine zipper-containing kinase AZK; Mixed lineage kinase-related kinase; MLK-like mitogen-activated protein triple kinase; Leucine zipper- and sterile alpha motif-containing kinase; Human cervical cancer suppressor gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527881 349.00
Check your price
  • XBP1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515626
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515626 349.00
Check your price
  • Vimentin Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527231
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
VIM, FLJ36605
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527231 349.00
Check your price
  • Villin Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529078
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
D2S1471, VIL, villin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529078 349.00
Check your price