Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  40  50  60  70  80  90  100  110  120  200  300  400  500  600  700  800  900  1000  1611 
691 - 720 of 48310 products displayed
  • CD90 Antibody, FITC (Monoclonal, F15-42-1)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA135866
  • Size 100 µg
Applications
Flow Cytometry (Assay dependent.)
Alternate names:
Thy-1 antigen; CDw90
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135866 600.00
Check your price
  • CD46 Antibody (Monoclonal, M177)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140183
  • Size 100 µg
Applications
ELISA (Assay Dependent.), Flow Cytometry (Assay Dependent.), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
Trophoblast leukocyte common antigen; TLX
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140183 600.00
Check your price
  • CD200R Antibody, FITC (Monoclonal, OX-108)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA135932
  • Size 100 µg
Applications
Flow Cytometry (10ul x 10^6 cells or 100ul whole blood)
Alternate names:
Cell surface glycoprotein OX2 receptor 1; CD200 cell surface glycoprotein receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135932 600.00
Check your price
  • BPDE Antibody (Monoclonal, 8E11)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA140270
  • Size 100 µg
Applications
ELISA (Assay Dependent.)
Alternate names:
Benzo[a]pyrenediol-epoxide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140270 600.00
Check your price
  • BPDE Antibody (Monoclonal, 5D11)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA140269
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Benzo[a]pyrenediol-epoxide modified DNA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140269 600.00
Check your price
  • ASGR1 Antibody (Monoclonal, 8D7)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA140244
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent.), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
Hepatic lectin 1; Hepatic lectin H1; C-type lectin domain family 4 member H1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140244 600.00
Check your price
  • CD8 alpha Antibody, PE-Cyanine5 (Monoclonal, OX-8)
  • Host Mouse
  • Target Species Rat
  • Conjugate PE-Cyanine5
  • Catalog # MA517401
  • Size 100 ug
Applications
Flow Cytometry (1 ug /10^6 cells)
Alternate names:
OX-8 membrane antigen; CD8 antigen 32 kDa chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517401 600.00
Check your price
  • CD4 Antibody, FITC (Monoclonal, vpg34)
  • Host Mouse
  • Target Species Cat
  • Conjugate FITC
  • Catalog # MA516970
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516970 600.00
Check your price
  • CD4 Antibody, FITC (Monoclonal, MIL17)
  • Host Mouse
  • Target Species Pig
  • Conjugate FITC
  • Catalog # MA516854
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516854 600.00
Check your price
  • CD180 Antibody, FITC (Monoclonal, MHR73)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516836
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 64
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516836 600.00
Check your price
  • CD3/CD4/CD8 Antibody, PE, PE-Cyanine5 (Monoclonal, 7E14, 17D8, M2AB)
  • Host Mouse
  • Target Species Human
  • Conjugate PE , PE-Cyanine5
  • Catalog # MA112563
  • Size 100 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-lymphocyte differentiation antigen T8/Leu-2; T-cell surface antigen T4/Leu-3; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112563 600.00
Check your price
  • CD130 (soluble) Antibody, FITC (Monoclonal, B-R3)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA181159
  • Size 100 Tests
Applications
Flow Cytometry (Neat)
Alternate names:
Oncostatin-M receptor subunit alpha; Membrane glycoprotein 130; Interleukin-6 signal transducer; CDw130
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181159 600.00
Check your price
  • CD100 Antibody, PE (Monoclonal, A8)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA516559
  • Size 100 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516559 600.00
Check your price
  • Myoglobin Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175298
  • Size 1 mL
Applications
ELISA (Assay Dependent)
Alternate names:
Human myoglobin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175298 600.00
Check your price
  • MRP6 Antibody (Monoclonal, M6II-21)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135686
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunocytochemistry (1:20-1:50), Immunohistochemistry (Frozen) (1:20-1:50), Western Blot (1:20-1:50)
Alternate names:
Multi-specific organic anion transporter E; Anthracycline resistance-associated protein; ATP-binding cassette sub-family C member 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135686 600.00
Check your price
  • MRP4 Antibody (Monoclonal, M4I-80)
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA135682
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunocytochemistry (1:20-50), Immunohistochemistry (Frozen) (1:20), Western Blot (1:20-50)
Alternate names:
Multi-specific organic anion transporter B; MRP/cMOAT-related ABC transporter; ATP-binding cassette sub-family C member 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135682 600.00
Check your price
  • MRP4 Antibody (Monoclonal, M4I-10)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135681
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunocytochemistry (1:20), Immunohistochemistry (1:20), Western Blot (1:20)
Alternate names:
Multi-specific organic anion transporter B; MRP/cMOAT-related ABC transporter; ATP-binding cassette sub-family C member 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135681 600.00
Check your price
  • MRP2 Antibody (Monoclonal, M2III-5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135678
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunocytochemistry (1:20-1:50), Immunohistochemistry (Paraffin, Frozen) (1:20), Western Blot (1:20-1:50)
Alternate names:
Multidrug resistance-associated protein 2; Canalicular multidrug resistance protein; ATP-binding cassette sub-family C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135678 600.00
Check your price
  • Epstein Barr Virus LMP Antibody (Monoclonal, CS1/CS2/CS3/CS4)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA180022
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:100-1:200)
Alternate names:
EBV, EBV Latent membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180022 600.00
Check your price
  • EPCR Antibody (Monoclonal, LMR-42)
  • Host Rat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA135363
  • Size 1 mL
Applications
Flow Cytometry (1:20), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Endothelial cell protein C receptor; Activated protein C receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135363 600.00
Check your price
  • Cyclin D1 Antibody (Monoclonal, SP4)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA139546
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA139546 600.00
Check your price
  • CD63 Antibody (Monoclonal, CLB-180)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135272
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunofluorescence (Assay dependent.)
Alternate names:
Tetraspanin-30; Ocular melanoma-associated antigen; OMA81H; Melanoma-associated antigen ME491; Lysosomal-associated membrane protein 3; Granulophysin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135272 600.00
Check your price
  • Caspase 8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137242
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137242 600.00
Check your price
  • B Cell Antigen Antibody (Monoclonal, 68-IB3)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA135018
  • Size 1 mL
Applications
Flow Cytometry (1:10), Immunohistochemistry (Frozen) (1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135018 600.00
Check your price
  • Transferrin Antibody (Monoclonal, F2H8G6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIT0603
  • Size 1 mg
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Siderophilin; Beta-1 metal-binding globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIT0603 600.00
Check your price
  • Thyroid Stimulating Hormone Antibody (Monoclonal, P2D9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIT0412
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIT0412 600.00
Check your price
  • Thyroid Stimulating Hormone Antibody (Monoclonal, Fc1005)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIT0401
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIT0401 600.00
Check your price
  • Thyroid Stimulating Hormone Antibody (Monoclonal, Fc1002)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIT0409
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIT0409 600.00
Check your price
  • Phenytoin Antibody (Monoclonal, 1P1H2)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MIP0802
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Phenytek®, Dilantin®, Kapseals®, Infatabs®, Epanutin®
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIP0802 600.00
Check your price
  • Mouse anti-Human IgM
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MII0402
  • Size 1 mg
Applications
ELISA (1 µg/ml), Flow Cytometry (1 µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (0.5-2ug/ml)
Alternate names:
Immunoglobulin M, IgM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MII0402 600.00
Check your price