Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  140  150  160  170  180  190  200  210  220  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1605 
7351 - 7380 of 48135 products displayed
  • p53 Antibody (Monoclonal, SP5)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516387
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516387 349.00
Check your price
  • p53 Antibody (Monoclonal, PAb 240)
  • Host Mouse
  • Target Species Human, Rhesus monkey
  • Conjugate Unconjugated
  • Catalog # MA515244
  • Size 500 µL
Applications
ChIP assay (1-3 µg x 10^6 cells), Flow Cytometry (Assay Dependent), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515244 349.00
Check your price
  • p130 Antibody (Monoclonal, 130P215 (DCS-215))
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514305
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
pRb2; Retinoblastoma-related protein 2; PPAR-alpha-interacting complex protein 128; 130 kDa retinoblastoma-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514305 349.00
Check your price
  • Nrf2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532523
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Nuclear factor, erythroid derived 2, like 2; HEBP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532523 349.00
Check your price
  • NGAL Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532476
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
p25; Siderocalin LCN2; Oncogene 24p3; Lipocalin-2; 25 kDa alpha-2-microglobulin-related subunit of MMP-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532476 349.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Fruit fly, Hamster, Human, Mouse, Pig, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA516545
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (5-10 µg/mL)
Alternate names:
XRel1; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516545 349.00
Check your price
  • MVP Antibody (Monoclonal, 1032)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA513871
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Gel Shift (1mg/mL), Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Lung resistance-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513871 349.00
Check your price
  • MSH3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532508
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Mismatch repair protein 1; Divergent upstream protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532508 349.00
Check your price
  • MMP9 Antibody (Monoclonal, GE-213)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513595
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (2 µg/ml), Inhibition Assays (2-4 µg/ml)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513595 349.00
Check your price
  • MiTF Antibody (Monoclonal, D5)
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # MA514154
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunohistochemistry (Paraffin) (4-8 µg/mL)
Alternate names:
Class E basic helix-loop-helix protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514154 349.00
Check your price
  • mGluR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121035
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121035 349.00
Check your price
  • J Chain Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532457
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Joining chain of multimeric IgA and IgM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532457 349.00
Check your price
  • IGF1R alpha Antibody (Monoclonal, 24-57)
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513812
  • Size 500 µL
Applications
Immunoprecipitation (Assay Dependent), Inhibition Assays (1.5 µg/mL)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513812 349.00
Check your price
  • IGF1R alpha Antibody (Monoclonal, 17-69)
  • Host Mouse
  • Target Species Human, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA513807
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Inhibition Assays (Assay Dependent)
Alternate names:
Insulin-like growth factor I receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513807 349.00
Check your price
  • ID2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532449
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Inhibitor of differentiation 2; Inhibitor of DNA binding 2; Class B basic helix-loop-helix protein 26
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532449 349.00
Check your price
  • HPV Antibody (Monoclonal, K1H8)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA512446
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:20)
Alternate names:
HPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512446 349.00
Check your price
  • HDJ2 Antibody (Monoclonal, KA2A5.6)
  • Host Mouse
  • Target Species Bacteria, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512748
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Heat shock protein J2; DnaJ-like protein 1; Human DnaJ protein 2; Heat shock 40 kDa protein 4; HSDJ; DnaJ protein homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512748 349.00
Check your price
  • Glypican 3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532432
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
MXR7; Intestinal protein OCI-5; GTR2-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532432 349.00
Check your price
  • GLUT1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516793
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:200), Western Blot (2-4 ug/ ml)
Alternate names:
Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516793 349.00
Check your price
  • GLUT1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532428
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
HepG2 glucose transporter; Glucose transporter type 1, erythrocyte/brain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532428 349.00
Check your price
  • FAS Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532336
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:160)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532336 349.00
Check your price
  • Estrogen Receptor alpha Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516440
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:250-500), Immunoprecipitation (10 µg/ml), Western Blot (2-5µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516440 349.00
Check your price
  • ErbB4 Antibody (Monoclonal, H4.77.16 (Ab77))
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA512888
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (2µg/mg protein lysate), Western Blot (1:50-1:500)
Alternate names:
p180erbB4; Tyrosine kinase-type cell surface receptor HER4; Proto-oncogene-like protein c-ErbB-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512888 349.00
Check your price
  • ErbB3 Antibody (Monoclonal, 2B5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA513040
  • Size 500 µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3; Glial growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513040 349.00
Check your price
  • ErbB2 Antibody (Monoclonal, N28)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513032
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513032 349.00
Check your price
  • ErbB2 Antibody (Monoclonal, N24)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513003
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunoprecipitation (2 µg/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513003 349.00
Check your price
  • ErbB2 Antibody (Monoclonal, N12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512998
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:200), Immunomicroscopy (Assay Dependent)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512998 349.00
Check your price
  • ErbB2 Antibody (Cocktail, e2-4001 + 3B5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514057
  • Size 500 µL
Applications
Flow Cytometry (1:10-1:100), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (0.25-0.5 µg/mL), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514057 349.00
Check your price
  • EGFR Antibody (Monoclonal, H9B4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513877
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513877 349.00
Check your price
  • EGFR Antibody (Monoclonal, 199.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513319
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513319 349.00
Check your price