Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  270  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1630 
7531 - 7560 of 48882 products displayed
  • RAD51 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514419
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514419 349.00
Check your price
  • Prolactin Receptor Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511955
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
PRL-R, delta 4-SF1b truncated prolactin receptor, delta 4-delta 7/11 truncated prolactin receptor, hPRL receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511955 349.00
Check your price
  • PMEL Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513232
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:40-1:80), Western Blot (1:50)
Alternate names:
Silver locus protein homolog; Premelanosome protein; P100; P1; Melanoma-associated ME20 antigen; Melanocytes lineage-specific antigen GP100; Melanocyte protein Pmel 17; ME20-M
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513232 349.00
Check your price
  • Plasminogen Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513849
  • Size 500µL
Applications
Western Blot (1-2 µg/ml)
Alternate names:
plasmin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513849 349.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516392
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516392 349.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514544
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514544 349.00
Check your price
  • Perlecan Monoclonal Antibody for IF, IHC (F) and IHC (P)
  • Host Rat
  • Target Species Bovine, Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA514641
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1-2 µg/ml)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514641 349.00
Check your price
  • Pds1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512252
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-200)
Alternate names:
T3M-11-derived growth factor; Somatomedin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512252 349.00
Check your price
  • PAR1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532612
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
Thrombin receptor; Coagulation factor II receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532612 349.00
Check your price
  • Pan-cadherin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA516766
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:200), Western Blot (2-4 µg/ml)
Alternate names:
Neural cadherin; Epithelial cadherin; Uvomorulin; ARC-1; Liver cell adhesion molecule; Placental cadherin; Retinal cadherin; xTCAD-1; CAM 120/80; CDw325
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516766 349.00
Check your price
  • p70 S6 Kinase Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532533
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532533 349.00
Check your price
  • p57 Kip2 Monoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511309
  • Size 500µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
p57Kip2; Cyclin-dependent kinase inhibitor p57
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511309 349.00
Check your price
  • Osteopontin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA516821
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-100), Western Blot (2-4 µg/ml)
Alternate names:
Secreted phosphoprotein 1; Bone sialoprotein 1; Uropontin; Urinary stone protein; Nephropontin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516821 349.00
Check your price
  • Occludin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532524
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532524 349.00
Check your price
  • Nestin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532516
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532516 349.00
Check your price
  • Myogenin Monoclonal Antibody for Western Blot, IF, IHC (P), IP and GS
  • Host Mouse
  • Target Species Cat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511486
  • Size 500µL
Applications
Gel Shift (1mg/mL), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:400-1:800), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Myogenic factor 4; Class C basic helix-loop-helix protein 3; MYOD1-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511486 349.00
Check your price
  • MUC3 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512350
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
MUC-3A,intestinal mucin-3A, mucin 3, intestinal, mucin 3A, intestinal, mucin-3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512350 349.00
Check your price
  • MUC2 Monoclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512345
  • Size 500µL
Applications
Flow Cytometry (0.5-1ug/test), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:10-1:500)
Alternate names:
Intestinal mucin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512345 349.00
Check your price
  • MMP13 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514247
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Matrix metalloproteinase-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514247 349.00
Check your price
  • MGMT Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513506
  • Size 500µL
Applications
Flow Cytometry (2 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:10-20), Western Blot (0.5-1.0 µg/ml)
Alternate names:
O-6-methylguanine-DNA-alkyltransferase; 6-O-methylguanine-DNA methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513506 349.00
Check your price
  • mGluR1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121035
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121035 349.00
Check your price
  • Melan-A Monoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515237
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (1:100-1:200), Immunoprecipitation (1:50), Western Blot (1:50-1:100)
Alternate names:
Protein Melan-A; Antigen SK29-AA; Antigen LB39-AA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515237 349.00
Check your price
  • Melan-A Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514017
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Protein Melan-A; Antigen SK29-AA; Antigen LB39-AA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514017 349.00
Check your price
  • LMO2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532477
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
T-cell translocation protein 2; LIM domain only protein 2; Cysteine-rich protein TTG-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532477 349.00
Check your price
  • L-Plastin Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511921
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 ug/ml), Western Blot (1-2 µg/ml)
Alternate names:
Lymphocyte cytosolic protein 1; LC64P; L-plastin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511921 349.00
Check your price
  • IRS1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532455
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
HIRS-1, IRS-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532455 349.00
Check your price
  • INSR alpha Monoclonal Antibody for IA
  • Host Mouse
  • Target Species Bovine, Human, Sheep, Pig
  • Conjugate Unconjugated
  • Catalog # MA513773
  • Size 500µL
Applications
Inhibition Assays (10nM)
Alternate names:
CD220, HHF5, IR, IR alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513773 349.00
Check your price
  • iNOS Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516524
  • Size 500µL
Applications
Immunohistochemistry (Assay Dependent)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516524 349.00
Check your price
  • HPV Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA512446
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:20)
Alternate names:
HPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512446 349.00
Check your price
  • GRP94 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rat
  • Target Species Bovine, Dog, Chicken, Horse, Guinea pig, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA514592
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (10 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
gp96 homolog; Heat shock protein 90 kDa beta member 1; 98 kDa protein kinase; 94 kDa glucose-regulated protein; Tumor rejection antigen 1; Tumor rejection antigen gp96; Polymorphic tumor rejection antigen 1; Endoplasmic reticulum resident protein 99; Transferrin-binding protein; Heat shock 108 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514592 349.00
Check your price