Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  280  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1611 
7891 - 7920 of 48310 products displayed
  • CCRL2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528743
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Lipopolysaccharide-inducible C-C chemokine receptor; G-protein coupled beta chemokine receptor; Chemokine receptor CCR11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528743 349.00
Check your price
  • CCKBR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527169
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cholecystokinin-2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527169 349.00
Check your price
  • CBS Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522273
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
HIP4, beta-thionase, cystathionine beta-synthase, methylcysteine synthase, serine sulfhydrase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522273 349.00
Check your price
  • Cathepsin D Antibody (Monoclonal, CTD-19)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126773
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000)
Alternate names:
CLN10, CPSD, MGC2311
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126773 349.00
Check your price
  • CAS Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521468
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Importin-alpha re-exporter; Chromosome segregation 1-like protein; Cellular apoptosis susceptibility protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521468 349.00
Check your price
  • CARD10 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529261
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (Assay-dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Bcl10-interacting MAGUK protein 1; CARD-containing MAGUK protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529261 349.00
Check your price
  • Carboxypeptidase M Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA521727
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
renal carboxypeptidase, urinary carboxypeptidase B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521727 349.00
Check your price
  • Cannabinoid Receptor 1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1743
  • Size 100 µL
Applications
Immunocytochemistry (1:1,000), Immunofluorescence (1:1000), Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Western Blot (1:250)
Alternate names:
Brain-type cannabinoid receptor; CANN6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1743 349.00
Check your price
  • Calpastatin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534692
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Sperm BS-17 component; Calpain inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534692 349.00
Check your price
  • Calpastatin Antibody (Monoclonal, 1F7E3D10)
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3944
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunohistochemistry (1:20), Western Blot (1:5,000)
Alternate names:
Sperm BS-17 component; Calpain inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3944 349.00
Check your price
  • C1S Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528253
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Complement component 1 subcomponent s; C1 esterase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528253 349.00
Check your price
  • C/EBP beta Antibody (Monoclonal, 3H9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515893
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Western Blot (1/500-1/2000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515893 349.00
Check your price
  • c-Cbl Antibody (Monoclonal, 3B12)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515885
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Signal transduction protein CBL; RING finger protein 55; Proto-oncogene c-Cbl; Casitas B-lineage lymphoma proto-oncogene
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515885 349.00
Check your price
  • BRD1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530491
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Bromodomain and PHD finger-containing protein 2; BR140-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530491 349.00
Check your price
  • BMPR2 Antibody (Monoclonal, 1F12)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515826
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Bone morphogenetic protein receptor type II; BRK-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515826 349.00
Check your price
  • BMP-4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527288
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Bone morphogenetic protein 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527288 349.00
Check your price
  • BLK Antibody (Monoclonal, 1E6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515725
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
p55-Blk; B lymphocyte kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515725 349.00
Check your price
  • beta Tubulin 2 Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA527552
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Tubulin beta class IIa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527552 349.00
Check your price
  • BAIAP2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530386
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Insulin receptor substrate protein of 53 kDa; Insulin receptor substrate p53/p58; Fas ligand-associated factor 3; Insulin receptor tyrosine kinase 53 kDa substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530386 349.00
Check your price
  • Axl Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528850
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
AXL oncogene
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528850 349.00
Check your price
  • Aurora A Antibody (Monoclonal, 4G10)
  • Host Mouse
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515804
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase 15; Breast tumor-amplified kinase; Aurora/IPL1-related kinase 1; Aurora 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515804 349.00
Check your price
  • ATRX Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521348
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
X-linked nuclear protein; Heterochromatin protein 2; HP1-BP38 protein; HP1 alpha-interacting protein; ATP-dependent helicase ATRX; Znf-HX; X-linked helicase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521348 349.00
Check your price
  • ATP Synthase gamma Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529975
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
F-ATPase gamma subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529975 349.00
Check your price
  • ATP Citrate Lyase Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529497
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Citrate cleavage enzyme; ATP-citrate (pro-S-)-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529497 349.00
Check your price
  • ATM Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515816
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Ataxia telangiectasia mutated
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515816 349.00
Check your price
  • ATF4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527576
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Tax-responsive enhancer element-binding protein 67; DNA-binding protein TAXREB67; Cyclic AMP-responsive element-binding protein 2; Activating transcription factor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527576 349.00
Check your price
  • ATF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527129
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000)
Alternate names:
cAMP response element-binding protein CRE-BP1; Histone acetyltransferase ATF2; HB16; Cyclic AMP-responsive element-binding protein 2; Activating transcription factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527129 349.00
Check your price
  • Ataxin 3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527391
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Spinocerebellar ataxia type 3 protein; Machado-Joseph disease protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527391 349.00
Check your price
  • ASL Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522052
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Arginosuccinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522052 349.00
Check your price
  • Arginase 2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531088
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Type II arginase; Non-hepatic arginase; Kidney-type arginase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531088 349.00
Check your price