Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  287  288  289  290  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1622 
8251 - 8280 of 48655 products displayed
  • Relaxin 1 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547858
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
H1, RLN1, RLXH1, bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, RLXH1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547858 349.00
Check your price
  • REG3B Polyclonal Antibody for Western Blot and IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547700
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Regenerating islet-derived protein III-beta; Pancreatitis-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547700 349.00
Check your price
  • RAP1A Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1013
  • Size 100 µg
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (2 µg), Western Blot (1:500-1:1000)
Alternate names:
Ras-related protein Krev-1; GTP-binding protein smg p21A; G-22K; C21KG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1013 349.00
Check your price
  • Rabies Virus Monoclonal Antibody for IF, ICC, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA121549
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121549 349.00
Check your price
  • RAB3d Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1773
  • Size 100 µg
Applications
Western Blot (2.0 µg/ml)
Alternate names:
Ras-Associated Protein 3D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1773 349.00
Check your price
  • PUMA Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520007
  • Size 100 µg
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (2 µg/ml)
Alternate names:
p53 up-regulated modulator of apoptosis; JFY-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520007 349.00
Check your price
  • PTGER2/PTGER3 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112677
  • Size 100 µg
Applications
Western Blot (1-5 µg/ml)
Alternate names:
Prostanoid EP2 receptor; Prostanoid EP3 receptor; PGE2-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112677 349.00
Check your price
  • Protogenin Polyclonal Antibody for IHC (F)
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547874
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml)
Alternate names:
Protein Shen-Dan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547874 349.00
Check your price
  • Proliferin Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547169
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Proliferin-1; Mitogen-regulated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547169 349.00
Check your price
  • PPP1R8 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547906
  • Size 100 µg
Applications
Western Blot (0.5 µg/ml)
Alternate names:
Protein phosphatase 1 regulatory inhibitor subunit 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547906 349.00
Check your price
  • POMC Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518368
  • Size 100 µg
Applications
Immunohistochemistry (4-6 µg/ml), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518368 349.00
Check your price
  • PLXNC1 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547549
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Virus-encoded semaphorin protein receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547549 349.00
Check your price
  • Plasma Kallikrein Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547334
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Plasma prekallikrein; Kininogenin; Fletcher factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547334 349.00
Check your price
  • Phospho-PSD95 (Thr19) Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519478
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:300), Immunoprecipitation (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519478 349.00
Check your price
  • Phospho-IRS1 (Tyr1179) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11060
  • Size 100 µg
Applications
Western Blot (Assay dependent)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11060 349.00
Check your price
  • Phospho-IRS1 (Ser636) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11055
  • Size 100 µg
Applications
Western Blot (2 ug/ml)
Alternate names:
Phospho-Insulin Receptor Substrate-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11055 349.00
Check your price
  • Phospho-HSP27 (Ser15) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1016
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (3 µg), Western Blot (1:500-1:2000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1016 349.00
Check your price
  • PD-L1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518337
  • Size 100 µg
Applications
Immunohistochemistry (4-6µg/ml), Western Blot (0.3-1.0 ug/ml)
Alternate names:
B7 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518337 349.00
Check your price
  • Parvalbumin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547693
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1-50 µg/ml)
Alternate names:
parvalbumin, HGNC:9704, PVALB, D22S749, parvalbumin alpha, MGC116759
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547693 349.00
Check your price
  • p300 Polyclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1848
  • Size 100 µg
Applications
ChIP assay (1-3 ul), Western Blot (1-2 µg/ml)
Alternate names:
E1A-associated protein p300
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1848 349.00
Check your price
  • OLIG1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547313
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Class E basic helix-loop-helix protein 21; Class B basic helix-loop-helix protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547313 349.00
Check your price
  • NOX2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA534600
  • Size 100 µg
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:2000), Western Blot (1 µg/ml)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534600 349.00
Check your price
  • Nogo-A Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520366
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Reticulon-5; RTN-x; Neuroendocrine-specific protein C homolog; Neuroendocrine-specific protein; Neurite outgrowth inhibitor; Foocen; Glut4 vesicle 20 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520366 349.00
Check your price
  • Ninjurin 2 Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547692
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Nerve injury-induced protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547692 349.00
Check your price
  • NELL2 Polyclonal Antibody for IHC (F)
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547868
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml)
Alternate names:
NEL-like protein 2; MEL91 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547868 349.00
Check your price
  • NEFM Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519783
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:2000), Western Blot (1 µg/ml)
Alternate names:
Neurofilament triplet M protein; Neurofilament 3; 160 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519783 349.00
Check your price
  • MyD88 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519918
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519918 349.00
Check your price
  • MUC5AC Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135708
  • Size 100 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Paraffin, Frozen) (Assay dependent.), Western Blot (Assay dependent.)
Alternate names:
Tracheobronchial mucin; Mucin-5 subtype AC, tracheobronchial; Major airway glycoprotein; Lewis B blood group antigen; Gastric mucin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135708 349.00
Check your price
  • Mouse anti-Rat Kappa Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # A15426
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Ig kappa chain, Igk, Ig-kappa
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15426 349.00
Check your price
  • MIP-3 beta Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547958
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small inducible cytokine A19, Macrophage inflammatory protein 3 beta, MIP-3-beta, EBI1-ligand chemokine, ELC, Beta chemokine exodus-3, CK beta-11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547958 349.00
Check your price