Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  180  190  200  210  220  230  240  250  260  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  287  288  289  290  291  292  293  294  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1605 
8371 - 8400 of 48136 products displayed
  • NOX2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519010
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.03-0.1 µg/ml)
Alternate names:
p22 phagocyte B-cytochrome; gp91-phox; gp91-1; Superoxide-generating NADPH oxidase heavy chain subunit; Neutrophil cytochrome b 91 kDa polypeptide; NADPH oxidase 2; Heme-binding membrane glycoprotein gp91phox; Cytochrome b(558) subunit beta; CGD91-phox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519010 349.00
Check your price
  • Neuropilin 2 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547762
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Vascular endothelial cell growth factor 165 receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547762 349.00
Check your price
  • Nectin 3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547441
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Poliovirus receptor-related protein 3; CDw113
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547441 349.00
Check your price
  • n-Myc Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519529
  • Size 100 µg
Applications
Western Blot (1-5 µg/ml)
Alternate names:
Class E basic helix-loop-helix protein 37
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519529 349.00
Check your price
  • Myoglobin Antibody (Monoclonal, 4E2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122740
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
MGC13548, PVALB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122740 349.00
Check your price
  • MyD88 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519918
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519918 349.00
Check your price
  • Mycobacterium tuberculosis ESAT-6 Antibody
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA119446
  • Size 100 µg
Applications
Western Blot (0.2-1.0 µg/ml)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA119446 349.00
Check your price
  • MUC1 Antibody (Monoclonal, GP1.4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106503
  • Size 100 µg
Applications
Flow Cytometry (1:25-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Tumor-associated mucin; Tumor-associated epithelial membrane antigen; Polymorphic epithelial mucin; Peanut-reactive urinary mucin; PEMT; Krebs von den Lungen-6; H23AG; Episialin; Carcinoma-associated mucin; Cancer antigen 15-3; Breast carcinoma-associated antigen DF3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106503 349.00
Check your price
  • Mouse anti-Rat Kappa Light Chain, FITC
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # A15426
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Ig kappa chain, Igk, Ig-kappa
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15426 349.00
Check your price
  • Mist1 Antibody (Monoclonal, 6E8/A12/C11P1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1517
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Western Blot (1:100-1:500)
Alternate names:
Muscle, intestine and stomach expression 1; Class B basic helix-loop-helix protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1517 349.00
Check your price
  • MDMX Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519473
  • Size 100 µg
Applications
Immunofluorescence (5 µg/ml), Western Blot (5 µg/ml)
Alternate names:
p53-binding protein Mdm4; Protein Mdmx; Mdm2-like p53-binding protein; Double minute 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519473 349.00
Check your price
  • MC4R Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520759
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
MC4-R; melanocortin receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520759 349.00
Check your price
  • Marburg Virus Antibody (Monoclonal, A93G)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17392
  • Size 100 µg
Applications
ELISA (1:20), Western Blot (1:10)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17392 349.00
Check your price
  • Macrophage Marker Antibody (Monoclonal, LN-5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135492
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin, Frozen) (Assay dependent.)
Alternate names:
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135492 349.00
Check your price
  • LRP1B Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547836
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Low-density lipoprotein receptor-related protein-deleted in tumor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547836 349.00
Check your price
  • LIN28A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1096
  • Size 100 µg
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (5 µg, 1:1000), Western Blot (1:1000)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1096 349.00
Check your price
  • LIF Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547337
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (0.04-0.08 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Melanoma-derived LPL inhibitor; Differentiation-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547337 349.00
Check your price
  • LHR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521271
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Luteinizing hormone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521271 349.00
Check your price
  • LGMN Antibody
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547244
  • Size 100 µg
Applications
Western Blot (0.2 µg/ml)
Alternate names:
Protease, cysteine 1; Asparaginyl endopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547244 349.00
Check your price
  • LCK Antibody, Alexa Fluor® 647 (Monoclonal, LCK-01)
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518166
  • Size 100 µg
Applications
Flow Cytometry (Assay-dependent)
Alternate names:
p56-LCK; T cell-specific protein-tyrosine kinase; Proto-oncogene Lck; Protein YT16; Lymphocyte cell-specific protein-tyrosine kinase; Leukocyte C-terminal Src kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518166 349.00
Check your price
  • LAX1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547652
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Membrane-associated adapter protein LAX; Linker for activation of X cells
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547652 349.00
Check your price
  • L-Thyroxine T4 Antibody (Monoclonal, T4YCH)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA514716
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514716 349.00
Check your price
  • KLRF1 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547219
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
C-type lectin domain family 5 member C; Activating coreceptor NKp80
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547219 349.00
Check your price
  • KCNMA1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1923
  • Size 100 µg
Applications
Immunocytochemistry (10.0 µg/ml), Western Blot (2.0 µg/ml)
Alternate names:
Slowpoke homolog; Slo1; Slo-alpha; Maxi K channel; KCa1.1; K(VCA)alpha; Calcium-activated potassium channel, subfamily M subunit alpha-1; BKCA alpha; BK channel
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1923 349.00
Check your price
  • KCNA1 Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519593
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin, Frozen) (1:3000 dilution), Western Blot (1 µg/ml)
Alternate names:
Voltage-gated potassium channel subunit Kv1.1; MKI; MBK1; Voltage-gated potassium channel HBK1; Voltage-gated K(+) channel HuKI; RCK1; RBKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519593 349.00
Check your price
  • ITGA8 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547572
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ITGA8, integrin alpha-8, integrin, alpha 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547572 349.00
Check your price
  • ITGA3 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547398
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547398 349.00
Check your price
  • IRF7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520281
  • Size 100 µg
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
IRF-7H, IRF7A; IRF-7; interferon regulatory factor-7H
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520281 349.00
Check your price
  • IRF3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520086
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520086 349.00
Check your price
  • IL5RA Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547750
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
CD125, CDw125, HSIL5R3, IL5R, CD125 antigen, IL-5 receptor subunit alpha, IL-5R subunit alpha, interleukin 5 receptor type 3, interleukin-5 receptor alpha chain, interleukin-5 receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547750 349.00
Check your price