Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  70  80  90  100  110  120  130  140  200  300  400  500  600  700  800  900  1000  1630 
1441 - 1470 of 48882 products displayed
  • MERTK Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131203
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (1:200), Western Blot (1:500)
Alternate names:
Receptor tyrosine kinase MerTK; Proto-oncogene c-Mer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131203 499.00
Check your price
  • MECP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130643
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
AUTSX3, DKFZp686A24160, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130643 499.00
Check your price
  • MCP-1 Polyclonal Antibody for Western Blot, ELISA and FN
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA184244
  • Size 100µg
Applications
ELISA (0.5-2 ug/ml), Functional Assay (4.0-6 ug/ml), Western Blot (0.1-0.2 ug/ml)
Alternate names:
Small-inducible cytokine A2; Monocyte chemotactic protein 1; Monocyte chemoattractant protein 1; Immediate-early serum-responsive protein JE
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184244 499.00
Check your price
  • LIPG Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA132483
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-20 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
Endothelial-derived lipase; Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132483 499.00
Check your price
  • LDB2 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131764
  • Size 100µg
Applications
ELISA (1:5000-1:20,000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
LIM domain-binding factor CLIM1; Carboxyl-terminal LIM domain-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131764 499.00
Check your price
  • LDB1 Polyclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131715
  • Size 100µg
Applications
ChIP assay (20 µg for 10^8 cells), Western Blot (Assay Dependent)
Alternate names:
Nuclear LIM interactor; LIM domain-binding factor CLIM2; Carboxyl-terminal LIM domain-binding protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131715 499.00
Check your price
  • Lactoferrin Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131254
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay dependent), Immunoprecipitation (1:200), Western Blot (1:500)
Alternate names:
Talalactoferrin; Growth-inhibiting protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131254 499.00
Check your price
  • IL-17F Polyclonal Antibody for Western Blot, ELISA and FN
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186898
  • Size 100µg
Applications
ELISA (0.5-2 ug/ml), Functional Assay (0.9-1.2 ug/ml), Western Blot (0.1-0.2 ug/ml)
Alternate names:
Cytokine ML-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186898 499.00
Check your price
  • GST Tag Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # MA181040
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (1/100-1/500)
Alternate names:
Glutathione-S-Transferase, GST protein, GST Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181040 499.00
Check your price
  • Giardia lamblia cysts Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA191188
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
G. lamblia
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191188 499.00
Check your price
  • Giardia lamblia cysts Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Fungi
  • Conjugate Unconjugated
  • Catalog # MA191200
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50)
Alternate names:
G. lamblia
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191200 499.00
Check your price
  • gamma Adaptin Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131775
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Golgi adaptor HA1/AP1 adaptin subunit gamma-1; Gamma1-adaptin; Clathrin assembly protein complex 1 gamma-1 large chain; Adaptor-related protein complex 1 subunit gamma-1; Adaptor protein complex AP-1 subunit gamma-1; Gamma-adaptin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131775 499.00
Check your price
  • FGFR4 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131991
  • Size 100µg
Applications
ELISA (1:900-1:1100), Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:500)
Alternate names:
Fibroblast Growth Factor Receptor 4, TKF, JTK2, MGC20292, Fibroblast growth factor receptor 4, FGF Receptor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131991 499.00
Check your price
  • Estrogen Receptor alpha Monoclonal Antibody for Western Blot, IP, ChIP, GS and Neu
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA112692
  • Size 100µg
Applications
Blocking Assay (Assay Dependent), ChIP assay (Assay Dependent), Gel Shift (Assay Dependent), Immunoprecipitation (2 µg/ml protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112692 499.00
Check your price
  • ErbB3 Polyclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131989
  • Size 100µg
Applications
ELISA (1:900-1:1100), Flow Cytometry (Assay-dependent), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:100-1:500)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3; Glial growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131989 499.00
Check your price
  • EphB2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130297
  • Size 100µg
Applications
ELISA (1:1000), Immunocytochemistry (Assay-dependent), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10), Western Blot (1:100)
Alternate names:
Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase TYRO5; Renal carcinoma antigen NY-REN-47; EPH-like kinase 5; EPH tyrosine kinase 3; ELK-related tyrosine kinase; Developmentally-regulated Eph-related tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130297 499.00
Check your price
  • EDG2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112306
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Lysophosphatidic acid receptor Edg-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112306 499.00
Check your price
  • Dengue Virus Type 2 Monoclonal Antibody for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA183114
  • Size 100µg
Applications
Immunofluorescence (1:100-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183114 499.00
Check your price
  • CNTF Polyclonal Antibody for Western Blot, IHC (P), ELISA and FN
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA184969
  • Size 100µg
Applications
ELISA (0.5-2 µg/ml), Functional Assay (7-11 ug/ml), Immunohistochemistry (Paraffin) (0.25 ug/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
HCNTF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184969 499.00
Check your price
  • CD51 Monoclonal Antibody for IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191669
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (2-3µg/ml), Immunohistochemistry (2-3µg/ml), Immunoprecipitation (2-3 µg/ml)
Alternate names:
Vitronectin receptor subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191669 499.00
Check your price
  • CD45 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA180304
  • Size 100µg
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180304 499.00
Check your price
  • CD4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Goat, Sheep
  • Conjugate FITC
  • Catalog # MA180901
  • Size 100µg
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180901 499.00
Check your price
  • CD34 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA181639
  • Size 100µg
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181639 499.00
Check your price
  • CD31 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Goat, Sheep
  • Conjugate FITC
  • Catalog # MA180360
  • Size 100µg
Applications
Flow Cytometry (Neat)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180360 499.00
Check your price
  • Caspase 13 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA130684
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130684 499.00
Check your price
  • CAIII (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75525
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Carbonic Anhydrase III
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75525 499.00
Check your price
  • BMP-8B Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131216
  • Size 100µg
Applications
ELISA (1:2000), Western Blot (1:500-1:1000)
Alternate names:
Osteogenic protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131216 499.00
Check your price
  • BMP-2 Polyclonal Antibody for Western Blot, IHC (P), ELISA and DB
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131215
  • Size 100µg
Applications
Dot blot (1:2000), ELISA (1:2000), Western Blot (1:500-1:1000)
Alternate names:
Bone morphogenetic protein 2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131215 499.00
Check your price
  • ATAD5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131987
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Chromosome fragility-associated gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131987 499.00
Check your price
  • Artemis Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131797
  • Size 100µg
Applications
ELISA (1:10,000-1:20,000), Immunohistochemistry (Paraffin) (1:1000), Western Blot (1:500-1:2000)
Alternate names:
SNM1-like protein; SNM1 homolog C; Protein A-SCID; DNA cross-link repair 1C protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131797 499.00
Check your price