Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  74  75  76  80  90  100  110  120  130  140  150  160  200  300  400  500  600  700  800  900  1000  1630 
1831 - 1860 of 48882 products displayed
  • AKAP3 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131348
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Sperm oocyte-binding protein; Protein kinase A-anchoring protein 3; Fibrousheathin-1; Fibrousheathin I; Fibrous sheath protein of 95 kDa; Cancer/testis antigen 82; A-kinase anchor protein 110 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131348 475.00
Check your price
  • Adiponectin Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129901
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Gelatin-binding protein; Adipose most abundant gene transcript 1 protein; Adipocyte, C1q and collagen domain-containing protein; Adipocyte complement-related 30 kDa protein; 30 kDa adipocyte complement-related protein; Adipocyte-specific protein AdipoQ
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129901 475.00
Check your price
  • Adenylate Cyclase 4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131192
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Adenylyl cyclase 4; Adenylate cyclase type IV; ATP pyrophosphate-lyase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131192 475.00
Check your price
  • ADAM17 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131481
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:2000)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131481 475.00
Check your price
  • XRCC1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112640
  • Size 100ul
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (4 µg/ml)
Alternate names:
X-ray repair cross-complementing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112640 475.00
Check your price
  • OX40 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Cat, Human
  • Conjugate FITC
  • Catalog # MA516547
  • Size 100ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
TAX transcriptionally-activated glycoprotein 1 receptor; OX40L receptor; ACT35 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516547 475.00
Check your price
  • HLA-DR Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Goat, Human, Sheep
  • Conjugate FITC
  • Catalog # MA516502
  • Size 100ug
Applications
Flow Cytometry (1:5-1:10)
Alternate names:
MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516502 475.00
Check your price
  • HEC1 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126543
  • Size 100ug
Applications
ELISA (Assay-dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Retinoblastoma-associated protein HEC; Kinetochore-associated protein 2; Kinetochore protein Hec1; Highly expressed in cancer protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126543 475.00
Check your price
  • F4/80 Monoclonal Antibody, FITC conjugate for IF and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA516627
  • Size 100ug
Applications
Flow Cytometry (Neat), Immunofluorescence (Assay Dependent)
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516627 475.00
Check your price
  • F4/80 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA516626
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516626 475.00
Check your price
  • CD8 alpha Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Avian, Chicken
  • Conjugate FITC
  • Catalog # MA516441
  • Size 100ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
CD8, Leu-2, MAL, p32, CD8a
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516441 475.00
Check your price
  • CD68 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA516675
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
GP110, LAMP4, SCARD1, CD68 antigen, macrophage antigen CD68, macrosialin, scavenger receptor class D member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516675 475.00
Check your price
  • CD5 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA181217
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181217 475.00
Check your price
  • CD5 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Cat
  • Conjugate FITC
  • Catalog # MA516681
  • Size 100ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516681 475.00
Check your price
  • CD3e Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Many
  • Conjugate FITC
  • Catalog # MA516623
  • Size 100ug
Applications
Flow Cytometry (1:5-1:10)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516623 475.00
Check your price
  • CD31 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Biotin
  • Catalog # MA516770
  • Size 100ug
Applications
Flow Cytometry (Neat-1:200)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516770 475.00
Check your price
  • CD204 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Fish, Mouse, Pig
  • Conjugate Biotin
  • Catalog # MA516493
  • Size 100ug
Applications
Flow Cytometry (Neat)
Alternate names:
Scavenger receptor type A; Macrophage acetylated LDL receptor I and II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516493 475.00
Check your price
  • CD200 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516897
  • Size 100ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
My033, MOX1, MOX2, MRC, OX-2, CD200 antigen, MRC OX-2 antigen, OX-2 membrane glycoprotein, antigen identified by monoclonal antibody MRC OX-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516897 475.00
Check your price
  • B7-H4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516845
  • Size 100ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
T-cell costimulatory molecule B7x; Protein B7S1; Immune costimulatory protein B7-H4; B7h.5; B7 homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516845 475.00
Check your price
  • MHC II Monomorphic Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA181228
  • Size 100Tests
Applications
Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181228 475.00
Check your price
  • KIR2DL3 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA181620
  • Size 100Tests
Applications
Flow Cytometry (Neat)
Alternate names:
p58.2 MHC class-I-specific NK receptor; p58 natural killer cell receptor clone CL-6; Natural killer-associated transcript 2; NKAT2b; NKAT2a; MHC class I NK cell receptor; Killer inhibitory receptor cl 2-3; KIR-023GB; CD158 antigen-like family member B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181620 475.00
Check your price
  • F4/80 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA516625
  • Size 100TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
EMR1 hormone receptor; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-like module receptor 1; Cell surface glycoprotein F4/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516625 475.00
Check your price
  • CD25 Monoclonal Antibody, PE-Cyanine5 conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE-Cyanine5
  • Catalog # MA516588
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
p55; TAC antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516588 475.00
Check your price
  • Rat anti-Mouse IgG (Kappa light chain) Secondary Antibody, HRP conjugate for IHC (F) and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # MA134732
  • Size 250µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134732 472.00
Check your price
  • Luteinizing Hormone Releasing Hormone Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PEP10933
  • Size 10mg
Applications
Control (Assay Dependent)
Alternate names:
LHRH, GnRH, Lutropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP10933 473.00
Check your price
  • Neuron Specific Enolase (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75669
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Neuronal Specific Enolase, NSE
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75669 473.00
Check your price
  • GST Omega 1 140D (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75567
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75567 473.00
Check your price
  • GST Omega 1 140A (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75564
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75564 473.00
Check your price
  • Mouse anti-Human IgG1 Secondary Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134581
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134581 472.00
Check your price
  • Granzyme B Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # GRB04
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell serine protease 1-3E; SECT; Human lymphocyte protein; Granzyme-2; Fragmentin-2; Cytotoxic T-lymphocyte proteinase 2; Cathepsin G-like 1; CTLA-1; C11
Catalog # Size Price (AUD) Your Price (AUD) Qty
GRB04 470.00
Check your price