Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  74  75  76  77  78  79  80  81  82  90  100  110  120  130  140  150  160  200  300  400  500  600  700  800  900  1000  1630 
2011 - 2040 of 48881 products displayed
  • CD26 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134137
  • Size 200µg
Applications
Flow Cytometry (1:50-1:100 (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
TP103; T-cell activation antigen CD26; Dipeptidyl peptidase IV; Adenosine deaminase complexing protein 2; ADABP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134137 449.00
Check your price
  • CD146 Monoclonal Antibody for IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134111
  • Size 200µg
Applications
ELISA (Assay dependent), Flow Cytometry (1:10-1:50), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
S-endo 1 endothelial-associated antigen; Melanoma-associated antigen MUC18; Melanoma-associated antigen A32; Melanoma cell adhesion molecule; Cell surface glycoprotein P1H12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134111 449.00
Check your price
  • CD11b Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA190974
  • Size 200µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Neutrophil adherence receptor; Leukocyte adhesion receptor MO1; Cell surface glycoprotein MAC-1 subunit alpha; CR-3 alpha chain; CD11 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190974 449.00
Check your price
  • Prolactin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA139446
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
PRL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA139446 449.00
Check your price
  • Poliovirus Type 1-3 Polyclonal Antibody, Biotin conjugate for IF and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA533245
  • Size 1mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
PV-1, PV-2, PV-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533245 449.00
Check your price
  • Nrf2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138312
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Nuclear factor, erythroid derived 2, like 2; HEBP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138312 449.00
Check your price
  • mGluR5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138132
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
glutamate receptor, metabotropic 5, GRM5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138132 449.00
Check your price
  • Lactoferrin Polyclonal Antibody, HRP conjugate for IHC (F), IHC (P) and ELISA
  • Host Sheep
  • Target Species Dog, Human
  • Conjugate HRP
  • Catalog # PA186864
  • Size 1mL
Applications
ELISA (1/50), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
Talalactoferrin; Growth-inhibiting protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186864 449.00
Check your price
  • JM4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137961
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
PRAF2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137961 449.00
Check your price
  • ITGB5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137932
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
FLJ26658
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137932 449.00
Check your price
  • ENO1 Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130493
  • Size 1mL
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:25-1:100), Western Blot (Assay dependent)
Alternate names:
Plasminogen-binding protein; Phosphopyruvate hydratase; Non-neural enolase; MPB-1; MBP-1; Enolase 1; C-myc promoter-binding protein; 2-phospho-D-glycerate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130493 449.00
Check your price
  • Endothelium Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA190996
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190996 449.00
Check your price
  • CD34 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA139456
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA139456 449.00
Check your price
  • CD34 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA137329
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137329 449.00
Check your price
  • Caveolin 1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA139458
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
BSCL3, CGL3, MSTP085, VIP21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA139458 449.00
Check your price
  • BRCA2 Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185156
  • Size 1mL
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/25-1/100)
Alternate names:
Fanconi anemia group D1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185156 449.00
Check your price
  • Annexin A1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137083
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
p35; Phospholipase A2 inhibitory protein; Lipocortin I; Chromobindin-9; Calpactin-2; Calpactin II; Annexin-1; Annexin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137083 449.00
Check your price
  • 14-3-3 zeta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137006
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:25)
Alternate names:
Protein kinase C inhibitor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137006 449.00
Check your price
  • 14-3-3 beta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137002
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
Protein kinase C inhibitor protein 1; Protein 1054
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137002 449.00
Check your price
  • Parainfluenza Virus Type 3 Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516499
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
hPIV-3, PIV-3, Parainfluenza Virus Type 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516499 449.00
Check your price
  • TIMP1 for Ctrl
  • Host Insect cells
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77537
  • Size 10µg
Applications
Control (Assay Dependent)
Alternate names:
TIMP 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77537 449.00
Check your price
  • Sphingosine Kinase 2, Long for Ctrl
  • Host Insect cells
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP12950
  • Size 10µg
Applications
Control (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP12950 449.00
Check your price
  • Sphingosine Kinase 1a for Ctrl
  • Host Baculovirus
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # RP12951
  • Size 10µg
Applications
Control (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP12951 449.00
Check your price
  • Sphingosine Kinase 1 for Ctrl
  • Host Insect cells
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP12948
  • Size 10µg
Applications
Control (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP12948 449.00
Check your price
  • MMP24 for Ctrl
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77536
  • Size 10µg
Applications
Control (Assay Dependent)
Alternate names:
MT5-MMP
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77536 449.00
Check your price
  • MMP17 for Ctrl
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77535
  • Size 10µg
Applications
Control (Assay Dependent)
Alternate names:
MT4-MMP
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77535 449.00
Check your price
  • MMP14 for Ctrl
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77532
  • Size 10µg
Applications
Control (Assay Dependent)
Alternate names:
MT1-MMP
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77532 449.00
Check your price
  • SPHK2 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112872
  • Size 10Miniblots
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112872 449.00
Check your price
  • SPHK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA112856
  • Size 10Miniblots
Applications
Western Blot (1:1,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112856 449.00
Check your price
  • SPHK1 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112719
  • Size 10Miniblots
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
RP23-193A16.4, 1110006G24Rik, Sk1, Spk1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112719 449.00
Check your price