Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  76  77  78  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  110  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1611 
2701 - 2730 of 48310 products displayed
  • CD62L Antibody, PE (Monoclonal, OX-85)
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA517576
  • Size 200 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
SELL, LAM-1, LAM1, LECAM1, LNHR, LSEL, LYAM1, Leu-8, Lyam-1, PLNHR, TQ1, hLHRc
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517576 425.00
Check your price
  • CD21 Antibody (Monoclonal, LB21)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134415
  • Size 200 tests
Applications
Flow Cytometry (Neat (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (1:10-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Epstein-Barr virus receptor; Complement C3d receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134415 425.00
Check your price
  • CaMKII alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137226
  • Size 1mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
CAMKA, KIAA0968
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137226 425.00
Check your price
  • FGFR1 Antibody (Monoclonal, M2F12)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA126256
  • Size 125 µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1 µg/ml)
Alternate names:
Proto-oncogene c-Fgr; MFR; Basic fibroblast growth factor receptor 1; N-sam; Fms-like tyrosine kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126256 425.00
Check your price
  • FGFR1 Antibody (Monoclonal, M1C4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190880
  • Size 125 µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Proto-oncogene c-Fgr; N-sam; Fms-like tyrosine kinase 2; Basic fibroblast growth factor receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190880 425.00
Check your price
  • Lamin C Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA106105
  • Size 100µL
Applications
Flow Cytometry (1:50-1:250), Immunohistochemistry (Frozen) (1:50-1:250), Western Blot (1:50-1:500)
Alternate names:
OR19-15, OR19-8, OR7G1P
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA106105 425.00
Check your price
  • CD11c Antibody, FITC (Monoclonal, 8A2)
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA516551
  • Size 100ug
Applications
Flow Cytometry (Neat)
Alternate names:
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516551 425.00
Check your price
  • MAdCAM1 (soluble) Antibody, RPE (Monoclonal, 314G8)
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA516498
  • Size 100TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
MACAM1,MAdCAM-1; hMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516498 425.00
Check your price
  • CD3z Antibody, PE (Monoclonal, G3)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate PE
  • Catalog # MA516689
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
T-cell receptor T3 zeta chain; T-cell receptor T3 eta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516689 425.00
Check your price
  • XAGE1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132314
  • Size 100 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
G antigen family D member 2; Cancer/testis antigen 12.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132314 425.00
Check your price
  • UCP1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA124894
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:500), Western Blot (1:1,000)
Alternate names:
Thermogenin; Solute carrier family 25 member 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124894 425.00
Check your price
  • Trenbolone Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175028
  • Size 100 µL
Applications
ELISA (2.5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175028 425.00
Check your price
  • Testosterone Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175024
  • Size 100 µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175024 425.00
Check your price
  • Tankyrase Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132225
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Tankyrase I; TRF1-interacting ankyrin-related ADP-ribose polymerase; TNKS-1; Poly [ADP-ribose] polymerase 5A; ADP-ribosyltransferase diphtheria toxin-like 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132225 425.00
Check your price
  • TACR1 Antibody
  • Host Rabbit
  • Target Species Guinea pig, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132229
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tachykinin receptor 1; NK-1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132229 425.00
Check your price
  • Sulfamerazine Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175430
  • Size 100 µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175430 425.00
Check your price
  • SR-BI Antibody
  • Host Rabbit
  • Target Species Hamster, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131393
  • Size 100 µL
Applications
Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (4-8 µg/ml of lysate), Western Blot (1:1000)
Alternate names:
SR-BI; Collagen type I receptor, thrombospondin receptor-like 1; CD36 antigen-like 1; CD36 and LIMPII analogous 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131393 425.00
Check your price
  • SLC4A1 Antibody (Monoclonal, BIII 136)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120211
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Solute carrier family 4 member 1; Anion exchange protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120211 425.00
Check your price
  • SCP3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131226
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:400), Western Blot (Assay-dependent)
Alternate names:
COR1, RNASCP3, Sycp3, Synaptonemal complex protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131226 425.00
Check your price
  • SCP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131221
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
Cancer/testis antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131221 425.00
Check your price
  • SCP1 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA131223
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:750), Immunofluorescence (1:100-1:750), Immunohistochemistry (Frozen) (1:750), Immunohistochemistry (Paraffin) (1:750)
Alternate names:
AW742678, DlkI, FA1, Ly107, Peg9, SCP1, ZOG, pG2, pref-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131223 425.00
Check your price
  • RCC1 Antibody
  • Host Rabbit
  • Target Species Human, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA15868
  • Size 100 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:1000)
Alternate names:
Chromosome condensation protein 1; Cell cycle regulatory protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15868 425.00
Check your price
  • RAN Antibody
  • Host Rabbit
  • Target Species Human, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA15864
  • Size 100 µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:500)
Alternate names:
Ras-related nuclear protein; Ras-like protein TC4; GTPase Ran; Androgen receptor-associated protein 24
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15864 425.00
Check your price
  • PP2A alpha Antibody (Monoclonal, 2G9)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA518064
  • Size 100 µL
Applications
Immunoprecipitation (Assay-dependent), Western Blot (1:1000)
Alternate names:
Replication protein C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518064 425.00
Check your price
  • PKP3 Antibody (Monoclonal, 7F6)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA518080
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
PKP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518080 425.00
Check your price
  • PKP1 Antibody (Monoclonal, 19F10)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA518082
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
Band 6 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518082 425.00
Check your price
  • Phospho-POLR2A (Ser5) Antibody (Monoclonal, 4H8)
  • Host Mouse
  • Target Species C. elegans, Fruit fly, Human, Mouse, Non-human primate, Plant, Yeast
  • Conjugate Unconjugated
  • Catalog # MA518089
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; RNA-directed RNA polymerase II subunit RPB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518089 425.00
Check your price
  • PARP Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3951
  • Size 100 µL
Applications
ELISA (Assay dependent), Western Blot (1:500)
Alternate names:
Poly[ADP-ribose] synthase 1; NAD(+) ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3951 425.00
Check your price
  • p19ARF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130670
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), ELISA (Assay dependent), Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (1 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:1500)
Alternate names:
p16-INK4a; Cyclin-dependent kinase 4 inhibitor A; p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p19ARF; Multiple tumor suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130670 425.00
Check your price
  • OVCA2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA15859
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15859 425.00
Check your price