Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1630 
2881 - 2910 of 48882 products displayed
  • FGFR4 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518084
  • Size 100µL
Applications
Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
TKF; JTK2; CD334; MGC20292; Fibroblast growth factor receptor 4; CD334
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518084 425.00
Check your price
  • Dynorphin B Polyclonal Antibody for IHC (F) and RIA
  • Host Rabbit
  • Target Species Guinea pig, Hamster, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA184561
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:250-1:2500), Radioimmune Assays (1/5000)
Alternate names:
Preprodynorphin; Beta-neoendorphin-dynorphin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184561 425.00
Check your price
  • Dopamine Transporter Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA518079
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:1000)
Alternate names:
Solute carrier family 6 member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518079 425.00
Check your price
  • DNMT3B Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132317
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
DNA methyltransferase HsaIIIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132317 425.00
Check your price
  • DNMT1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132315
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
MCMT; DNA methyltransferase HsaI; CXXC-type zinc finger protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132315 425.00
Check your price
  • DLX4 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA132425
  • Size 100µL
Applications
Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay-Dependent)
Alternate names:
Homeobox protein DLX-7; Homeobox protein DLX-8; Beta protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132425 425.00
Check your price
  • DJ-1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Fish, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132440
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Parkinson disease protein 7 homolog; Fertility protein SP22; Contraception-associated protein 1; Parkinson disease protein 7; Oncogene DJ1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132440 425.00
Check your price
  • Cytokeratin 13 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15764
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:1000-1:3000)
Alternate names:
Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15764 425.00
Check your price
  • Cytokeratin 10 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15766
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1:1000)
Alternate names:
Keratin-10; Cytokeratin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15766 425.00
Check your price
  • CYP46A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132446
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:1000)
Alternate names:
Cytochrome P450 46A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132446 425.00
Check your price
  • Complement C4d Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518044
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:25), Immunohistochemistry (Paraffin) (1:25)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; Acidic complement C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518044 425.00
Check your price
  • Collagen IV Polyclonal Antibody for IF, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA185320
  • Size 100µL
Applications
ELISA (1:100-1:400), Immunofluorescence (1:40-1:160), Immunohistochemistry (Paraffin) (1:250-1:1000)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185320 425.00
Check your price
  • CKAP5 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Mammal, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA132456
  • Size 100µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
Colonic and hepatic tumor overexpressed gene protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132456 425.00
Check your price
  • CDH11 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518085
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
Osteoblast cadherin; OSF-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518085 425.00
Check your price
  • CD34 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA518091
  • Size 100µL
Applications
Flow Cytometry (1:50), Immunohistochemistry (Frozen) (Assay-dependent), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518091 425.00
Check your price
  • CD33 Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518056
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (2 ug/ml)
Alternate names:
gp67; Sialic acid-binding Ig-like lectin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518056 425.00
Check your price
  • Calnexin Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Avian, Bovine, Guinea pig, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA130197
  • Size 100µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Assay-dependent), Immunoprecipitation (1:100), Western Blot (1:5000)
Alternate names:
p90; Major histocompatibility complex class I antigen-binding protein p88; IP90
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130197 425.00
Check your price
  • Brugia malayi filaria Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA518034
  • Size 100µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
philariasis, filariasis, Brugia malayi filaria, B. malayi
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518034 425.00
Check your price
  • BRIP1 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132422
  • Size 100µL
Applications
Immunohistochemistry (Assay-dependent), Immunoprecipitation (2 ul/IP), Western Blot (1:2000)
Alternate names:
BRCA1-interacting protein C-terminal helicase 1; BRCA1-associated C-terminal helicase 1; ATP-dependent RNA helicase BRIP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132422 425.00
Check your price
  • APOE Monoclonal Antibody for Western Blot, Flow, IP, ELISA and DB
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518068
  • Size 100µL
Applications
Dot blot (Assay-dependent), ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:64,000)
Alternate names:
AD2; LDLCQ5; LPG; apo-E; apolipoprotein E4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518068 425.00
Check your price
  • AJUBA Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518087
  • Size 100µL
Applications
Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
LIM domain-containing protein ajuba; JUB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518087 425.00
Check your price
  • ABCE1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132357
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:250-1:1000)
Alternate names:
Ribonuclease 4 inhibitor; RNase L inhibitor; HuHP68; 2'-5'-oligoadenylate-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132357 425.00
Check your price
  • ABCA1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132129
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132129 425.00
Check your price
  • 14-3-3 gamma Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA186556
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1/800), Western Blot (1:6000-1:12,000)
Alternate names:
Protein kinase C inhibitor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186556 425.00
Check your price
  • ZNF683 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130046
  • Size 100µg
Applications
Immunohistochemistry (4-8 µg/ml), Western Blot (1.25 µg/ml)
Alternate names:
RP11-569G9.6, MGC33414
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130046 425.00
Check your price
  • VSV-G Tag Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA129903
  • Size 100µg
Applications
ELISA (1:1000-1:30,000), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400), Western Blot (1:1000-1:30,000)
Alternate names:
Vesicular stomatitis virus, VSV, VSV glycoprotein, YTDIEMNRLGK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129903 425.00
Check your price
  • VEGF Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA121465
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121465 425.00
Check your price
  • VAMP1 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA134662
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:1000)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134662 425.00
Check your price
  • TLX Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 434500
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein tailless homolog; Nuclear receptor TLX
Catalog # Size Price (AUD) Your Price (AUD) Qty
434500 425.00
Check your price
  • TLR7 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA181963
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (1:1000-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181963 425.00
Check your price