Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1100  1622 
3061 - 3090 of 48655 products displayed
  • CD154 Monoclonal Antibody, FITC conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # HMCD15401
  • Size 500 µg
Applications
Flow Cytometry (0.1-0.2 ug per 1x10^6 cells)
Alternate names:
Tumor necrosis factor ligand superfamily member 5; TNF-related activation protein; T-cell antigen Gp39
Catalog # Size Price (AUD) Your Price (AUD) Qty
HMCD15401 400.00
Check your price
  • CTLA-4 Monoclonal Antibody, FITC conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # HMCD15201
  • Size 500 µg
Applications
Flow Cytometry (0.1-0.2 ug per 1x10^6 cells)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
HMCD15201 400.00
Check your price
  • beta Galactosidase Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), IP, ELISA and DB
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA121477
  • Size 1 mg
Applications
Dot blot (Assay dependent), ELISA (1:4000-1:16,000), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:500-1:3000), Immunoprecipitation (Assay dependent), Western Blot (1:2000-1:5000)
Alternate names:
Beta-Gal, RP3-337O18.1, GLB2, GSL, NGBE, PPCA, PPGB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121477 399.00
Check your price
  • Ferritin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191847
  • Size 1 mg
Applications
ELISA (Assay dependent)
Alternate names:
MGC71996
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191847 399.00
Check your price
  • Goat anti-Donkey IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Donkey
  • Conjugate AP
  • Catalog # PA130377
  • Size 1 mg
Applications
ELISA (1:2000-1:6000), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130377 399.00
Check your price
  • Rabbit anti-Hamster IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Rabbit
  • Target Species Hamster
  • Conjugate AP
  • Catalog # PA130348
  • Size 1 mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130348 399.00
Check your price
  • Rabbit anti-Rat IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Rabbit
  • Target Species Rat
  • Conjugate AP
  • Catalog # PA130338
  • Size 1 mg
Applications
ELISA (1:2000-1:10,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunomicroscopy (Assay-dependent), Western Blot (1:500-1:2500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130338 399.00
Check your price
  • RSV NP Monoclonal Antibody for IF, IHC (F) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA180039
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:100)
Alternate names:
human respiratory syncytial virus, RSV, RSV NP, RSV Nuclear Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180039 399.00
Check your price
  • TSH beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183492
  • Size 1 mg
Applications
ELISA (Assay Dependent)
Alternate names:
Thyrotropin beta chain; Thyroid-stimulating hormone subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183492 399.00
Check your price
  • alpha Tubulin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA138814
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Tubulin alpha-1 chain; Tubulin H2-alpha; Testis-specific alpha-tubulin; Alpha-tubulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138814 399.00
Check your price
  • Amylin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137075
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Insulinoma amyloid peptide; Diabetes-associated peptide; Amylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137075 399.00
Check your price
  • Apolipoprotein B Polyclonal Antibody for Western Blot and IDF
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA127284
  • Size 1 mL
Applications
Immunodiffusion (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
mCG_129875, AI315052, MGC176318, apob-100, apob-48
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127284 399.00
Check your price
  • Bcl-xL Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137161
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:100)
Alternate names:
Apoptosis regulator Bcl-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137161 399.00
Check your price
  • beta Amyloid Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137077
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137077 399.00
Check your price
  • C1q Polyclonal Antibody, FITC conjugate for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate FITC
  • Catalog # PA129720
  • Size 1 mL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (1:20-1:40)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129720 399.00
Check your price
  • Calretinin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA137223
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
29 kDa calbindin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137223 399.00
Check your price
  • CaMKIV Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA137224
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
CaM kinase-GR; Calspermin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137224 399.00
Check your price
  • CAS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137229
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Importin-alpha re-exporter; Chromosome segregation 1-like protein; Cellular apoptosis susceptibility protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137229 399.00
Check your price
  • CD138 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137379
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
SDC1, SYND1, SDC, CD antigen 138
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137379 399.00
Check your price
  • CD19 Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135112
  • Size 1 mL
Applications
Flow Cytometry (Assay dependent.), Immunohistochemistry (Frozen) (1:5-1:20)
Alternate names:
T-cell surface antigen Leu-12; Differentiation antigen CD19; B-lymphocyte surface antigen B4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135112 399.00
Check your price
  • CD20 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137312
  • Size 1 mL
Applications
Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1:300), Western Blot (1:25)
Alternate names:
Membrane-spanning 4-domains subfamily A member 1; Leukocyte surface antigen Leu-16; Bp35; B-lymphocyte surface antigen B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137312 399.00
Check your price
  • CD3z Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137284
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
T-cell receptor T3 zeta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137284 399.00
Check your price
  • CD40 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137334
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
CDw40; CD40L receptor; Bp50; B-cell surface antigen CD40
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137334 399.00
Check your price
  • CD45RO Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA133243
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133243 399.00
Check your price
  • CDK1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137386
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137386 399.00
Check your price
  • Chlamydia trachomatis MOMP Polyclonal Antibody for IF, IHC (F) and ELISA
  • Host Goat
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA533131
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C. trachomatis major outer membrane protein, C. trachomatis MOMP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533131 399.00
Check your price
  • Claudin 3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137469
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Rat ventral prostate.1 protein homolog; Clostridium perfringens enterotoxin receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137469 399.00
Check your price
  • Claudin 4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137471
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Williams-Beuren syndrome chromosomal region 8 protein; Clostridium perfringens enterotoxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137471 399.00
Check your price
  • Claudin 5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137473
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Transmembrane protein deleted in VCFS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137473 399.00
Check your price
  • Complement C3c Polyclonal Antibody, FITC conjugate for IF and ICC
  • Host Rabbit
  • Target Species Bovine, Dog, Cat, Guinea pig, Goat, Human, Mink, Mouse, Sheep, Pig, Rat
  • Conjugate FITC
  • Catalog # PA129718
  • Size 1 mL
Applications
Immunocytochemistry (1:75-1:100), Immunofluorescence (1:75-1:100)
Alternate names:
HSE-MSF; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129718 399.00
Check your price