Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  80  90  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  112  113  114  115  116  117  118  119  120  121  122  123  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1100  1630 
3241 - 3270 of 48877 products displayed
  • TNF alpha Monoclonal Antibody, Biotin conjugate for Flow and ELISA
  • Host Rat
  • Target Species Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA120303
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay-dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120303 399.00
Check your price
  • TNFR1 Monoclonal Antibody for IHC (F), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA135667
  • Size 100µg
Applications
ELISA (1:10-1:50), Flow Cytometry (1:10), Immunohistochemistry (Frozen) (1:10), Immunoprecipitation (Assay dependent.)
Alternate names:
p60; p55; Tumor necrosis factor receptor type I; Tumor necrosis factor receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135667 399.00
Check your price
  • TRF2 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, IP, ChIP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA141001
  • Size 100µg
Applications
ChIP assay (1:10-1:500), ELISA (2 µg/ml), Flow Cytometry (0.1 µg/10^6 cells), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (5 µg/ml), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (2 µg/10^6 cells), Western Blot (2-4 µg/ml)
Alternate names:
Telomeric DNA-binding protein; TTAGGG repeat-binding factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141001 399.00
Check your price
  • Trypsin Pan Polyclonal Antibody for Western Blot, IHC (P), IP and Neu
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546939
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (0.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Trypsin II; Serine protease 2; Anionic trypsinogen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546939 399.00
Check your price
  • TXNDC5 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129840
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (0.1-1 µg/ml)
Alternate names:
Thioredoxin-like protein p46; Endoplasmic reticulum resident protein 46
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129840 399.00
Check your price
  • UGCG Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112760
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
UDP-glucose:N-acylsphingosine D-glucosyltransferase; UDP-glucose ceramide glucosyltransferase; Glucosylceramide synthase; GLCT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112760 399.00
Check your price
  • Varicella zoster Virus Monoclonal Antibody, FITC conjugate for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA183517
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
Varicellovirus, VZV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183517 399.00
Check your price
  • VCAM-1 Polyclonal Antibody for Western Blot, IHC (F), Flow and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547029
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
CD106, DKFZp779G2333, INCAM-100, MGC99561, CD106 antigen, vascular cell adhesion protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547029 399.00
Check your price
  • VISTA Polyclonal Antibody for Western Blot, IF and ICC
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547032
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Protein L259
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547032 399.00
Check your price
  • VMAT2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127477
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Vesicular amine transporter 2; Solute carrier family 18 member 2; Monoamine transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127477 399.00
Check your price
  • VSV-G Tag Polyclonal Antibody, FITC conjugate for IF and ICC
  • Host Goat
  • Target Species Tag
  • Conjugate FITC
  • Catalog # PA131803
  • Size 100µg
Applications
Immunocytochemistry (1:200-1:2000), Immunofluorescence (Assay-dependent)
Alternate names:
Vesicular stomatitis virus, VSV, VSV glycoprotein, YTDIEMNRLGK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131803 399.00
Check your price
  • XPA Monoclonal Antibody for Western Blot, IF, ICC, ELISA and DB
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121460
  • Size 100µg
Applications
Dot blot (Assay dependent), ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (1:1000-1:1500)
Alternate names:
Xeroderma pigmentosum group A-complementing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121460 399.00
Check your price
  • XPG Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126186
  • Size 100µg
Applications
Immunoprecipitation (2 µg/mg of protein lysate), Western Blot (1:625-1:1250)
Alternate names:
Xeroderma pigmentosum group G-complementing protein; DNA excision repair protein ERCC-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126186 399.00
Check your price
  • ZNF174 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124365
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Zinc finger and SCAN domain-containing protein 8; AW-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124365 399.00
Check your price
  • ABO Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537352
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
NAGAT; Histo-blood group B transferase; Histo-blood group A transferase; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein 3-alpha-galactosyltransferase; Cis-AB transferase; NAGAT 1; N-acetylgalactosaminyltransferase A blood group-like enzyme; Cis-AB transferase 1; Blood group A glycosyltransferase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537352 399.00
Check your price
  • Afadin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125075
  • Size 100µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Assay Dependent), Western Blot (1:2000)
Alternate names:
Protein Af-6; ALL1-fused gene from chromosome 6 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125075 399.00
Check your price
  • alpha Catenin Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA125081
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Renal carcinoma antigen NY-REN-13; Cadherin-associated protein; Alpha E-catenin; CAP102; 102 kDa cadherin-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125081 399.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118264
  • Size 100µL
Applications
Immunohistochemistry (1:500-1:3000), Western Blot (1:500-1:3000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118264 399.00
Check your price
  • Amitryptyline Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175475
  • Size 100µL
Applications
ELISA (Assay Dependent)
Alternate names:
Amitryptyline, Elavil, Tryptanol, Endep, Elatrol, Tryptizol, Trepiline, Laroxyl
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175475 399.00
Check your price
  • Androstanedione Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175046
  • Size 100µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175046 399.00
Check your price
  • ATG16L1 Polyclonal Antibody for Western Blot, IF, IHC, IP and ELISA
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118296
  • Size 100µL
Applications
ELISA (Assay dependent), Immunofluorescence (1:100-1:3000), Immunohistochemistry (1:100-1:3000), Western Blot (1:100-1:3000)
Alternate names:
APG16-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118296 399.00
Check your price
  • ATP6AP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537328
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Vacuolar proton pump subunit S1; V-ATPase S1 accessory protein; V-ATPase Ac45 subunit; Protein C7-1; C7-1 protein; Protein XAP-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537328 399.00
Check your price
  • Bcl-10 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120160
  • Size 100µL
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1-2 µg/ml)
Alternate names:
Mammalian CARD-containing adapter molecule E10; Cellular-E10; Cellular homolog of vCARMEN; CED-3/ICH-1 prodomain homologous E10-like regulator; CARD-like apoptotic protein; CARD-containing molecule enhancing NF-kappa-B; B-cell CLL/lymphoma 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120160 399.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118357
  • Size 100µL
Applications
ELISA (1:1,000), Immunohistochemistry (1:1,000), Inhibition Assays (1:10-1:50), Western Blot (1:1,000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118357 399.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118364
  • Size 100µL
Applications
ELISA (1:200-1:2000), Immunohistochemistry (1:200-1:2000), Inhibition Assays (1:10-1:50), Western Blot (1:200-1:2000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118364 399.00
Check your price
  • BDNF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118375
  • Size 100µL
Applications
Western Blot (1:500-1:2,000)
Alternate names:
Abrineurin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118375 399.00
Check your price
  • Bestrophin 1 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA191895
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Vitelliform macular dystrophy protein 2; TU15B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191895 399.00
Check your price
  • beta Catenin Monoclonal Antibody, Cyanine3 conjugate for IF, ICC and IP
  • Host Mouse
  • Target Species Avian, Chicken, Mammal
  • Conjugate Cyanine3
  • Catalog # MA134999
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134999 399.00
Check your price
  • BNIP3 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124688
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay Dependent), Western Blot (2-4 µg/ml)
Alternate names:
NIP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124688 399.00
Check your price
  • BVES Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537134
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Popeye domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537134 399.00
Check your price