Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  115  116  117  118  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  150  160  170  180  190  200  210  220  300  400  500  600  700  800  900  1000  1100  1630 
3871 - 3900 of 48877 products displayed
  • NGF Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Avian, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118377
  • Size 500µg
Applications
ELISA (1-3 µg/ml), Immunohistochemistry (1-3 µg/ml), Inhibition Assays (1-3 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
beta Nerve Growth Factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118377 399.00
Check your price
  • Pancreatic Amylase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110619
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
1,4-alpha-D-glucan glucanohydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110619 399.00
Check your price
  • Plasminogen Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120079
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
RP1-81D8.1, DKFZp779M0222
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120079 399.00
Check your price
  • RCA60 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Plant
  • Conjugate Unconjugated
  • Catalog # MA191116
  • Size 500µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191116 399.00
Check your price
  • Staphylococcus aureus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110708
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
S. aureus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110708 399.00
Check your price
  • Staphylococcus aureus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110710
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
S. aureus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110710 399.00
Check your price
  • Streptavidin Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA120010
  • Size 500µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120010 399.00
Check your price
  • Streptavidin Monoclonal Antibody for Western Blot, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA120011
  • Size 500µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay dependent), Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120011 399.00
Check your price
  • Streptococcus agalactiae Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110871
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
S. agalactiae
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110871 399.00
Check your price
  • Transferrin Receptor Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA190784
  • Size 500µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Western Blot (Assay dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190784 399.00
Check your price
  • Trypsin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110616
  • Size 500µg
Applications
ELISA (1:12800)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110616 399.00
Check your price
  • Yersinia pestis F1 Antigen Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA123074
  • Size 500µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Bubonic Plague, Pasturella pestis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123074 399.00
Check your price
  • Actin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511866
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 ug), Western Blot (1:5000-1:20,000)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511866 399.00
Check your price
  • ADAM17 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516551
  • Size 500µL
Applications
Immunoprecipitation (10 µg/ml), Western Blot (5 µg/mL)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516551 399.00
Check your price
  • alpha Sarcomeric Actinin Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA122863
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:25-1:50)
Alternate names:
Non-muscle alpha-actinin-1; F-actin cross-linking protein; Alpha-actinin cytoskeletal isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122863 399.00
Check your price
  • alpha Tubulin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Amphibian, Bovine, Gerbil, Guinea pig, Human, Mouse, Pig, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA120988
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Tubulin alpha-4 chain; Alpha-tubulin 4; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-4; Tubulin H2-alpha; Testis-specific alpha-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120988 399.00
Check your price
  • Bax Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513991
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Bcl-2-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513991 399.00
Check your price
  • Bax Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA514000
  • Size 500µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:10-1:100)
Alternate names:
Bcl-2-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514000 399.00
Check your price
  • Bcl-X Monoclonal Antibody, Biotin conjugate for IF, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA514008
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml)
Alternate names:
Apoptosis regulator Bcl-X
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514008 399.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA121167
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121167 399.00
Check your price
  • Blood Group Lewis A Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126522
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:40)
Alternate names:
Fucosyltransferase III; Fucosyltransferase 3; Blood group Lewis alpha-4-fucosyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126522 399.00
Check your price
  • Blood Group Lewis Y Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126525
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (1:40), Western Blot (Assay Dependent)
Alternate names:
Fucosyltransferase III; Fucosyltransferase 3; Blood group Lewis alpha-4-fucosyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126525 399.00
Check your price
  • Borrelia burgdorferi Polyclonal Antibody, HRP conjugate for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate HRP
  • Catalog # PA127289
  • Size 500µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:20-1:200)
Alternate names:
B. burgdorferi, Lyme disease, Borrelia
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127289 399.00
Check your price
  • c-Abl Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # MA514395
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (12-24 µg/ml), Immunoprecipitation (2 µg/mg lysate), Western Blot (1 µg/mL)
Alternate names:
p150; Proto-oncogene c-Abl; Abelson tyrosine-protein kinase 1; Abelson murine leukemia viral oncogene homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514395 399.00
Check your price
  • Calretinin Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512539
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:10-1:20)
Alternate names:
29 kDa calbindin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512539 399.00
Check your price
  • Caspase 3 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA129157
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (1:2000), Western Blot (1:500)
Alternate names:
Cysteine protease CPP32; Apopain; SREBP cleavage activity 1; Protein Yama; LICE; IRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129157 399.00
Check your price
  • Caspase 3 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # PA516332
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
Cysteine protease CPP32; Apopain; SREBP cleavage activity 1; Protein Yama; LICE; IRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516332 399.00
Check your price
  • Caspase 8 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129159
  • Size 500µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (1:2000), Western Blot (1:500)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129159 399.00
Check your price
  • Caspase 9 Polyclonal Antibody, Biotin conjugate for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Biotin
  • Catalog # PA516355
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/mL), Immunoprecipitation (10 µg/ml), Western Blot (2.5-5.0 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516355 399.00
Check your price
  • CD146 Monoclonal Antibody for IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134759
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (Assay-dependent)
Alternate names:
S-endo 1 endothelial-associated antigen; Melanoma-associated antigen MUC18; Melanoma-associated antigen A32; Melanoma cell adhesion molecule; Cell surface glycoprotein P1H12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134759 399.00
Check your price