Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  119  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  150  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1605 
3991 - 4020 of 48123 products displayed
  • Cdc6 Antibody, Biotin (Monoclonal, DCS-180)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA514311
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p62(cdc6); Cdc18-related protein; CDC6-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514311 399.00
Check your price
  • CDK1 Antibody, Biotin (Monoclonal, A17.1.1)
  • Host Mouse
  • Target Species Chicken, Guinea pig, Human, Mouse, Rodent, Rat, Xenopus
  • Conjugate Biotin
  • Catalog # MA511469
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog; p34 protein kinase 1; Cell division protein kinase 1-A; Cell division control protein 2-A; Cell division control protein 2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511469 399.00
Check your price
  • CDK4 Antibody, Biotin (Monoclonal, DCS-35)
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA512981
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
PSK-J3; Cell division protein kinase 4; CRK3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512981 399.00
Check your price
  • CDK6 Antibody, Biotin (Monoclonal, K6.83 (DCS-83))
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA513330
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Serine/threonine-protein kinase PLSTIRE; Cell division protein kinase 6; CR2 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513330 399.00
Check your price
  • CDK8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121780
  • Size 500 µL
Applications
ChIP assay (1-3 ul), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (10 µg/mg of lysate), Western Blot (1-2 µg/ml)
Alternate names:
Protein kinase K35; Mediator of RNA polymerase II transcription subunit CDK8; Mediator complex subunit CDK8; Cell division protein kinase 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121780 399.00
Check your price
  • Cholera Toxin beta Antibody
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA125635
  • Size 500 µL
Applications
ELISA (1:1000), Immunohistochemistry (Assay Dependent)
Alternate names:
beta subunit cholera toxin, CTB, VC1456
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125635 399.00
Check your price
  • Collagen II Antibody, Biotin (Monoclonal, 2B1.5)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA512786
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512786 399.00
Check your price
  • COX2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120955
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Prostaglandin-endoperoxide synthase 2; Prostaglandin H2 synthase 2; PHS II; Cyclooxygenase-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120955 399.00
Check your price
  • Cyclin D3 Antibody, Biotin (Monoclonal, DCS-28.1)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # MA512723
  • Size 500 µL
Applications
Western Blot (1-2 µg/ml)
Alternate names:
D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3,OTTHUMP00000223415
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512723 399.00
Check your price
  • Cyclin E Antibody, Biotin (Monoclonal, HE12)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # MA514333
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
G1/S-specific cyclin-E1, cyclin Es, cyclin Et
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514333 399.00
Check your price
  • Cytochrome C Antibody, Biotin (Monoclonal, CTC03 (2B5))
  • Host Mouse
  • Target Species Bovine, Dog, Horse, Human, Mouse
  • Conjugate Biotin
  • Catalog # MA511820
  • Size 500 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunoprecipitation (2 µg/ml)
Alternate names:
cytochrome c
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511820 399.00
Check your price
  • Cytokeratin 8 Antibody (Monoclonal, 35bH11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126238
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (1:10-25), Immunohistochemistry (Paraffin) (1:10-25)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126238 399.00
Check your price
  • Cytokeratin Pan Type II Antibody (Monoclonal, AE3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134220
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134220 399.00
Check your price
  • DNA-PK Antibody, Biotin (Monoclonal, 18-2)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # MA513235
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513235 399.00
Check your price
  • E. coli serotype O/K Antibody
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA125636
  • Size 500 µL
Applications
ELISA (1:2000), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Escherichia coli
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125636 399.00
Check your price
  • E2F1 Antibody, Biotin (Monoclonal, KH95)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # MA514341
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-100), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
pRB-binding protein E2F-1; Retinoblastoma-binding protein 3; Retinoblastoma-associated protein 1; PBR3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514341 399.00
Check your price
  • EGFR Antibody, Biotin (Monoclonal, 111.6)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513266
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513266 399.00
Check your price
  • EGFR Antibody, Biotin (Monoclonal, 528 + 199.12)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513694
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml)
Alternate names:
Receptor tyrosine-protein kinase erbB-1; Proto-oncogene c-ErbB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513694 399.00
Check your price
  • ErbB2 Antibody, Biotin (Monoclonal, 3B5)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Biotin
  • Catalog # MA513672
  • Size 500 µL
Applications
Immunohistochemistry (1:1000-1:1500), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513672 399.00
Check your price
  • ErbB2 Antibody, Biotin (Monoclonal, e2-4001)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA513102
  • Size 500 µL
Applications
Immunohistochemistry (1:200-400), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513102 399.00
Check your price
  • ErbB2 Antibody, Biotin (Monoclonal, N12)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512995
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:50-1:200), Immunomicroscopy (Assay Dependent)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512995 399.00
Check your price
  • ErbB3 Antibody, Biotin (Monoclonal, 2B5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA513037
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3; Glial growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513037 399.00
Check your price
  • ErbB3 Antibody, Biotin (Monoclonal, H3.90.6 (Ab90))
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512864
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512864 399.00
Check your price
  • ErbB4 Antibody, Biotin (Monoclonal, H4.72.8 (Ab72))
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513013
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Western Blot (1:50-1:500)
Alternate names:
p180erbB4; Tyrosine kinase-type cell surface receptor HER4; Proto-oncogene-like protein c-ErbB-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513013 399.00
Check your price
  • ErbB4 Antibody, Biotin (Monoclonal, H4.77.16 (Ab77))
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA512885
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
p180erbB4; Tyrosine kinase-type cell surface receptor HER4; Proto-oncogene-like protein c-ErbB-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512885 399.00
Check your price
  • ERCC1 Antibody, Biotin (Monoclonal, 3H11)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513827
  • Size 500 µL
Applications
Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
DNA excision repair protein ERCC-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513827 399.00
Check your price
  • Estrogen Receptor alpha Antibody, Biotin (Monoclonal, TE111.5D11)
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA513062
  • Size 500 µL
Applications
ChIP assay (Assay Dependent), Gel Shift (1mg/mL), Immunoprecipitation (2 µg/ml), Western Blot (1:1000-1:5000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513062 399.00
Check your price
  • Ezrin Antibody, Biotin (Monoclonal, 3C12)
  • Host Mouse
  • Target Species Bovine, Hamster, Human, Marsupial, Mouse, Non-human primate, Rat
  • Conjugate Biotin
  • Catalog # MA513859
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
p81; Villin-2; Cytovillin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513859 399.00
Check your price
  • Gastrin Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516409
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516409 399.00
Check your price
  • GLUT1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA121041
  • Size 500 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1:200), Western Blot (0.5 µg/ml for 2 hrs)
Alternate names:
Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121041 399.00
Check your price