Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
4021 - 4050 of 48881 products displayed
  • APC Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191039
  • Size 50µg
Applications
Western Blot (1-1.5 µg/ml)
Alternate names:
Deleted in polyposis 2.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191039 399.00
Check your price
  • Apolipoprotein J Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125833
  • Size 50µg
Applications
Immunocytochemistry (1:2,000), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:2000)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125833 399.00
Check your price
  • Aquaporin 1 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA124915
  • Size 50µg
Applications
ELISA (1:1000-1:10,000), Immunoprecipitation (1:1000-1:10,000), Western Blot (1:1000-1:2000)
Alternate names:
Water channel protein for red blood cells and kidney proximal tubule; Aquaporin-CHIP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124915 399.00
Check your price
  • ARFGAP3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131452
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
RP11-261N11.1, ARF1GAP, HRIHFB2281, MGC39924
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131452 399.00
Check your price
  • ARIH1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131804
  • Size 50µg
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitin-conjugating enzyme E2-binding protein 1; UbcM4-interacting protein 77; UbcH7-binding protein; Protein ariadne-1 homolog; UbcM4-interacting protein; Monocyte protein 6; HHARI; H7-AP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131804 399.00
Check your price
  • ARL2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131653
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
ARFL2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131653 399.00
Check your price
  • ARL4A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131566
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
ARL4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131566 399.00
Check your price
  • BAF53A Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131249
  • Size 50µg
Applications
ELISA (1:64,000), Western Blot (1-3 µg/ml)
Alternate names:
INO80 complex subunit K; BRG1-associated factor 53A; ArpNbeta; Actin-related protein Baf53a; 53 kDa BRG1-associated factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131249 399.00
Check your price
  • Bcl-rambo Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125847
  • Size 50µg
Applications
Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Protein Mil1; Bcl-rambo
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125847 399.00
Check your price
  • beta Amyloid Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA125493
  • Size 50µg
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125493 399.00
Check your price
  • beta Amyloid Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134553
  • Size 50µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (Assay Dependent), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134553 399.00
Check your price
  • BLNK Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131493
  • Size 50µg
Applications
Western Blot (3-5 µg/ml)
Alternate names:
Src homology 2 domain-containing leukocyte protein of 65 kDa; Lymphocyte antigen 57; Cytoplasmic adapter protein; B-cell adapter containing a Src homology 2 domain protein; B-cell adapter containing a SH2 domain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131493 399.00
Check your price
  • BOB-1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131664
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
OCT-binding factor 1; OCA-B; BOB-1; B-cell-specific coactivator OBF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131664 399.00
Check your price
  • BPOZ Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131433
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Elongation factor 1A-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131433 399.00
Check your price
  • C1q Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (F), ELISA and FN
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA140312
  • Size 50µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140312 399.00
Check your price
  • CABP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131613
  • Size 50µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Caldendrin; Calbrain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131613 399.00
Check your price
  • Calprotectin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA140314
  • Size 50µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Urinary stone protein band A; S100 calcium-binding protein A8; Migration inhibitory factor-related protein 8; Leukocyte L1 complex light chain; Cystic fibrosis antigen; Calprotectin L1L subunit; Calgranulin-A; S100 calcium-binding protein A9; Migration inhibitory factor-related protein 14; Leukocyte L1 complex heavy chain; Calprotectin L1H subunit; Calgranulin-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140314 399.00
Check your price
  • Caspase 7 (active) Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126433
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (10-20 µg/ml), Immunoprecipitation (10-20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cysteine protease LICE2; Apoptotic protease Mch-3; ICE-like apoptotic protease 3; CMH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126433 399.00
Check your price
  • Caspase 9 (active) Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126434
  • Size 50µg
Applications
Immunocytochemistry (10-20 µg/ml), Immunofluorescence (10-20 ug/ml), Immunoprecipitation (10-20 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126434 399.00
Check your price
  • CD180 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA140035
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent.)
Alternate names:
Radioprotective 105 kDa protein; Lymphocyte antigen 78
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140035 399.00
Check your price
  • Cdc27 Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132473
  • Size 50µg
Applications
ELISA (1:5000-1:40,000), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (1:500-1:2000)
Alternate names:
H-NUC; CDC27 homolog; Anaphase-promoting complex subunit 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132473 399.00
Check your price
  • CENPA Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Neu
  • Host Mouse
  • Target Species Chicken, Human
  • Conjugate Unconjugated
  • Catalog # MA120832
  • Size 50µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Neutralization (Assay-dependent), Western Blot (1 µg/ml)
Alternate names:
Centromere protein A; Centromere autoantigen A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120832 399.00
Check your price
  • cGKI Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128083
  • Size 50µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (1:800), Immunoprecipitation (4 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
cGMP-dependent protein kinase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128083 399.00
Check your price
  • CTGF Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA122376
  • Size 50µg
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:5000)
Alternate names:
Insulin-like growth factor-binding protein 8; Hypertrophic chondrocyte-specific protein 24; CCN family member 2; Connective tissue growth-related protein; Protein FISP-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122376 399.00
Check your price
  • CXCR7 Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Rabbit
  • Target Species Cat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA129377
  • Size 50µg
Applications
Immunohistochemistry (Frozen) (1:200), Immunoprecipitation (1:250), Western Blot (1:500)
Alternate names:
G-protein coupled receptor RDC1 homolog; G-protein coupled receptor 159; Chemokine orphan receptor 1; C-X-C chemokine receptor type 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129377 399.00
Check your price
  • DDX5 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131019
  • Size 50µg
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (0.1-0.3 µg/ml)
Alternate names:
RNA helicase p68; DEAD box protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131019 399.00
Check your price
  • DIAPH2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131027
  • Size 50µg
Applications
Western Blot (Assay dependent)
Alternate names:
Diaphanous-related formin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131027 399.00
Check your price
  • DKK4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131527
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
DKK-4, MGC129562, MGC129563
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131527 399.00
Check your price
  • DLL4 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186897
  • Size 50µg
Applications
ELISA (1:3000-1:15,000), Immunohistochemistry (Paraffin) (5-15 ug/ml), Western Blot (1:300-1:1500)
Alternate names:
Drosophila Delta homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186897 399.00
Check your price
  • DOCK1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131461
  • Size 50µg
Applications
Western Blot (3-4 µg/ml)
Alternate names:
180 kDa protein downstream of CRK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131461 399.00
Check your price