Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
4081 - 4110 of 48882 products displayed
  • GRB2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131495
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
SH2/SH3 adapter GRB2; Protein Ash; Adapter protein GRB2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131495 399.00
Check your price
  • HCCS Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA126475
  • Size 50µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (5-10 µg/ml)
Alternate names:
Holocytochrome c-type synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126475 399.00
Check your price
  • Hepatitis C Virus Core Antigen Monoclonal Antibody, FITC conjugate for IF, ICC, IHC (F) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA190321
  • Size 50µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
HepC, HepC core, HCV core
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190321 399.00
Check your price
  • HLTF Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131579
  • Size 50µg
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Sucrose nonfermenting protein 2-like 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; RING finger protein 80; HIP116; DNA-binding protein/plasminogen activator inhibitor 1 regulator; TNF-response element-binding protein; P113
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131579 399.00
Check your price
  • HMOX1 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), ELISA and IA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125475
  • Size 50µg
Applications
ELISA (1:10,000), Immunohistochemistry (Frozen) (5-10 µg/ml), Immunohistochemistry (Paraffin) (5-10 µg/ml), Inhibition Assays (Assay Dependent), Western Blot (5-10 µg/ml)
Alternate names:
P32 protein; HSP32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125475 399.00
Check your price
  • hnRNP H1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA133147
  • Size 50µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA133147 399.00
Check your price
  • HSP20 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Cat, Guinea pig, Human, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA129447
  • Size 50µg
Applications
Western Blot (0.05 µg/ml)
Alternate names:
Heat shock 20 kDa-like protein p20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129447 399.00
Check your price
  • HSPB8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131842
  • Size 50µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Small stress protein-like protein HSP22; Protein kinase H11; E2-induced gene 1 protein; Alpha-crystallin C chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131842 399.00
Check your price
  • IL-1RAcP Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129969
  • Size 50µg
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Interleukin-1 receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129969 399.00
Check your price
  • IL-22 Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121357
  • Size 50µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121357 399.00
Check your price
  • IL-22 Polyclonal Antibody for IF, ICC, IHC (P) and ELISA
  • Host Goat
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121356
  • Size 50µg
Applications
ELISA (1:100,000), Immunocytochemistry (1:300), Immunofluorescence (1:300), Immunohistochemistry (Paraffin) (Assay-dependent)
Alternate names:
Interleukin-22a; IL-TIF alpha; IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121356 399.00
Check your price
  • IL21R Polyclonal Antibody for IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120730
  • Size 50µg
Applications
ELISA (1:100,000), Immunohistochemistry (Paraffin) (1:300)
Alternate names:
Novel interleukin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120730 399.00
Check your price
  • IRF2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131652
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
DKFZp686F0244, IRF-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131652 399.00
Check your price
  • IRF8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131665
  • Size 50µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Interferon consensus sequence-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131665 399.00
Check your price
  • JNK2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA121411
  • Size 50µg
Applications
ELISA (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121411 399.00
Check your price
  • KIN Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124441
  • Size 50µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (0.2-5 µg/ml)
Alternate names:
KIN, antigenic determinant of recA protein homolog; Binding to curved DNA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124441 399.00
Check your price
  • KLF16 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131564
  • Size 50µg
Applications
Western Blot (2-4 µg/ml)
Alternate names:
Transcription factor NSLP2; Transcription factor BTEB4; Novel Sp1-like zinc finger transcription factor 2; Basic transcription element-binding protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131564 399.00
Check your price
  • LIMK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131026
  • Size 50µg
Applications
Western Blot (1:500)
Alternate names:
KIZ-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131026 399.00
Check your price
  • LIS1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131561
  • Size 50µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
PAF-AH alpha; PAF acetylhydrolase 45 kDa subunit; Lissencephaly-1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131561 399.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot, IHC (F), Flow and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA184920
  • Size 50µg
Applications
Flow Cytometry (1:10), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Neat), Western Blot (1:10-1:100)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184920 399.00
Check your price
  • MAD2L2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131766
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500)
Alternate names:
REV7 homolog; Mitotic arrest deficient 2-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131766 399.00
Check your price
  • MAGE1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129668
  • Size 50µg
Applications
ELISA (0.1-1 µg/ml), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (3-5 µg x 10^7 cells), Western Blot (0.5-2 µg/ml)
Alternate names:
MAGE-1 antigen; Cancer/testis antigen 1.1; Antigen MZ2-E
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129668 399.00
Check your price
  • MAX Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Human
  • Conjugate Unconjugated
  • Catalog # PA132194
  • Size 50µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Myc-associated factor X; Class D basic helix-loop-helix protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132194 399.00
Check your price
  • MBD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129386
  • Size 50µg
Applications
Western Blot (0.625 µg/ml)
Alternate names:
Methyl-CpG-binding protein MBD2; Demethylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129386 399.00
Check your price
  • MD1 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA140041
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent.)
Alternate names:
Protein MD-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140041 399.00
Check your price
  • Metallothionein Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Horse, Fish, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA125479
  • Size 50µg
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1 µg/ml)
Alternate names:
Metallothionein-I; Metallothionein-IA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125479 399.00
Check your price
  • MLH1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190218
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (9-11 µg/ml), Western Blot (1-1.5 µg/ml)
Alternate names:
MutL protein homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190218 399.00
Check your price
  • MMP2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA122527
  • Size 50µg
Applications
Immunoprecipitation (1:500), Western Blot (1:2000)
Alternate names:
Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122527 399.00
Check your price
  • MST4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131507
  • Size 50µg
Applications
Western Blot (0.5-1.5 µg/ml)
Alternate names:
Serine/threonine-protein kinase MASK; Mammalian STE20-like protein kinase 4; MST3 and SOK1-related kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131507 399.00
Check your price
  • Nestin Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IM
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191657
  • Size 50µg
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Frozen) (1:10-1:100), Immunohistochemistry (Paraffin) (1:10-1:100), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191657 399.00
Check your price