Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
4141 - 4170 of 48881 products displayed
  • gp210 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA129888
  • Size 50µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
Pore membrane protein of 210 kDa; Nucleoporin Nup210; Nuclear envelope pore membrane protein POM 210
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129888 399.00
Check your price
  • HIV1 p24 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14951
  • Size 50µL
Applications
ELISA (Assay dependent), Western Blot (1/2000-1/5000)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14951 399.00
Check your price
  • HIV1 Reverse Transcriptase Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14948
  • Size 50µL
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Human immunodeficiency virus, HIV RT
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14948 399.00
Check your price
  • HIV2 Nef Monoclonal Antibody for Western Blot and IP
  • Host Rat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA517015
  • Size 50µL
Applications
Immunoprecipitation (Assay-Dependent), Western Blot (1:1000-1:5000)
Alternate names:
F-protein, HIV2 nef protein, Human immunodeficiency virus 2 nef, Nefprotein, Negative factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517015 399.00
Check your price
  • hnRNP A2B1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA134789
  • Size 50µL
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134789 399.00
Check your price
  • HnRNP Q Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190980
  • Size 50µL
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Synaptotagmin-binding, cytoplasmic RNA-interacting protein; NS1-associated protein 1; Glycine- and tyrosine-rich RNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190980 399.00
Check your price
  • HSP25 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Guinea pig, Hamster, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA125494
  • Size 50µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Heat shock 27 kDa protein; Heat shock 25 kDa protein; Growth-related 25 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125494 399.00
Check your price
  • IkB beta Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132136
  • Size 50µL
Applications
ELISA (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
I-kappa-B-beta; Thyroid receptor-interacting protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132136 399.00
Check your price
  • MAP2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Chicken
  • Target Species Bovine, Chicken, Horse, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA110005
  • Size 50µL
Applications
Immunocytochemistry (1:10,000-1:20,000), Immunofluorescence (1:10,000-1:20,000), Immunohistochemistry (1:10,000-1:20,000), Western Blot (1:20,000)
Alternate names:
Microtubule Associated Protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110005 399.00
Check your price
  • MCPH1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518057
  • Size 50µL
Applications
ELISA (Assay-dependent), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay-dependent), Western Blot (1:1000)
Alternate names:
Microcephalin; FLJ12847; MCT
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518057 399.00
Check your price
  • MDC1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120205
  • Size 50µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Western Blot (1:2000)
Alternate names:
Nuclear factor with BRCT domains 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120205 399.00
Check your price
  • MUS81 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15837
  • Size 50µL
Applications
Immunoprecipitation (1:10000), Western Blot (1:10000)
Alternate names:
FLJ21012, FLJ44872 MUS81
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15837 399.00
Check your price
  • NCoR2 Polyclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123286
  • Size 50µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Thyroid-, retinoic-acid-receptor-associated corepressor; T3 receptor-associating factor; Silencing mediator of retinoic acid and thyroid hormone receptor; SMAP270; CTG repeat protein 26
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123286 399.00
Check your price
  • Nesprin 1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130283
  • Size 50µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (1:25-1:50), Immunohistochemistry (Frozen) (1:25-1:50), Western Blot (1:500-1:750)
Alternate names:
Synaptic nuclear envelope protein 1; Nuclear envelope spectrin repeat protein 1; Myocyte nuclear envelope protein 1; Enaptin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130283 399.00
Check your price
  • NFkB p65 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA124691
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124691 399.00
Check your price
  • OPRD1 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA127970
  • Size 50µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000)
Alternate names:
Opioid receptor A; MSL-2; K56
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127970 399.00
Check your price
  • p130 Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132114
  • Size 50µL
Applications
ELISA (1:500-1:2000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:1000)
Alternate names:
pRb2; Retinoblastoma-related protein 2; PPAR-alpha-interacting complex protein 128; 130 kDa retinoblastoma-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132114 399.00
Check your price
  • P2X1 Polyclonal Antibody for IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124622
  • Size 50µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Frozen) (1:1000)
Alternate names:
RP-2 protein; Purinergic receptor; ATP receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124622 399.00
Check your price
  • PABP Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Rabbit, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA134079
  • Size 50µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Cytoplasmic poly(A)-binding protein 1-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134079 399.00
Check your price
  • PGP9.5 (soluble) Polyclonal Antibody for IF, ICC and IHC (F)
  • Host Guinea Pig
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA124674
  • Size 50µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:500)
Alternate names:
Ubiquitin thioesterase L1; PGP 9.5; Neuron cytoplasmic protein 9.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124674 399.00
Check your price
  • PGP9.5 Polyclonal Antibody for Western Blot, IF and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA120158
  • Size 50µL
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:2000), Western Blot (1:1000)
Alternate names:
Ubiquitin thioesterase L1; PGP 9.5; Neuron cytoplasmic protein 9.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120158 399.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126727
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
CAMKA, KIAA0968
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126727 399.00
Check your price
  • Phospho-Progesterone Receptor (Ser294) Polyclonal Antibody for Western Blot and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125647
  • Size 50µL
Applications
Dot blot (1:1000-1:2000), Western Blot (1:1000-1:2000)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125647 399.00
Check your price
  • Prodynorphin Polyclonal Antibody for IF, ICC and IHC
  • Host Guinea Pig
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA130935
  • Size 50µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130935 399.00
Check your price
  • RNA polymerase Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA125426
  • Size 50µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
RNA Pol sigma 70
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125426 399.00
Check your price
  • TAP1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA128108
  • Size 50µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay-dependent), Immunoprecipitation (1:100), Western Blot (1:4000)
Alternate names:
Really interesting new gene 4 protein; Peptide transporter involved in antigen processing 1; Peptide transporter TAP1; Peptide transporter PSF1; Peptide supply factor 1; ATP-binding cassette sub-family B member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128108 399.00
Check your price
  • Tapasin Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129450
  • Size 50µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
TAP-binding protein; TAP-associated protein; NGS-17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129450 399.00
Check your price
  • TBP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA125427
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Transcription initiation factor TFIID TBP subunit; TATA-box factor; TATA-binding factor; TATA sequence-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125427 399.00
Check your price
  • VAMP1 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA126000
  • Size 50µL
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (1:100), Western Blot (1:2000)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126000 399.00
Check your price
  • Calcitonin Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Dog, Horse, Human, Mouse, Non-human primate, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA190591
  • Size 6mL
Applications
Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190591 399.00
Check your price