Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  170  180  190  200  210  220  230  240  300  400  500  600  700  800  900  1000  1100  1630 
4381 - 4410 of 48881 products displayed
  • alpha Actinin 4 Monoclonal Antibody for Western Blot, IF, IHC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1800
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1µg per 10^6 cells), Immunofluorescence (4 ug/mL), Immunohistochemistry (1:20), Immunohistochemistry (Paraffin) (4 µg/ml), Western Blot (2 ug/mL)
Alternate names:
Non-muscle alpha-actinin 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1800 375.00
Check your price
  • alpha COP Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1067
  • Size 100µg
Applications
Immunocytochemistry (3 µg/ml), Immunofluorescence (3 ug/ml), Western Blot (1 µg/ml)
Alternate names:
HEP-COP; Alpha-coat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1067 375.00
Check your price
  • alpha Synuclein Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118256
  • Size 100µg
Applications
Immunohistochemistry (Paraffin, Frozen) (3 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118256 375.00
Check your price
  • alpha-2c Adrenergic Receptor Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14518
  • Size 100µg
Applications
Immunohistochemistry (1:100-1:300), Western Blot (1:500-1:1000)
Alternate names:
Alpha-2C adrenoreceptor; Alpha-2 adrenergic receptor subtype C4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14518 375.00
Check your price
  • ALPL Monoclonal Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA135004
  • Size 100µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135004 375.00
Check your price
  • ALPL Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547419
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547419 375.00
Check your price
  • Androgen Receptor Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1111B
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (2 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1111B 375.00
Check your price
  • Angiopoietin 1 Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA546968
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
AGP1, AGPT, ANG1, ANG-1, angiopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546968 375.00
Check your price
  • ANGPTL4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11053
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Secreted protein Bk89; Hepatic fibrinogen/angiopoietin-related protein; Fasting-induced adipose factor; Angiopoietin-like protein 4; 425O18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11053 375.00
Check your price
  • Annexin A4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547586
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Annexin-4; Annexin IV; Protein II; Placental anticoagulant protein II; PP4-X; P32.5; Lipocortin IV; Endonexin I; Chromobindin-4; Carbohydrate-binding protein p33/p41; 35-beta calcimedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547586 375.00
Check your price
  • APC Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116631
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
Deleted in polyposis 2.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116631 375.00
Check your price
  • APC Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA126195
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Deleted in polyposis 2.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126195 375.00
Check your price
  • APH1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA12010
  • Size 100µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
Aph-1alpha; Presenilin-stabilization factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA12010 375.00
Check your price
  • Apolipoprotein A1 Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123058
  • Size 100µg
Applications
ELISA (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123058 375.00
Check your price
  • Apolipoprotein A1 Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA122558
  • Size 100µg
Applications
ELISA (1:10,000-1:20,000), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122558 375.00
Check your price
  • Apolipoprotein A1 Polyclonal Antibody for Western Blot, ELISA and IDF
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA123059
  • Size 100µg
Applications
ELISA (Assay dependent), Immunodiffusion (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123059 375.00
Check your price
  • Apolipoprotein A2 Polyclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126900
  • Size 100µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Apolipoprotein A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126900 375.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112800
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112800 375.00
Check your price
  • Apolipoprotein J Polyclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126903
  • Size 100µg
Applications
ELISA (1:5000-1:10,000), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Testosterone-repressed prostate message 2; NA1/NA2; Ku70-binding protein 1; Complement-associated protein SP-40,40; Complement cytolysis inhibitor; Apolipoprotein J; Aging-associated gene 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126903 375.00
Check your price
  • ASAH2 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547509
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Non-lysosomal ceramidase; N-acylsphingosine amidohydrolase 2; LCDase; BCDase; Acylsphingosine deacylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547509 375.00
Check your price
  • ASGR1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547385
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Hepatic lectin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547385 375.00
Check your price
  • ATP Synthase beta Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1930
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (2-5 ug), Western Blot (1-2 µg/ml)
Alternate names:
ATP5B, ATPMB, ATPSB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1930 375.00
Check your price
  • ATP1A1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3929
  • Size 100µg
Applications
Flow Cytometry (1/20-1/100), Immunocytochemistry (1/50-1/200), Immunofluorescence (1/50-1/200), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1/100-1/2000), Western Blot (1:200-1:2000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3929 375.00
Check your price
  • ATP1A3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Guinea pig, Human, Mouse, Non-human primate, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3915
  • Size 100µg
Applications
Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (5 µg/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (Frozen) (3 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Sodium pump subunit alpha-3; Na(+)/K(+) ATPase alpha(III) subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3915 375.00
Check your price
  • ATP1B1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Sheep, Pig
  • Conjugate Unconjugated
  • Catalog # MA3930
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:1000), Immunohistochemistry (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:10,000)
Alternate names:
Sodium/potassium-dependent ATPase subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3930 375.00
Check your price
  • ATR Monoclonal Antibody for Western Blot, IF, ICC, IP and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123158
  • Size 100µg
Applications
Functional Assay (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1000)
Alternate names:
FRAP-related protein 1; Ataxia telangiectasia and Rad3-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123158 375.00
Check your price
  • B7-H3 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547132
  • Size 100µg
Applications
Neutralization (0.15-0.6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Costimulatory molecule; B7 homolog 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547132 375.00
Check your price
  • BACE1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112529
  • Size 100µg
Applications
Western Blot (0.5 -1 µg/ml)
Alternate names:
Membrane-associated aspartic protease 2; Memapsin-2; Beta-site amyloid precursor protein cleaving enzyme 1; Aspartyl protease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112529 375.00
Check your price
  • BACE1 Polyclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1757
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunoprecipitation (Assay dependent), Western Blot (1:200-1:2000)
Alternate names:
Membrane-associated aspartic protease 2; Memapsin-2; Beta-site amyloid precursor protein cleaving enzyme 1; Aspartyl protease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1757 375.00
Check your price
  • BACE2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112530
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Theta-secretase; Membrane-associated aspartic protease 1; Memapsin-1; Beta-site amyloid precursor protein cleaving enzyme 2; Aspartyl protease 1; Down region aspartic protease; Aspartic-like protease 56 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112530 375.00
Check your price