Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  50  60  70  80  90  100  110  120  130  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1630 
4711 - 4740 of 48882 products displayed
  • IL-21 Polyclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546962
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL-21, Za11, interleukin-21, interleukin-21 isoform, CX2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546962 375.00
Check your price
  • IL-26 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546973
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Protein AK155
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546973 375.00
Check your price
  • IL-27 p28 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546981
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (1-5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546981 375.00
Check your price
  • IL-27 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546993
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p28; Interleukin-30; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546993 375.00
Check your price
  • IL-3 Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546918
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.05-0.25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
P-cell-stimulating factor; Multipotential colony-stimulating factor; Mast cell growth factor; Hematopoietic growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546918 375.00
Check your price
  • IL-33 Polyclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547006
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (0.75-3.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Nuclear factor from high endothelial venules; Interleukin-1 family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547006 375.00
Check your price
  • IL-7 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546944
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.2- 0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-7, IL7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546944 375.00
Check your price
  • IL10RB Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547727
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
CDW210B, CRF2-4, CRFB4, D21S58, D21S66, IL-10R2, IL-10 receptor subunit beta, IL-10R subunit 2, IL-10R subunit beta, IL-10RB, cytokine receptor class-II CRF2-4, cytokine receptor class-II member 4, cytokine receptor family 2 member 4, cytokine receptor family II, member 4, interleukin-10 receptor subunit 2, inte
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547727 375.00
Check your price
  • IL13RA2 Polyclonal Antibody for Western Blot, IHC (P), Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546976
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.2-10 µg/ml)
Alternate names:
Interleukin-13-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546976 375.00
Check your price
  • IL17RB Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547093
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-17B receptor; IL-17 receptor homolog 1; Cytokine receptor-like 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547093 375.00
Check your price
  • IL1F9 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546988
  • Size 100µg
Applications
Blocking Assay (10 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Interleukin-1 homolog 1; Interleukin-1 family member 9; Interleukin-1 epsilon; IL-1-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546988 375.00
Check your price
  • IL1R1 Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546930
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.5-2.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p80; Interleukin-1 receptor type I; Interleukin-1 receptor alpha; CD121 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546930 375.00
Check your price
  • IL1RA Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Horse
  • Conjugate Unconjugated
  • Catalog # PA546924
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL1 inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546924 375.00
Check your price
  • IL20RA Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141167
  • Size 100µg
Applications
Flow Cytometry (2-5 µg/10^6 cells), Western Blot (1-3 µg/ml)
Alternate names:
ZcytoR7; IL-20R1; Cytokine receptor family 2 member 8; Cytokine receptor class-II member 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141167 375.00
Check your price
  • Ileal Fatty Acid Binding Protein (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75561
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Fatty Acid Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75561 375.00
Check your price
  • Importin beta-1 Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and IA
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3070
  • Size 100µg
Applications
Flow Cytometry (1 ug/test), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Immunoprecipitation (Assay dependent), Inhibition Assays (Assay Dependent), Western Blot (1:5,000)
Alternate names:
Pore targeting complex 97 kDa subunit; Nuclear factor p97; Karyopherin subunit beta-1; Importin-90; SCG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3070 375.00
Check your price
  • Influenza A H6N1 MP1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA141124
  • Size 100µg
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (0.5-2 µg/ml)
Alternate names:
Influenza H6N1, H6N1 MP1, H6N1 Major Protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141124 375.00
Check your price
  • Influenza A HA Monoclonal Antibody for Western Blot, IF, ICC, IHC, ELISA, HA and IA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122684
  • Size 100µg
Applications
ELISA (Assay dependent), Hemagglutination Assay (Assay Dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay-Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H1N1, H1N1 HA, H1N1 hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122684 375.00
Check your price
  • Influenza A M2 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA1082
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 ug/ml), Immunoprecipitation (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Influenza Matrix protein M2, Influenza M2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1082 375.00
Check your price
  • Influenza A NP Monoclonal Antibody, FITC conjugate for IF, Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17322
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17322 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17323
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17323 375.00
Check your price
  • Influenza A NP Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA126958
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126958 375.00
Check your price
  • Influenza A NP Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141071
  • Size 100µg
Applications
ELISA (1:100-1:2000), Western Blot (1-3 µg/ml)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141071 375.00
Check your price
  • Influenza B NP Monoclonal Antibody, FITC conjugate for IF and ICC
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17306
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:5-1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17306 375.00
Check your price
  • INHBA Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547004
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (2-6 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Activin beta-A chain; Erythroid differentiation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547004 375.00
Check your price
  • Insulin Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA135525
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135525 375.00
Check your price
  • Insulin/Proinsulin Monoclonal Antibody, Biotin conjugate for IHC (F) and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA127288
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Ins1, Ins2, ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127288 375.00
Check your price
  • IRAK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130549
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
IRAK-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130549 375.00
Check your price
  • IRF7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112810
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
IRF-7H, IRF7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112810 375.00
Check your price
  • IRF7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112811
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
IRF-7H, IRF7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112811 375.00
Check your price