Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1630 
4741 - 4770 of 48882 products displayed
  • ITGA3 Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127165
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127165 375.00
Check your price
  • JAB1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Mouse
  • Target Species Fruit fly, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA123244
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Immunoprecipitation (0.5-1 µg/ml), Western Blot (1:1000)
Alternate names:
Jun activation domain-binding protein 1; Kip1 C-terminus-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123244 375.00
Check your price
  • Jagged1 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546970
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
protein jagged-1, JAG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546970 375.00
Check your price
  • Kallikrein 7 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546998
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Stratum corneum chymotryptic enzyme; Serine protease 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546998 375.00
Check your price
  • KCNMB1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1924
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Western Blot (2.0 µg/ml)
Alternate names:
Slo-beta-1; Maxi K channel subunit beta-1; K(VCA)beta-1; Charybdotoxin receptor subunit beta-1; Calcium-activated potassium channel, subfamily M subunit beta-1; BK channel subunit beta-1; Slowpoke-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1924 375.00
Check your price
  • KIDINS220 Monoclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA190667
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Ankyrin repeat-rich membrane-spanning protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190667 375.00
Check your price
  • KIF5A Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1642
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Neuronal kinesin heavy chain; Kinesin heavy chain neuron-specific 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1642 375.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1643
  • Size 100µg
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunoprecipitation (5 µg), Western Blot (1:4000-1:6000 )
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1643 375.00
Check your price
  • KIF5C Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1644
  • Size 100µg
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Kinesin heavy chain neuron-specific 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1644 375.00
Check your price
  • KSV Cyclin D Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA191431
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Kaposi's Sarcoma Virus Type D Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191431 375.00
Check your price
  • L-Myc Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547567
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
V-myc myelocytomatosis viral oncogene homolog; Protein L-Myc-1; Class E basic helix-loop-helix protein 38
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547567 375.00
Check your price
  • LALBA Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191042
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Lysozyme-like protein 7; Lactose synthase B protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191042 375.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA106102
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, PRO1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106102 375.00
Check your price
  • LAMP1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1654A
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (1:250-1:1000)
Alternate names:
CD107 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1654A 375.00
Check your price
  • Langerin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547250
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Langerin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547250 375.00
Check your price
  • Langerin Polyclonal Antibody for Western Blot, IHC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA141053
  • Size 100µg
Applications
Flow Cytometry (1:10-1:1000), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:500), Western Blot (1-3 µg/ml)
Alternate names:
Langerin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141053 375.00
Check your price
  • LDLR Polyclonal Antibody for Western Blot, IHC (F), Flow and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546987
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
FH, FHC, LDLCQ2, LDL receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546987 375.00
Check your price
  • LDLR Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546985
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
FH, FHC, LDLCQ2, LDL receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546985 375.00
Check your price
  • Leptin Polyclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547023
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.05-0.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Obesity factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547023 375.00
Check your price
  • LGR7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141532
  • Size 100µg
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Relaxin family peptide receptor 1; Leucine-rich repeat-containing G-protein coupled receptor 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141532 375.00
Check your price
  • LILRB2 Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546983
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.3-1.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
CD85d, ILT4, LIR2, MIR10, CD85 antigen-like family member D, CD85d antigen, Ig-like transcript 4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546983 375.00
Check your price
  • LPL Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547033
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
HDLCQ11, LIPD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547033 375.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127198
  • Size 100µg
Applications
Blocking Assay (Assay-dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin, Frozen) (1-10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127198 375.00
Check your price
  • LRP6 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547145
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547145 375.00
Check your price
  • LTBR Polyclonal Antibody for Western Blot, Flow and IA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547028
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Inhibition Assays (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Tumor necrosis factor receptor type III; Tumor necrosis factor receptor 2-related protein; Tumor necrosis factor C receptor; Lymphotoxin-beta receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547028 375.00
Check your price
  • Lubricin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3118
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Superficial zone proteoglycan; Megakaryocyte-stimulating factor; Lubricin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3118 375.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA1332
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1332 375.00
Check your price
  • M-cadherin Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547578
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Muscle cadherin; Cadherin-14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547578 375.00
Check your price
  • Macrophages / Monocytes Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Dog, Deer, Horse, Cat, Guinea pig, Goat, Human, Non-human primate, Pig, Rabbit, Rat
  • Conjugate FITC
  • Catalog # MA180263
  • Size 100µg
Applications
Flow Cytometry (Neat)
Alternate names:
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180263 375.00
Check your price
  • MAdCAM1 (soluble) Monoclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191737
  • Size 100µg
Applications
ELISA (Assay-dependent), Flow Cytometry (1:10-1:100 (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (Assay dependent)
Alternate names:
MACAM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191737 375.00
Check your price