Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1605 
4771 - 4800 of 48136 products displayed
  • GnRH1 Antibody
  • Host Rabbit
  • Target Species Amphibian, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1121
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:200-1:1000)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1121 375.00
Check your price
  • GPNMB Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546926
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Transmembrane glycoprotein HGFIN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546926 375.00
Check your price
  • GPR77 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141397
  • Size 100 µg
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
G-protein coupled receptor 77; Complement component 5a receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141397 375.00
Check your price
  • GRAF Antibody
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131264
  • Size 100 µg
Applications
ELISA (Assay-dependent), Western Blot (0.3-1 µg/ml)
Alternate names:
Rho-type GTPase-activating protein 26; Oligophrenin-1-like protein; GTPase regulator associated with focal adhesion kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131264 375.00
Check your price
  • Gram Positive Bacteria LTA Antibody (Monoclonal, G35C)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17401
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Lipoteichoic acid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17401 375.00
Check your price
  • Granzyme B Antibody (Monoclonal, GB11)
  • Host Mouse
  • Target Species Human, Non-human primate, Rhesus monkey
  • Conjugate Unconjugated
  • Catalog # MA180734
  • Size 100 µg
Applications
ELISA (2ug/ml-5ug/ml), Flow Cytometry (1/10-1/100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
T-cell serine protease 1-3E; SECT; Human lymphocyte protein; Granzyme-2; Fragmentin-2; Cytotoxic T-lymphocyte proteinase 2; Cathepsin G-like 1; CTLA-1; C11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180734 375.00
Check your price
  • Granzyme B Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547214
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (3.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Fragmentin-2; Cytotoxic cell protease 1; CTLA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547214 375.00
Check your price
  • GREM1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141012
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (2.5-10 µg/ml)
Alternate names:
Increased in high glucose protein 2; Down-regulated in Mos-transformed cells protein; DAN domain family member 2; Cysteine knot superfamily 1, BMP antagonist 1; Cell proliferation-inducing gene 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141012 375.00
Check your price
  • GRK1 Antibody (Monoclonal, G8)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1720
  • Size 100 µg
Applications
Immunocytochemistry (2-3 µg/ml), Immunofluorescence (2-3 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
G protein-coupled receptor kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1720 375.00
Check your price
  • Growth Hormone Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546911
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Growth hormone
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546911 375.00
Check your price
  • H-cadherin Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547472
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Truncated cadherin; P105; Heart cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547472 375.00
Check your price
  • HA Tag Antibody (Monoclonal, 12CA5)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA112429
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112429 375.00
Check your price
  • hCdc4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123468
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
hCdc4; SEL-10; F-box protein FBX30; F-box and WD-40 domain-containing protein 7; Archipelago homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123468 375.00
Check your price
  • hCV-2 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546982
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Protein crossveinless-2; Bone morphogenetic protein-binding endothelial cell precursor-derived regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546982 375.00
Check your price
  • HDAC1 Antibody
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1860
  • Size 100 µg
Applications
ChIP assay (Assay dependent), Immunocytochemistry (2.0 µg/mL), Immunofluorescence (Assay dependent), Immunohistochemistry (1:200), Immunohistochemistry (Frozen) (0.2 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Histone Deacetylase 1, DKFZp686H12203, GON-10, HD1, RPD3, RPD3L1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1860 375.00
Check your price
  • HDAC11 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141123
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
FLJ22237, HD11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141123 375.00
Check your price
  • HDAC2 Antibody
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1861
  • Size 100 µg
Applications
ChIP assay (1-3 ul), Immunocytochemistry (1.0 µg/ml), Immunofluorescence (1:50-1:200), Immunohistochemistry (0.4 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (0.5 µg/ml)
Alternate names:
YY1 transcription factor-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1861 375.00
Check your price
  • HDAC4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141092
  • Size 100 µg
Applications
ChIP assay (Assay Dependent.), Immunoprecipitation (Assay-Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
HA6116, HD4, HDAC-A, HDACA, KIAA0288
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141092 375.00
Check your price
  • HDAC5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141117
  • Size 100 µg
Applications
ChIP assay (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1-3 µg/ml)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141117 375.00
Check your price
  • HDAC6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141056
  • Size 100 µg
Applications
ChIP assay (1:10-1:500), Immunoprecipitation (Assay-Dependent), Western Blot (2-5 µg/ml)
Alternate names:
Histone deacetylase mHDA2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141056 375.00
Check your price
  • Helicobacter pylori 19 kDa Antibody (Monoclonal, F92B)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17455
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
H. pylori, Campylobacter pyloridis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17455 375.00
Check your price
  • Helicobacter pylori 54 kDa Antibody (Monoclonal, G56H)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17457
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
H. pylori, Campylobacter pyloridis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17457 375.00
Check your price
  • Hepatitis A Virus Vp3 Antibody (Monoclonal, C22H)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17371
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50)
Alternate names:
HepA, HAVgp1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17371 375.00
Check your price
  • Hepatitis B Virus E Antibody (Monoclonal, H06A)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17374
  • Size 100 µg
Applications
ELISA (1:20-1:200)
Alternate names:
HepB, HBVgp4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17374 375.00
Check your price
  • Hepatitis B Virus preS2 Antibody (Monoclonal, J68F)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17377
  • Size 100 µg
Applications
ELISA (1:20), Immunofluorescence (1:10), Western Blot (1:10)
Alternate names:
HepB, HBVgp4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17377 375.00
Check your price
  • Hepatitis B Virus X Antibody (Monoclonal, X36C)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA1081
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
HBV x Core Protein, HBVgp3, HepB X
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1081 375.00
Check your price
  • Hepatitis C Virus Core Antigen Antibody (Monoclonal, C7-50)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA1080
  • Size 100 µg
Applications
ELISA (Assay dependent), Flow Cytometry (1/100), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
HepC, HepC core, HCV core
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1080 375.00
Check your price
  • Hepatitis C Virus Core Antigen Antibody (Monoclonal, D95B)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17366
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
HepC, HepC core, HCV core
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17366 375.00
Check your price
  • Hepatitis C Virus Core Antigen Antibody (Monoclonal, F66G)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17356
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
HepC, HepC core, HCV core
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17356 375.00
Check your price
  • Hepatitis C Virus E1 Antibody (Monoclonal, E72J)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17370
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
HepC, HCV2a Envelope glycoprotein E1, HCV E1, HCV genotype 2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17370 375.00
Check your price