Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1630 
5011 - 5040 of 48882 products displayed
  • TLR6 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112828
  • Size 100µg
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Toll-like Receptor 6, CD antigen 286
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112828 375.00
Check your price
  • TMPRSS15 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547162
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Transmembrane protease serine 15; Serine protease 7; Enterokinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547162 375.00
Check your price
  • TNF alpha Monoclonal Antibody for IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126970
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126970 375.00
Check your price
  • TNF alpha Polyclonal Antibody for Western Blot, IF, ICC, Flow, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546945
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546945 375.00
Check your price
  • TNFAIP6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547253
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tumor necrosis factor alpha-induced protein 6; TNF-stimulated gene 6 protein; Hyaluronate-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547253 375.00
Check your price
  • TNFRSF13C Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546904
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
BLyS receptor 3; BAFF receptor; B-cell-activating factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546904 375.00
Check your price
  • Toxoplasma gondii Monoclonal Antibody, FITC conjugate for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate FITC
  • Catalog # MA17436
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5), Western Blot (Assay Dependent)
Alternate names:
T. gondii
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17436 375.00
Check your price
  • TPX2 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1802
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (4 ug/mL), Immunohistochemistry (1:200), Immunohistochemistry (Paraffin) (0.5-1.0 ug/ml), Immunoprecipitation (Assay dependent), Western Blot (2 ug/mL)
Alternate names:
Restricted expression proliferation-associated protein 100; Protein fls353; Hepatocellular carcinoma-associated antigen 90; Hepatocellular carcinoma-associated antigen 519; Differentially expressed in cancerous and non-cancerous lung cells 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1802 375.00
Check your price
  • TR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124354
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Testicular receptor 2; Orphan nuclear receptor TR2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124354 375.00
Check your price
  • TRAG3 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112868
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CSAG3B, CT24.2, MGC149851, MGC149852, TRAG3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112868 375.00
Check your price
  • Transferrin Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120108
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
Siderophilin; Beta-1 metal-binding globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120108 375.00
Check your price
  • TRAP1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1010
  • Size 100µg
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Frozen) (1:20), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Tumor necrosis factor type 1 receptor-associated protein; TNFR-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1010 375.00
Check your price
  • TREM1 Polyclonal Antibody for Western Blot, Flow, ELISA and FN
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547112
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Functional Assay (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Triggering receptor expressed on monocytes 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547112 375.00
Check your price
  • TREM2 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546980
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.2 µg/ml)
Alternate names:
Triggering receptor expressed on monocytes 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546980 375.00
Check your price
  • TREM2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546978
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Triggering receptor expressed on monocytes 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546978 375.00
Check your price
  • TREML2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547471
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Triggering receptor expressed on myeloid cells-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547471 375.00
Check your price
  • TRIM28 Monoclonal Antibody for Western Blot, IF, ICC, IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA12023
  • Size 100µg
Applications
ChIP assay (Assay Dependent), Gel Shift (Assay Dependent), Immunocytochemistry (2-4 µg/ml), Immunofluorescence (2-4 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1 ug/ml)
Alternate names:
Tripartite motif-containing protein 28; RING finger protein 96; Nuclear corepressor KAP-1; KRAB-interacting protein 1; KRAB-associated protein 1; E3 SUMO-protein ligase TRIM28; KRIP-1; KRAB-A-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA12023 375.00
Check your price
  • TROP2 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547030
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Pancreatic carcinoma marker protein GA733-1; Membrane component chromosome 1 surface marker 1; Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547030 375.00
Check your price
  • TROP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547074
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547074 375.00
Check your price
  • TSG101 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA123296
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:3000)
Alternate names:
ESCRT-I complex subunit TSG101
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123296 375.00
Check your price
  • TSLP Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546975
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (0.05-0.25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
thymic stroma-derived lymphopoietin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546975 375.00
Check your price
  • TYROBP Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112701
  • Size 100µg
Applications
Immunoprecipitation (2-4µg/ml protein lysate), Western Blot (Assay Dependent)
Alternate names:
Killer-activating receptor-associated protein; DNAX-activation protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112701 375.00
Check your price
  • Ubiquilin 1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1759
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
1110046H03Rik, 1810030E05Rik, AU019746, C77538, D13Ertd372e, DA41, Dsk2, Plic-1, XDRP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1759 375.00
Check your price
  • UCP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1055
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Solute carrier family 25 member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1055 375.00
Check your price
  • UGT1A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547844
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
UDP-glucuronosyltransferase 1A1; UDP-glucuronosyltransferase 1-A; Bilirubin-specific UDPGT isozyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547844 375.00
Check your price
  • ULBP1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547131
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
UL16-binding protein 1; Retinoic acid early transcript 1I; ALCAN-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547131 375.00
Check your price
  • ULBP2/ULBP5/ULBP6 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547118
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
UL-16 binding protein 5; UL16-binding protein 2; Retinoic acid early transcript 1H; ALCAN-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547118 375.00
Check your price
  • ULBP3 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547148
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Retinoic acid early transcript 1N; ALCAN-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547148 375.00
Check your price
  • Uromodulin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546959
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (2-15 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Tamm-Horsfall urinary glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546959 375.00
Check your price
  • V5 Tag Monoclonal Antibody, FITC conjugate for IF
  • Host Mouse
  • Target Species Tag
  • Conjugate FITC
  • Catalog # MA180281
  • Size 100µg
Applications
Immunofluorescence (1/100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180281 375.00
Check your price