Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  190  200  210  220  230  240  250  260  270  300  400  500  600  700  800  900  1000  1100  1630 
5161 - 5190 of 48882 products displayed
  • Blimp-1 Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Pig
  • Conjugate DyLight® 650
  • Catalog # MA516114
  • Size 100µL
Applications
ELISA (1:100-1:2000), Flow Cytometry (8 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000)
Alternate names:
Positive regulatory domain I-binding factor 1; PR domain-containing protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP-1; B lymphocyte-induced maturation protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516114 375.00
Check your price
  • Blimp-1 Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Pig
  • Conjugate HRP
  • Catalog # MA516116
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Positive regulatory domain I-binding factor 1; PR domain-containing protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP-1; B lymphocyte-induced maturation protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516116 375.00
Check your price
  • BMI-1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116973
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (2 µg/ml)
Alternate names:
Polycomb group RING finger protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116973 375.00
Check your price
  • BMI-1 Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522844
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Polycomb group RING finger protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522844 375.00
Check your price
  • BMI-1 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522846
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Polycomb group RING finger protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522846 375.00
Check your price
  • BMI-1 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522848
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Polycomb group RING finger protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522848 375.00
Check your price
  • BMI-1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522845
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Polycomb group RING finger protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522845 375.00
Check your price
  • BMI-1 Polyclonal Antibody, HRP conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522847
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Polycomb group RING finger protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522847 375.00
Check your price
  • BNIP3 Polyclonal Antibody, Biotin conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA522859
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522859 375.00
Check your price
  • BNIP3 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # PA522861
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522861 375.00
Check your price
  • BNIP3 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # PA522863
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522863 375.00
Check your price
  • BNIP3 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # PA522860
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522860 375.00
Check your price
  • BNIP3 Polyclonal Antibody, HRP conjugate for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA522862
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
NIP3, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522862 375.00
Check your price
  • BRCA1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP, ChIP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123160
  • Size 100µl
Applications
ChIP assay (Assay-dependent), ELISA (Assay dependent), Immunocytochemistry (1 ug/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:3000)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123160 375.00
Check your price
  • BRCA1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123164
  • Size 100µl
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:400), Immunofluorescence (1:400), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123164 375.00
Check your price
  • BRCA1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA516050
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516050 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # MA516052
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516052 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # MA516054
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516054 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # MA516051
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516051 375.00
Check your price
  • BRCA1 Monoclonal Antibody, HRP conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA516053
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516053 375.00
Check your price
  • BRCA1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA516055
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516055 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # MA516057
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516057 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # MA516059
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516059 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # MA516056
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516056 375.00
Check your price
  • BRCA1 Monoclonal Antibody, HRP conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA516058
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516058 375.00
Check your price
  • BRCA1 Monoclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116617
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116617 375.00
Check your price
  • BRCA1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA516045
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516045 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # MA516047
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516047 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # MA516049
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516049 375.00
Check your price
  • BRCA1 Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # MA516046
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2-4 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516046 375.00
Check your price