Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  200  210  220  230  240  250  260  270  300  400  500  600  700  800  900  1000  1100  1611 
5221 - 5250 of 48310 products displayed
  • Advanced Glycated End Products Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186850
  • Size 100 µL
Applications
ELISA (1:4000), Western Blot (1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186850 375.00
Check your price
  • Afadin Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125076
  • Size 100 µL
Applications
Immunocytochemistry (1:250-1:500), Immunofluorescence (1:500-1:250), Western Blot (1:1000)
Alternate names:
Protein Af-6; ALL1-fused gene from chromosome 6 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125076 375.00
Check your price
  • Aflatoxin Antibody (Monoclonal, 6A10)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA116885
  • Size 100 µL
Applications
ELISA (5 µg/ml), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116885 375.00
Check your price
  • Aflatoxin Antibody, Biotin (Monoclonal, 6A10)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Biotin
  • Catalog # MA516119
  • Size 100 µL
Applications
ELISA (5 µg/ml), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516119 375.00
Check your price
  • Aflatoxin Antibody, DyLight® 488 (Monoclonal, 6A10)
  • Host Mouse
  • Target Species Chemical
  • Conjugate DyLight® 488
  • Catalog # MA516121
  • Size 100 µL
Applications
ELISA (5 µg/ml), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516121 375.00
Check your price
  • Aflatoxin Antibody, DyLight® 550 (Monoclonal, 6A10)
  • Host Mouse
  • Target Species Chemical
  • Conjugate DyLight® 550
  • Catalog # MA516123
  • Size 100 µL
Applications
ELISA (5 µg/ml), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516123 375.00
Check your price
  • Aflatoxin Antibody, DyLight® 650 (Monoclonal, 6A10)
  • Host Mouse
  • Target Species Chemical
  • Conjugate DyLight® 650
  • Catalog # MA516120
  • Size 100 µL
Applications
ELISA (5 µg/ml), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516120 375.00
Check your price
  • Aflatoxin Antibody, HRP (Monoclonal, 6A10)
  • Host Mouse
  • Target Species Chemical
  • Conjugate HRP
  • Catalog # MA516122
  • Size 100 µL
Applications
ELISA (5 µg/ml), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516122 375.00
Check your price
  • AGPAT6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA116960
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Lysophosphatidic acid acyltransferase zeta; Acyl-CoA:glycerol-3-phosphate acyltransferase 4; 1-acylglycerol-3-phosphate O-acyltransferase 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116960 375.00
Check your price
  • AHR Antibody (Monoclonal, RPT1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1514
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (1:10-1:100), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:2000)
Alternate names:
Class E basic helix-loop-helix protein 76
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1514 375.00
Check your price
  • AHR Antibody (Monoclonal, RPT9)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1513
  • Size 100 µL
Applications
ChIP assay (1:100), Flow Cytometry (1:50), Gel Shift (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:10-1:100), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:2000)
Alternate names:
Class E basic helix-loop-helix protein 76
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1513 375.00
Check your price
  • AKR1B10 Antibody, Biotin
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA523018
  • Size 100 µL
Applications
Immunocytochemistry (1:5-1:50), Immunofluorescence (1:5-1:50), Immunohistochemistry (Frozen) (1:5-1:50), Immunohistochemistry (Paraffin) (1:5-1:50), Western Blot (1:500-1:1000)
Alternate names:
Small intestine reductase; Aldose reductase-related protein; Aldose reductase-like; ARL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523018 375.00
Check your price
  • AKR1B10 Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # PA523020
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:5-1:50), Immunofluorescence (1:5-1:50), Immunohistochemistry (Frozen) (1:5-1:50), Immunohistochemistry (Paraffin) (1:5-1:50), Immunoprecipitation (5 µg), Western Blot (1:500-1:1000)
Alternate names:
Small intestine reductase; Aldose reductase-related protein; Aldose reductase-like; ARL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523020 375.00
Check your price
  • AKR1B10 Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # PA523022
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:5-1:50), Immunofluorescence (1:5-1:50), Immunohistochemistry (Frozen) (1:5-1:50), Immunohistochemistry (Paraffin) (1:5-1:50), Immunoprecipitation (5 µg), Western Blot (1:500-1:1000)
Alternate names:
Small intestine reductase; Aldose reductase-related protein; Aldose reductase-like; ARL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523022 375.00
Check your price
  • AKR1B10 Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # PA523019
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:5-1:50), Immunofluorescence (1:5-1:50), Immunohistochemistry (Frozen) (1:5-1:50), Immunohistochemistry (Paraffin) (1:5-1:50), Immunoprecipitation (5 µg), Western Blot (1:500-1:1000)
Alternate names:
Small intestine reductase; Aldose reductase-related protein; Aldose reductase-like; ARL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523019 375.00
Check your price
  • alpha Internexin Antibody
  • Host Rabbit
  • Target Species Bovine, Mammal
  • Conjugate Unconjugated
  • Catalog # PA130608
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:1000-1:2500)
Alternate names:
66 kDa neurofilament protein; Neurofilament 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130608 375.00
Check your price
  • alpha N-Catenin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA120251
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Alpha N-catenin; Alpha-catenin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120251 375.00
Check your price
  • alpha-1a Adrenergic Receptor Antibody
  • Host Rabbit
  • Target Species Fruit fly, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA1047
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:1000), Immunofluorescence (1:10-1:1000), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:400)
Alternate names:
Alpha-1C adrenergic receptor; Alpha-1A adrenoreceptor; Alpha-adrenergic receptor 1c
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1047 375.00
Check your price
  • Amyloid beta 40 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA316760
  • Size 100 µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunoprecipitation (1:100), Western Blot (1:500-1:1000)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316760 375.00
Check your price
  • Amyloid Precursor Protein Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14648
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14648 375.00
Check your price
  • Angiopoietin 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA129640
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500)
Alternate names:
AGPT2, ANG2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129640 375.00
Check your price
  • APE1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA116500
  • Size 100 µL
Applications
Blocking Assay (50-200 molar excess), ChIP assay (1:10-1:500), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (7 µg/ml), Neutralization (50-200 molar excess), Western Blot (1:1000)
Alternate names:
Redox factor-1; REF-1; Apurinic-apyrimidinic endonuclease 1; APEX nuclease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116500 375.00
Check your price
  • Apolipoprotein A1 Antibody (Monoclonal, 2G4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0127
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1:2000)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0127 375.00
Check your price
  • Arg3.1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118301
  • Size 100 µL
Applications
Immunohistochemistry (1:50-1:1000)
Alternate names:
Activity-regulated gene 3.1 protein; ARC/ARG3.1; Activity-regulated gene 3.1 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118301 375.00
Check your price
  • Aromatase Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA116532
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1 µg/ml)
Alternate names:
Estrogen synthase; Cytochrome P450 19A1; Cytochrome P-450AROM; CYPXIX
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116532 375.00
Check your price
  • ATG5 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA146178
  • Size 100 µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (1:400), Western Blot (1:500)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146178 375.00
Check your price
  • ATG5 Antibody, Biotin
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish
  • Conjugate Biotin
  • Catalog # PA522854
  • Size 100 µL
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1:500)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522854 375.00
Check your price
  • ATG5 Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish
  • Conjugate DyLight® 488
  • Catalog # PA522856
  • Size 100 µL
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1:500)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522856 375.00
Check your price
  • ATG5 Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish
  • Conjugate DyLight® 550
  • Catalog # PA522858
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (2.5 µg/ml), Western Blot (1:500)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522858 375.00
Check your price
  • ATG5 Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish
  • Conjugate DyLight® 650
  • Catalog # PA522855
  • Size 100 µL
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1:500)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522855 375.00
Check your price