Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  200  210  220  230  240  250  260  270  300  400  500  600  700  800  900  1000  1100  1605 
5251 - 5280 of 48123 products displayed
  • ATG5 Antibody, HRP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate, Pig, Rat, Xenopus, Zebrafish
  • Conjugate HRP
  • Catalog # PA522857
  • Size 100 µL
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (2.5 µg/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1:500)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522857 375.00
Check your price
  • ATG7 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118299
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunofluorescence (1:200-3000), Immunohistochemistry (1:200-3000), Western Blot (Assay Dependent)
Alternate names:
Autophagy-related protein 7; ATG12-activating enzyme E1 ATG7; Ubiquitin-activating enzyme E1-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118299 375.00
Check your price
  • ATP13A2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116975
  • Size 100 µL
Applications
Immunohistochemistry (10 µg/ml), Western Blot (2 µg/ml)
Alternate names:
AT132 antibody, HSA9947 antibody
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116975 375.00
Check your price
  • ATP13A2 Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522849
  • Size 100 µL
Applications
Immunohistochemistry (10-20 µg/ml), Western Blot (2 µg/ml)
Alternate names:
RP1-37C10.4, HSA9947, KRPPD, PARK9, probable cation-transporting ATPase 13A2, putative ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522849 375.00
Check your price
  • ATP13A2 Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522851
  • Size 100 µL
Applications
Immunohistochemistry (10-20 µg/ml), Western Blot (2 µg/ml)
Alternate names:
RP1-37C10.4, HSA9947, KRPPD, PARK9, probable cation-transporting ATPase 13A2, putative ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522851 375.00
Check your price
  • ATP13A2 Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522853
  • Size 100 µL
Applications
Immunohistochemistry (10-20 µg/ml), Western Blot (2 µg/ml)
Alternate names:
RP1-37C10.4, HSA9947, KRPPD, PARK9, probable cation-transporting ATPase 13A2, putative ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522853 375.00
Check your price
  • ATP13A2 Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522850
  • Size 100 µL
Applications
Immunohistochemistry (10-20 µg/ml), Western Blot (2 µg/ml)
Alternate names:
RP1-37C10.4, HSA9947, KRPPD, PARK9, probable cation-transporting ATPase 13A2, putative ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522850 375.00
Check your price
  • ATP13A2 Antibody, HRP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522852
  • Size 100 µL
Applications
Immunohistochemistry (10-20 µg/ml), Western Blot (2 µg/ml)
Alternate names:
RP1-37C10.4, HSA9947, KRPPD, PARK9, probable cation-transporting ATPase 13A2, putative ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522852 375.00
Check your price
  • ATP1A1 Antibody (Monoclonal, 9-A5)
  • Host Mouse
  • Target Species Dog, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA3924
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1/50), Immunocytochemistry (Assay dependent), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:1,000), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/100), Immunoprecipitation (Assay dependent), Inhibition Assays (Assay De
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3924 375.00
Check your price
  • ATP1B1 Antibody (Monoclonal, 464.8 (aka 8A))
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Pig, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA116732
  • Size 100 µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:1000-1:5000)
Alternate names:
Sodium/potassium-dependent ATPase subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116732 375.00
Check your price
  • ATP4B Antibody (Monoclonal, 2G11)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mustelid, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3923
  • Size 100 µL
Applications
ChIP assay (Assay dependent), Flow Cytometry (1/50), Immunocytochemistry (Assay dependent), Immunofluorescence (1:2,000), Immunohistochemistry (Paraffin, Frozen) (1:2,000), Immunoprecipitation (Assay dependent), Western Blot (1:4,000)
Alternate names:
Proton pump beta chain; Gastric H(+)/K(+) ATPase subunit beta; gp60-90
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3923 375.00
Check your price
  • ATP7B Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA116584
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:1000)
Alternate names:
Wilson disease-associated protein; Copper pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116584 375.00
Check your price
  • ATP7B Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Biotin
  • Catalog # PA522767
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:1000)
Alternate names:
Wilson disease-associated protein; Copper pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522767 375.00
Check your price
  • ATP7B Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522769
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:1000)
Alternate names:
Wilson disease-associated protein; Copper pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522769 375.00
Check your price
  • ATP7B Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522771
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:1000)
Alternate names:
Wilson disease-associated protein; Copper pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522771 375.00
Check your price
  • ATP7B Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522768
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:1000)
Alternate names:
Wilson disease-associated protein; Copper pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522768 375.00
Check your price
  • ATP7B Antibody, HRP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate HRP
  • Catalog # PA522770
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Western Blot (1:1000)
Alternate names:
Wilson disease-associated protein; Copper pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522770 375.00
Check your price
  • ATR Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1450
  • Size 100 µL
Applications
Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
FRAP-related protein 1; Ataxia telangiectasia and Rad3-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1450 375.00
Check your price
  • AUP1 Antibody
  • Host Sheep
  • Target Species Bovine, Human, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA118257
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:500-1:6000), Western Blot (1:500-1:6000)
Alternate names:
60S ribosomal protein L40, HUBCEP52, RPL40, Ubiquitin 52 amino acid fusion protein, Ubiquitin A52 residue ribosomal protein fusion product 1, Ubiquiti
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118257 375.00
Check your price
  • AXUD1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121872
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
TGF-beta-induced apoptosis protein 3; Protein URAX1; Axin-1 up-regulated gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121872 375.00
Check your price
  • Beclin 1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116939
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500)
Alternate names:
Coiled-coil myosin-like BCL2-interacting protein; Protein GT197
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116939 375.00
Check your price
  • Beclin 1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA118287
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (1:100)
Alternate names:
Protein GT197; Coiled-coil myosin-like BCL2-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118287 375.00
Check your price
  • Beclin 1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116857
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:400), Immunoprecipitation (1:80), Western Blot (1:500-1:2000)
Alternate names:
Protein GT197; Coiled-coil myosin-like BCL2-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116857 375.00
Check your price
  • Bestrophin 1 Antibody (Monoclonal, E6-6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116739
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (1:1000)
Alternate names:
Vitelliform macular dystrophy protein 2; TU15B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116739 375.00
Check your price
  • beta COP Antibody (Monoclonal, maD)
  • Host Mouse
  • Target Species Hamster, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA122151
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1/80), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (1/1000)
Alternate names:
Beta-coat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122151 375.00
Check your price
  • beta COP Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1061
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:2,000), Western Blot (1:1,000)
Alternate names:
Beta-coat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1061 375.00
Check your price
  • beta Synuclein Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118259
  • Size 100 µL
Applications
Immunohistochemistry (1:500), Western Blot (1:500)
Alternate names:
Phosphoneuroprotein 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118259 375.00
Check your price
  • beta-1 Adaptin Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118307
  • Size 100 µL
Applications
Immunohistochemistry (1:1000-1:5000), Western Blot (1:100 -1:10000)
Alternate names:
Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118307 375.00
Check your price
  • Blimp-1 Antibody (Monoclonal, 3H2-E8)
  • Host Mouse
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA116874
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Flow Cytometry (8 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000)
Alternate names:
Positive regulatory domain I-binding factor 1; PR domain-containing protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP-1; B lymphocyte-induced maturation protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116874 375.00
Check your price
  • Blimp-1 Antibody, DyLight® 488 (Monoclonal, 3H2-E8)
  • Host Mouse
  • Target Species Human, Mouse, Pig
  • Conjugate DyLight® 488
  • Catalog # MA516115
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Flow Cytometry (8 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000)
Alternate names:
Positive regulatory domain I-binding factor 1; PR domain-containing protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP-1; B lymphocyte-induced maturation protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516115 375.00
Check your price