Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1630 
5491 - 5520 of 48882 products displayed
  • LC3A/LC3B Polyclonal Antibody, HRP conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat, Zebrafish
  • Conjugate HRP
  • Catalog # PA522732
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of lysate), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A; Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522732 375.00
Check your price
  • LC3B Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118293
  • Size 100µL
Applications
Immunofluorescence (1:100-1:1000), Immunohistochemistry (1:100-1:1000), Western Blot (1:100-1:1000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118293 375.00
Check your price
  • LC3B Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and IM
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA146286
  • Size 100µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (0.1-2 µg/ml), Immunofluorescence (0.1-2 µg/ml), Immunohistochemistry (Frozen) (1:400), Immunohistochemistry (Paraffin) (1:200-1:400), Immunomicroscopy (Assay-Dependent), Western Blot (1:1000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146286 375.00
Check your price
  • LC3B Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Bacteria, Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA116930
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (0.5-2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116930 375.00
Check your price
  • LC3B Polyclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Biotin
  • Catalog # PA522936
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (Paraffin) (1:2000), Western Blot (1:3000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522936 375.00
Check your price
  • LC3B Polyclonal Antibody, Biotin conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Bacteria, Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate Biotin
  • Catalog # PA522709
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522709 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate DyLight® 488
  • Catalog # PA522938
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Western Blot (1:3000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522938 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Rat, Zebrafish, Bacteria, Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig
  • Conjugate DyLight® 488
  • Catalog # PA522711
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522711 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Bacteria, Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate DyLight® 550
  • Catalog # PA522713
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522713 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate DyLight® 550
  • Catalog # PA522940
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Western Blot (1:3000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522940 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate DyLight® 650
  • Catalog # PA522937
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Western Blot (1:3000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522937 375.00
Check your price
  • LC3B Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Bacteria, Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate DyLight® 650
  • Catalog # PA522710
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of protein), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522710 375.00
Check your price
  • LC3B Polyclonal Antibody, HRP conjugate for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish
  • Conjugate HRP
  • Catalog # PA522939
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (Frozen) (1:2000), Western Blot (1:3000)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522939 375.00
Check your price
  • LC3B Polyclonal Antibody, HRP conjugate for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Pig, Rat, Zebrafish, Bacteria
  • Conjugate HRP
  • Catalog # PA522712
  • Size 100µL
Applications
Immunoprecipitation (1:200-1:400), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522712 375.00
Check your price
  • LCAT Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA316791
  • Size 100µL
Applications
ELISA (2 µg/ml), Western Blot (1:1000)
Alternate names:
Phospholipid-cholesterol acyltransferase; Lecithin-cholesterol acyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316791 375.00
Check your price
  • Legionella pneumophila Polyclonal Antibody, FITC conjugate for IF and ICC
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # PA128970
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
L. pneumophila, Legionellosis, Legionnaires disease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128970 375.00
Check your price
  • Leptin Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA1052
  • Size 100µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Western Blot (1:500-1:5000)
Alternate names:
Obesity factor; Obese protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1052 375.00
Check your price
  • Leptin Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1053
  • Size 100µL
Applications
Western Blot (1:2,000)
Alternate names:
OB receptor; HuB219; B219
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1053 375.00
Check your price
  • LIMP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Bovine, Hamster, Human, Mink, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA316787
  • Size 100µL
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Immunohistochemistry (Paraffin) (1:25), Western Blot (1:1000)
Alternate names:
Scavenger receptor class B member 2; Lysosome membrane protein II; 85 kDa lysosomal membrane sialoglycoprotein; CD36 antigen-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316787 375.00
Check your price
  • LIMP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA316802
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Scavenger receptor class B member 2; Lysosome membrane protein II; 85 kDa lysosomal membrane sialoglycoprotein; CD36 antigen-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316802 375.00
Check your price
  • LIPG Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116793
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116793 375.00
Check your price
  • LIPG Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116799
  • Size 100µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116799 375.00
Check your price
  • LIPG Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA522911
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522911 375.00
Check your price
  • LIPG Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA522916
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522916 375.00
Check your price
  • LIPG Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # PA522918
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522918 375.00
Check your price
  • LIPG Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # PA522913
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522913 375.00
Check your price
  • LIPG Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # PA522915
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522915 375.00
Check your price
  • LIPG Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # PA522920
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522920 375.00
Check your price
  • LIPG Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # PA522912
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522912 375.00
Check your price
  • LIPG Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # PA522917
  • Size 100µL
Applications
Western Blot (1:500)
Alternate names:
Endothelial cell-derived lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522917 375.00
Check your price