Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1630 
5521 - 5550 of 48881 products displayed
  • LOX Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116953
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-4 µg/ml)
Alternate names:
Ras excision protein; Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116953 375.00
Check your price
  • LPIN1 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116997
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (1.0-2 µg/ml)
Alternate names:
Lipin-1; Fatty liver dystrophy protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116997 375.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Bovine, C. elegans, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116770
  • Size 100µL
Applications
Immunocytochemistry (1:1000-1:2000), Immunofluorescence (1:1000-1:2000), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:500-1:1000), Immunoprecipitation (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116770 375.00
Check your price
  • LYVE1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116635
  • Size 100µL
Applications
Flow Cytometry (1:50-1:500), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116635 375.00
Check your price
  • LYVE1 Polyclonal Antibody, Biotin conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522785
  • Size 100µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522785 375.00
Check your price
  • LYVE1 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IHC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522783
  • Size 100µL
Applications
Flow Cytometry (1:25), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522783 375.00
Check your price
  • LYVE1 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # PA522787
  • Size 100µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522787 375.00
Check your price
  • LYVE1 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522784
  • Size 100µL
Applications
Flow Cytometry (1:25), Immunohistochemistry (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522784 375.00
Check your price
  • LYVE1 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # PA522789
  • Size 100µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522789 375.00
Check your price
  • LYVE1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522782
  • Size 100µL
Applications
Flow Cytometry (1:25), Immunohistochemistry (1:100-1:200), Western Blot (1:500-1:2000)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522782 375.00
Check your price
  • LYVE1 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522786
  • Size 100µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522786 375.00
Check your price
  • LYVE1 Polyclonal Antibody, HRP conjugate for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522788
  • Size 100µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Hyaluronic acid receptor; Extracellular link domain-containing protein 1; Cell surface retention sequence-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522788 375.00
Check your price
  • M13 phage coat protein g8p Monoclonal Antibody for Western Blot, ICC and Flow
  • Host Mouse
  • Target Species Bacteria, Virus
  • Conjugate Unconjugated
  • Catalog # MA106603
  • Size 100µL
Applications
Flow Cytometry (1:25-1:200), Immunocytochemistry (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106603 375.00
Check your price
  • M13 phage coat protein g8p Monoclonal Antibody for Western Blot, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Bacteria, Virus
  • Conjugate Unconjugated
  • Catalog # MA106604
  • Size 100µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1:25-1:200), Immunocytochemistry (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106604 375.00
Check your price
  • MAP1LC3C Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA118286
  • Size 100µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (1:100), Western Blot (1:100)
Alternate names:
Microtubule-associated protein 1 light chain 3 gamma; MAP1A/MAP1B light chain 3 C; MAP1 light chain 3-like protein 3; Autophagy-related ubiquitin-like modifier LC3 C; Autophagy-related protein LC3 C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118286 375.00
Check your price
  • mDIC Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3840
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:500)
Alternate names:
Solute carrier family 25 member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3840 375.00
Check your price
  • mGluR1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # OPA104060
  • Size 100µL
Applications
Immunohistochemistry (1:500), Western Blot (1:1,000)
Alternate names:
mGluR1a, GPRC1A, GRM1A, Human metabotropic glutamate receptor 1 alpha mRNA complete cds
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA104060 375.00
Check your price
  • mGluR1/mGluR5 Polyclonal Antibody for Western Blot, IHC and DB
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14663
  • Size 100µL
Applications
Dot blot (1:1000), Immunohistochemistry (1:500), Western Blot (1:1000)
Alternate names:
Metabotropic glutamate receptor 5, mGluR5, GRM5, GPRC1E, MGLUR5, Metabotropic glutamate receptor 1, mGluR1, GRM1, GPRC1A, MGLUR1, mGlurR1a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14663 375.00
Check your price
  • mGluR2/mGluR3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # OPA104061
  • Size 100µL
Applications
Immunohistochemistry (1:500), Western Blot (1:1,000)
Alternate names:
MGLUR2, mGlu2, Metabotropic glutamate receptor 2, Metabotropic glutamate receptor 3, GPRC1B, GRM2, GRM3, GPRC1C
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA104061 375.00
Check your price
  • mGluR6 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146253
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (1:1000)
Alternate names:
mGluR6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146253 375.00
Check your price
  • MGMT Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA316537
  • Size 100µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:500-1:1000), Western Blot (1:1000)
Alternate names:
O-6-methylguanine-DNA-alkyltransferase; 6-O-methylguanine-DNA methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA316537 375.00
Check your price
  • Mitofilin Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Dog, Cat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116918
  • Size 100µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunoprecipitation (Assay-Dependent), Western Blot (1:2000)
Alternate names:
Mitofilin; Mitochondrial inner membrane protein; p87/89; Cell proliferation-inducing gene 4/52 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116918 375.00
Check your price
  • MMP2 Monoclonal Antibody for IF, ICC, IHC, IHC (F), IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116641
  • Size 100µL
Applications
ELISA (0.03 µg/ml), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (2 µg/ml), Immunohistochemistry (Paraffin) (2 µg/ml), Immunoprecipitation (1-2 µg/100-500 µg total protein), Western Blot (1-2 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116641 375.00
Check your price
  • MMP2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116667
  • Size 100µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (2-4 µg/ml), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (1:1500-1:3500)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116667 375.00
Check your price
  • mtHSP70 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA3028
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Hsp68; Heat shock 70 kDa protein 3; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3028 375.00
Check your price
  • MYL1 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA124940
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (Assay Dependent)
Alternate names:
Myosin light chain alkali 1/2; Skeletal-muscle myosin L-4 light chain; Myosin light chain alkali 2; Myosin light chain 3f; Alkali myosin light chain 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124940 375.00
Check your price
  • n-Myc Monoclonal Antibody for Western Blot, IF, ICC, IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA116638
  • Size 100µL
Applications
ChIP assay (1:10-1:500), Gel Shift (1:1-1:100), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunoprecipitation (1:10-1:100), Western Blot (1:500-1:1000)
Alternate names:
Class E basic helix-loop-helix protein 37
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116638 375.00
Check your price
  • NCK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFPA0075
  • Size 100µL
Applications
Western Blot (1:2000)
Alternate names:
NCK adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0075 375.00
Check your price
  • NEFM Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Chicken, Horse, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA110015
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (1:500-1:1000), Western Blot (1:20,000)
Alternate names:
Neurofilament triplet M protein; 160 kDa neurofilament protein; Neurofilament 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110015 375.00
Check your price
  • Nestin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA516102
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:1000-1:5000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516102 375.00
Check your price