Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1630 
5551 - 5580 of 48882 products displayed
  • n-Myc Monoclonal Antibody for Western Blot, IF, ICC, IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA116638
  • Size 100µL
Applications
ChIP assay (1:10-1:500), Gel Shift (1:1-1:100), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunoprecipitation (1:10-1:100), Western Blot (1:500-1:1000)
Alternate names:
Class E basic helix-loop-helix protein 37
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116638 375.00
Check your price
  • NCK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFPA0075
  • Size 100µL
Applications
Western Blot (1:2000)
Alternate names:
NCK adaptor protein 1; SH2/SH3 adaptor protein NCK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0075 375.00
Check your price
  • NEFM Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Chicken, Horse, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA110015
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (1:500-1:1000), Western Blot (1:20,000)
Alternate names:
Neurofilament triplet M protein; 160 kDa neurofilament protein; Neurofilament 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110015 375.00
Check your price
  • Nestin Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA516102
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:1000-1:5000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516102 375.00
Check your price
  • Nestin Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 488
  • Catalog # MA516104
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:1000-1:5000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516104 375.00
Check your price
  • Nestin Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 550
  • Catalog # MA516106
  • Size 100µl
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:1000-1:5000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516106 375.00
Check your price
  • Nestin Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate DyLight® 650
  • Catalog # MA516103
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:1000-1:5000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516103 375.00
Check your price
  • Nestin Monoclonal Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # MA516105
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:1000-1:5000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516105 375.00
Check your price
  • Neurturin Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118368
  • Size 100µL
Applications
ELISA (1:2000-1:4000), Immunohistochemistry (1:2000-1:3000), Western Blot (1:2000-1:3000)
Alternate names:
Neurturin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118368 375.00
Check your price
  • NFATC1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IHC (PFA), IP, ChIP and GS
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3024
  • Size 100µL
Applications
ChIP assay (Assay dependent), Gel Shift (Assay dependent), Immunocytochemistry (1:250), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:20-200), Immunohistochemistry (Paraffin) (Assay Dependent), Immunohistochemistry (PFA fixed) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blo
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3024 375.00
Check your price
  • NKB Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116745
  • Size 100µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:500-1:1000), Immunohistochemistry (Paraffin) (1:500-1:1000), Western Blot (1:100-1:2000)
Alternate names:
Preprotachykinin-B; Neurokinin B-like protein Zneurok1; ZNEUROK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116745 375.00
Check your price
  • Nkx3.1 Monoclonal Antibody for Western Blot, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116906
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2 µg/ml)
Alternate names:
Homeobox protein NK-3 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116906 375.00
Check your price
  • nNOS Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA3032A
  • Size 100µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Frozen) (1:800), Immunohistochemistry (Paraffin) (1:200-400), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS1; Neuronal NOS; NOS type I; NC-NOS; Constitutive NOS; BNOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3032A 375.00
Check your price
  • Nociceptin Polyclonal Antibody for IHC and ELISA
  • Host Rabbit
  • Target Species Guinea pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA118311
  • Size 100µL
Applications
ELISA (1:1000-1:3000), Immunohistochemistry (1:1000-1:3000)
Alternate names:
Orphanin FQ, PPNOC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118311 375.00
Check your price
  • NOD1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141036
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000-1:2500)
Alternate names:
Caspase recruitment domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141036 375.00
Check your price
  • NOD2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116859
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (Assay-Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Inflammatory bowel disease protein 1; Caspase recruitment domain-containing protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116859 375.00
Check your price
  • NOX4 Polyclonal Antibody for IF, ICC, IHC, IHC (F), IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Bovine, Non-human primate, Pig, Rat, Sheep
  • Conjugate
  • Catalog # PA146014
  • Size 100µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (2-5 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Kidney superoxide-producing NADPH oxidase; Kidney oxidase-1; Superoxide-generating NADPH oxidase 4; Renal NAD(P)H-oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146014 375.00
Check your price
  • NOXA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA118331
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Protein Noxa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118331 375.00
Check your price
  • NPC1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP and IM
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116817
  • Size 100µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (5-10 µg/ml), Immunomicroscopy (Assay-Dependent), Immunoprecipitation (1:10-1:500), Western Blot (1:1000-1:3000)
Alternate names:
FLJ98532, NPC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116817 375.00
Check your price
  • NQO1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA116672
  • Size 100µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116672 375.00
Check your price
  • NQO1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Biotin
  • Catalog # MA516097
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516097 375.00
Check your price
  • NQO1 Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 488
  • Catalog # MA516099
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516099 375.00
Check your price
  • NQO1 Monoclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516101
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516101 375.00
Check your price
  • NQO1 Monoclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516098
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516098 375.00
Check your price
  • NQO1 Monoclonal Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate HRP
  • Catalog # MA516100
  • Size 100µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516100 375.00
Check your price
  • NR3C2 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and IA
  • Host Mouse
  • Target Species Chicken, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1620
  • Size 100µL
Applications
Blocking Assay (Assay Dependent), Flow Cytometry (1/20), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (1:100), Immunohistochemistry (Paraffin) (1/100), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1620 375.00
Check your price
  • NSE Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0108
  • Size 100µL
Applications
Western Blot (1:2000)
Alternate names:
Neuron-specific enolase; Neural enolase; Enolase 2; 2-phospho-D-glycerate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0108 375.00
Check your price
  • NSE Polyclonal Antibody for Western Blot and IF
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA110010
  • Size 100µL
Applications
Immunofluorescence (1:1000), Western Blot (1:2000)
Alternate names:
Neuron-specific enolase; Neural enolase; Enolase 2; 2-phospho-D-glycerate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110010 375.00
Check your price
  • NTF3 Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Chicken, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118390
  • Size 100µL
Applications
ELISA (1:500), Immunohistochemistry (1:500), Inhibition Assays (1:10), Western Blot (1:500)
Alternate names:
Neurotrophic factor; Nerve growth factor 2; HDNF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118390 375.00
Check your price
  • NTF4 Polyclonal Antibody for Western Blot, IHC, ELISA and IA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA118394
  • Size 100µL
Applications
ELISA (1:500), Immunohistochemistry (1:500), Inhibition Assays (1:10), Western Blot (1:500)
Alternate names:
Neutrophic factor 4; Neurotrophin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118394 375.00
Check your price