Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1606 
5611 - 5640 of 48165 products displayed
  • OGT Monoclonal Antibody for Western Blot, IF, IHC (P), IP, ChIP and DB
  • Host Mouse
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA1072
  • Size 100µL
Applications
ChIP assay (Assay Dependent), Dot blot (1/800), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
O-linked N-acetylglucosamine transferase 110 kDa subunit; O-GlcNAc transferase subunit p110
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1072 375.00
Check your price
  • OLFR749 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00007W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Odorant receptor S1, Olfactory receptor MOR106-1, ORL461. GENE NAME: Olfr749
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00007W 375.00
Check your price
  • OMP Polyclonal Antibody for Western Blot and IHC (PFA)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00001W
  • Size 100µL
Applications
Immunohistochemistry (PFA fixed) (1:200-1:2000), Western Blot (1:200-1:2000)
Alternate names:
Olfactory neuronal-specific protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00001W 375.00
Check your price
  • OXPAT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA146216
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Lipid storage droplet protein 5; Myocardial LD protein; Lipid droplet-associated protein PAT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146216 375.00
Check your price
  • p23 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Chicken, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3414
  • Size 100µL
Applications
Flow Cytometry (1/100), Immunocytochemistry (1:200-1:2000), Immunofluorescence (1:200-1:2000), Immunohistochemistry (1:1000), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Telomerase-binding protein p23; Progesterone receptor complex p23; Hsp90 co-chaperone; Cytosolic prostaglandin E2 synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3414 375.00
Check your price
  • Parkin Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Guinea pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA118265
  • Size 100µL
Applications
ELISA (1:500-1:2000), Immunohistochemistry (1:500-1:2000), Western Blot (1:500-1:2000)
Alternate names:
Ubiquitin E3 ligase PRKN, Parkinson juvenile disease protein 2, Parkinson disease protein 2, Prkn, Park2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118265 375.00
Check your price
  • Parkin Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118266
  • Size 100µL
Applications
Immunohistochemistry (1:1,000), Western Blot (1:2,000)
Alternate names:
Parkinson juvenile disease protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118266 375.00
Check your price
  • PARP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14280
  • Size 100µL
Applications
Western Blot (1:250-1:500)
Alternate names:
Poly[ADP-ribose] synthase 2; NAD(+) ADP-ribosyltransferase 2; ADP-ribosyltransferase diphtheria toxin-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14280 375.00
Check your price
  • Paxillin Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bovine, Chicken, Hamster, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA124952
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500)
Alternate names:
hCG_1778014, FLJ16691
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124952 375.00
Check your price
  • PDI Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Dog, Hamster, Human, Mouse, Pig, Rat, Clawed frog
  • Conjugate Unconjugated
  • Catalog # MA3018
  • Size 100µL
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:200-1:2000)
Alternate names:
Prolyl 4-hydroxylase subunit beta; Cellular thyroid hormone-binding protein; p55; Endoplasmic reticulum resident protein 59
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3018 375.00
Check your price
  • PDI Monoclonal Antibody for Western Blot, IF, ICC, IHC (P, F), Flow, IP and IM
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3019
  • Size 100µL
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin, Frozen) (1:100), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:200-1:2000)
Alternate names:
Prolyl 4-hydroxylase subunit beta; Cellular thyroid hormone-binding protein; p55; Endoplasmic reticulum resident protein 59
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3019 375.00
Check your price
  • PDK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116945
  • Size 100µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Pyruvate dehydrogenase kinase isoform 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116945 375.00
Check your price
  • PDZD3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA316819
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Sodium-hydrogen exchanger regulatory factor 4; PDZ domain-containing protein 3; PDZ domain-containing protein 2; Natrium-phosphate cotransporter IIa C-terminal-associated protein 2; Intestinal and kidney-enriched PDZ protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316819 375.00
Check your price
  • PDZK1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA316818
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:400), Western Blot (1:1000)
Alternate names:
Sodium-hydrogen exchanger regulatory factor 3; PDZ domain-containing protein 1; Na/Pi cotransporter C-terminal-associated protein 1; Na(+)/H(+) exchanger regulatory factor 3; CFTR-associated protein of 70 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316818 375.00
Check your price
  • PEN2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA316762
  • Size 100µL
Applications
Immunoprecipitation (1:50-1:100), Western Blot (1:1000-1:2500)
Alternate names:
Presenilin enhancer protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316762 375.00
Check your price
  • PER2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116513
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:1000)
Alternate names:
Circadian clock protein PERIOD 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116513 375.00
Check your price
  • Peripherin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA110012
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Western Blot (1:20,000)
Alternate names:
Neurofilament 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110012 375.00
Check your price
  • Peripherin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA110018
  • Size 100µL
Applications
Immunocytochemistry (1:2000), Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Western Blot (1:20,000)
Alternate names:
Neurofilament 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110018 375.00
Check your price
  • Perlecan Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rat
  • Target Species Bovine, Fish, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA106821
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:25-1:200), Immunohistochemistry (Frozen) (1:25-1:200), Immunohistochemistry (Paraffin) (1:25-1:200), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106821 375.00
Check your price
  • Phospho-14-3-3 beta (Ser58) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA14612
  • Size 100µL
Applications
Immunohistochemistry (1:500), Western Blot (1:1000)
Alternate names:
Protein kinase C inhibitor protein 1; Protein 1054; Prepronerve growth factor RNH-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14612 375.00
Check your price
  • Phospho-AKT1 (Ser473) Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # OMA103061
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
RAC-PK-alpha; Proto-oncogene c-Akt; Protein kinase B alpha; Protein kinase B; Thymoma viral proto-oncogene; AKT1 kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
OMA103061 375.00
Check your price
  • Phospho-alpha Synuclein (Ser129) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14686
  • Size 100µL
Applications
Western Blot (1/1000)
Alternate names:
Alpha-synuclein, Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14686 375.00
Check your price
  • Phospho-ATM (Ser1981) Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA146069
  • Size 100µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:10-1:500), Western Blot (1:1000)
Alternate names:
Ataxia telangiectasia mutated; Ataxia telangiectasia mutated homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146069 375.00
Check your price
  • Phospho-CaMKII alpha (Thr286) Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1047
  • Size 100µL
Applications
ELISA (Assay dependent), Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:100), Immunofluorescence (1:50), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2,000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, Phospho-CaMKI
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1047 375.00
Check your price
  • Phospho-CaMKII alpha/beta (Thr286) Polyclonal Antibody for Western Blot and DB
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14614
  • Size 100µL
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
phospho-Ca2+/Calmodulin-Dependent Protein Kinase II (Thr286)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14614 375.00
Check your price
  • Phospho-CDK1 (Tyr15) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat, Clawed frog
  • Conjugate Unconjugated
  • Catalog # PA14617
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
p34 protein kinase; Cell division protein kinase 1; Cell division control protein 2 homolog; p34 protein kinase 1; Cell division protein kinase 1-A; Cell division control protein 2-A; Cell division control protein 2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14617 375.00
Check your price
  • Phospho-CREB (Ser133) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14619
  • Size 100µL
Applications
Immunohistochemistry (1:100), Western Blot (1:1000)
Alternate names:
phospho-cyclic AMP response element binding protein (Ser133)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14619 375.00
Check your price
  • Phospho-CREB (Ser133) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1851B
  • Size 100µL
Applications
Western Blot (1:250)
Alternate names:
Phospho-Cyclic AMP Response Element Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1851B 375.00
Check your price
  • Phospho-DARPP-32 (Thr34) Polyclonal Antibody for Western Blot and DB
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14609
  • Size 100µL
Applications
Dot blot (1:1000), Western Blot (1:1000)
Alternate names:
Dopamine- and cAMP-regulated neuronal phosphoprotein; DARPP-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14609 375.00
Check your price
  • Phospho-DARPP-32 (Thr75) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14611
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Dopamine- and cAMP-regulated neuronal phosphoprotein; DARPP-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14611 375.00
Check your price