Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  210  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1605 
5701 - 5730 of 48136 products displayed
  • NKB Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116745
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:500-1:1000), Immunohistochemistry (Paraffin) (1:500-1:1000), Western Blot (1:100-1:2000)
Alternate names:
Preprotachykinin-B; Neurokinin B-like protein Zneurok1; ZNEUROK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116745 375.00
Check your price
  • Nkx3.1 Antibody (Monoclonal, 0361)
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116906
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Western Blot (2 µg/ml)
Alternate names:
Homeobox protein NK-3 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116906 375.00
Check your price
  • nNOS Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA3032A
  • Size 100 µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Frozen) (1:800), Immunohistochemistry (Paraffin) (1:200-400), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS1; Neuronal NOS; NOS type I; NC-NOS; Constitutive NOS; BNOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3032A 375.00
Check your price
  • Nociceptin Antibody
  • Host Rabbit
  • Target Species Guinea pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA118311
  • Size 100 µL
Applications
ELISA (1:1000-1:3000), Immunohistochemistry (1:1000-1:3000)
Alternate names:
Orphanin FQ, PPNOC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118311 375.00
Check your price
  • NOD1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141036
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000-1:2500)
Alternate names:
Caspase recruitment domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141036 375.00
Check your price
  • NOD2 Antibody (Monoclonal, 2D9)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA116611
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
Inflammatory bowel disease protein 1; Caspase recruitment domain-containing protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116611 375.00
Check your price
  • NOD2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116859
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (Assay-Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Inflammatory bowel disease protein 1; Caspase recruitment domain-containing protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116859 375.00
Check your price
  • NOXA Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA118331
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
Protein Noxa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118331 375.00
Check your price
  • NPC1 Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116817
  • Size 100 µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (5-10 µg/ml), Immunomicroscopy (Assay-Dependent), Immunoprecipitation (1:10-1:500), Western Blot (1:1000-1:3000)
Alternate names:
FLJ98532, NPC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116817 375.00
Check your price
  • NQO1 Antibody (Monoclonal, A180)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA116672
  • Size 100 µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116672 375.00
Check your price
  • NQO1 Antibody, Biotin (Monoclonal, A180)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Biotin
  • Catalog # MA516097
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516097 375.00
Check your price
  • NQO1 Antibody, DyLight® 488 (Monoclonal, A180)
  • Host Mouse
  • Target Species Mouse, Non-human primate, Rat, Dog, Human
  • Conjugate DyLight® 488
  • Catalog # MA516099
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516099 375.00
Check your price
  • NQO1 Antibody, DyLight® 550 (Monoclonal, A180)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516101
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516101 375.00
Check your price
  • NQO1 Antibody, DyLight® 650 (Monoclonal, A180)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516098
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516098 375.00
Check your price
  • NQO1 Antibody, HRP (Monoclonal, A180)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate HRP
  • Catalog # MA516100
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516100 375.00
Check your price
  • NR3C2 Antibody (Monoclonal, H10E4C9F)
  • Host Mouse
  • Target Species Chicken, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1620
  • Size 100 µL
Applications
Blocking Assay (Assay Dependent), Flow Cytometry (1/20), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (1:100), Immunohistochemistry (Paraffin) (1/100), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1620 375.00
Check your price
  • NSE Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0108
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
Neuron-specific enolase; Neural enolase; Enolase 2; 2-phospho-D-glycerate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0108 375.00
Check your price
  • NSE Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA110010
  • Size 100 µL
Applications
Immunofluorescence (1:1000), Western Blot (1:2000)
Alternate names:
Neuron-specific enolase; Neural enolase; Enolase 2; 2-phospho-D-glycerate hydro-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110010 375.00
Check your price
  • NTF3 Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118390
  • Size 100 µL
Applications
ELISA (1:500), Immunohistochemistry (1:500), Inhibition Assays (1:10), Western Blot (1:500)
Alternate names:
Neurotrophic factor; Nerve growth factor 2; HDNF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118390 375.00
Check your price
  • NTF4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA118394
  • Size 100 µL
Applications
ELISA (1:500), Immunohistochemistry (1:500), Inhibition Assays (1:10), Western Blot (1:500)
Alternate names:
Neutrophic factor 4; Neurotrophin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118394 375.00
Check your price
  • Nucleolin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Plant, Rat
  • Conjugate Unconjugated
  • Catalog # PA316875
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100), Immunomicroscopy (1:10-1:500), Western Blot (1:4000)
Alternate names:
Protein C23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316875 375.00
Check your price
  • NuMA Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA316829
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (Assay-Dependent), Immunoprecipitation (1:10-1:500), Western Blot (1:1000)
Alternate names:
SP-H antigen; Nuclear matrix protein-22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316829 375.00
Check your price
  • NUTF2 Antibody (Monoclonal, 4F5)
  • Host Mouse
  • Target Species Xenopus, Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA125324
  • Size 100 µL
Applications
Blocking Assay (Assay-dependent), ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-2 µg/ml)
Alternate names:
Placental protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125324 375.00
Check your price
  • O-linked N-acetylglucosamine (O-GlcNAc) Antibody (Monoclonal, RL2)
  • Host Mouse
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA1072
  • Size 100 µL
Applications
ChIP assay (Assay Dependent), Dot blot (1/800), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
O-linked N-acetylglucosamine transferase 110 kDa subunit; O-GlcNAc transferase subunit p110
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1072 375.00
Check your price
  • OATP2 Antibody (Monoclonal, ESL)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3934
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
Solute carrier family 21 member 6; Sodium-independent organic anion-transporting polypeptide 2; OATP-C; Liver-specific organic anion transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3934 375.00
Check your price
  • OCT4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116943
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (2 µg/ml)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116943 375.00
Check your price
  • OCT4 Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522735
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522735 375.00
Check your price
  • OCT4 Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522736
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522736 375.00
Check your price
  • OCT4 Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522734
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522734 375.00
Check your price
  • OLFR749 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00007W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Odorant receptor S1, Olfactory receptor MOR106-1, ORL461. GENE NAME: Olfr749
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00007W 375.00
Check your price