Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1630 
5851 - 5880 of 48882 products displayed
  • Synapsin 1 Polyclonal Antibody for Western Blot, IF, IHC and IP
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14673
  • Size 10µg
Applications
Immunofluorescence (1:2000), Immunohistochemistry (1:2000), Immunoprecipitation (1ug per 200 ug lysate), Western Blot (1:1000)
Alternate names:
Synapsin I; Brain protein 4.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14673 375.00
Check your price
  • AKAP12 Polyclonal Antibody for Western Blot, IHC and IP
  • Host Sheep
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA129372
  • Size 125µg
Applications
Immunohistochemistry (Assay-dependent), Immunoprecipitation (2-5 µg/ml), Western Blot (2-5 µg/ml)
Alternate names:
AKAP12A, AKAP12B, AKAP12G
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129372 375.00
Check your price
  • Bcl-2 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Sheep
  • Target Species Dog, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA128275
  • Size 125µg
Applications
Immunocytochemistry (1:5000), Immunofluorescence (1:5000), Immunoprecipitation (Assay dependent), Western Blot (1:5000)
Alternate names:
Bcl-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128275 375.00
Check your price
  • Tomato Spotted Wilt Virus Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA120368
  • Size 125µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120368 375.00
Check your price
  • ATG12 Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSA00025W
  • Size 150µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Autophagy-related protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSA00025W 375.00
Check your price
  • ATG7 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSA00019W
  • Size 150µL
Applications
Immunohistochemistry (1:100-1:2000)
Alternate names:
Ubiquitin-activating enzyme E1-like protein; Autophagy-related protein 7; ATG12-activating enzyme E1 ATG7
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSA00019W 375.00
Check your price
  • CAP-18 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSC00009W
  • Size 150µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:100-1:2000), Western Blot (1:100-1:2000)
Alternate names:
18 kDa cationic antimicrobial protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSC00009W 375.00
Check your price
  • Phospho-GluR1 (Ser845) Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # OPA104118
  • Size 150µL
Applications
Immunohistochemistry (Frozen) (1:1000), Western Blot (1: 1,000)
Alternate names:
Glutamate receptor ionotropic, AMPA 1; GluR-K1; GluR-A; AMPA-selective glutamate receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA104118 375.00
Check your price
  • NMDAR1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OMA104010
  • Size 15µg
Applications
Immunoprecipitation (3 µg/200µg lysate), Western Blot (1:1000)
Alternate names:
N-methyl-D-aspartate receptor subunit NR1; Glutamate [NMDA] receptor subunit zeta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OMA104010 375.00
Check your price
  • Adenovirus Type 3 Monoclonal Antibody for IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA180653
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
HADV, AdV, Adeno Virus, HADV-3, AdV-3, Adeno Virus 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180653 375.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA126909
  • Size 1mg
Applications
ELISA (1:50,000), Western Blot (1:2000-1:10,000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126909 375.00
Check your price
  • Avidin Polyclonal Antibody for Western Blot, IP, ELISA, Neu and IA
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA126812
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Inhibition Assays (Assay Dependent), Neutralization (Assay-dependent), Western Blot (Assay Dependent)
Alternate names:
AVD, avID
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126812 375.00
Check your price
  • Bovine Serum Albumin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # MA182941
  • Size 1mg
Applications
ELISA (1:250-1:1000), Western Blot (Assay Dependent)
Alternate names:
BSA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182941 375.00
Check your price
  • Chicken anti-Rat IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Chicken
  • Target Species Rat
  • Conjugate AP
  • Catalog # PA130741
  • Size 1mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130741 375.00
Check your price
  • Complement C3c Polyclonal Antibody, FITC conjugate for IF and ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate FITC
  • Catalog # PA533132
  • Size 1mg
Applications
ELISA (1:500-1:50,000), Immunofluorescence (1:10-1:50)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533132 375.00
Check your price
  • F(ab')2-Rabbit anti-Rat IgG Secondary Antibody, HRP conjugate for IHC, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate HRP
  • Catalog # PA533281
  • Size 1mg
Applications
ELISA (1:500-1:1000), Immunohistochemistry (1:50), Immunohistochemistry (Frozen) (1:50), Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533281 375.00
Check your price
  • Goat anti-Feline IgG Secondary Antibody, HRP conjugate for ELISA
  • Host Goat
  • Target Species Cat
  • Conjugate HRP
  • Catalog # PA184673
  • Size 1mg
Applications
ELISA (1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184673 375.00
Check your price
  • hCG alpha Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191691
  • Size 1mg
Applications
ELISA (Assay dependent)
Alternate names:
Thyrotropin alpha chain; Thyroid-stimulating hormone alpha chain; Lutropin alpha chain; Luteinizing hormone alpha chain; Follitropin alpha chain; Follicle-stimulating hormone alpha chain; Chorionic gonadotrophin subunit alpha; Choriogonadotropin alpha chain; Anterior pituitary glycoprotein hormones common subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191691 375.00
Check your price
  • HSV Type 1 VP21/VP22a Monoclonal Antibody for Western Blot, IF, IP and IM
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516798
  • Size 1mg
Applications
Immunofluorescence (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516798 375.00
Check your price
  • HSV Type 2 gG Monoclonal Antibody for Western Blot, IF, IHC (F) and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516575
  • Size 1mg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, Glycoprotein G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516575 375.00
Check your price
  • Influenza B NP Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA180712
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180712 375.00
Check your price
  • Intestinal Alkaline Phosphatase Polyclonal Antibody, HRP conjugate for Western Blot, IHC (F), ELISA and IM
  • Host Goat
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # PA126878
  • Size 1mg
Applications
ELISA (1:500-1:1000), Immunohistochemistry (Frozen) (Assay Dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay Dependent)
Alternate names:
IAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126878 375.00
Check your price
  • Mouse anti-Human IgA Secondary Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516451
  • Size 1mg
Applications
ELISA (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516451 375.00
Check your price
  • Protein A Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Bacteria, Tag
  • Conjugate Unconjugated
  • Catalog # PA127415
  • Size 1mg
Applications
ELISA (1:200-1:500), Western Blot (0.1-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127415 375.00
Check your price
  • Protein A Polyclonal Antibody, HRP conjugate for Western Blot, IHC (F) and ELISA
  • Host Goat
  • Target Species Bacteria, Tag
  • Conjugate HRP
  • Catalog # PA126853
  • Size 1mg
Applications
Dot blot (Assay Dependent), ELISA (1:10,000-1:40,000), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126853 375.00
Check your price
  • Rabbit anti-Donkey IgG Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Rabbit
  • Target Species Donkey
  • Conjugate AP
  • Catalog # PA130343
  • Size 1mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130343 375.00
Check your price
  • Rat Albumin Polyclonal Antibody, FITC conjugate for IF and ELISA
  • Host Sheep
  • Target Species Bovine, Rat
  • Conjugate FITC
  • Catalog # PA186695
  • Size 1mg
Applications
ELISA (1/1000-1/25000), Immunofluorescence (1/10 -1/50)
Alternate names:
BSA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186695 375.00
Check your price
  • Sheep anti-Canine IgG Secondary Antibody, HRP conjugate for ELISA
  • Host Sheep
  • Target Species Dog
  • Conjugate HRP
  • Catalog # PA184644
  • Size 1mg
Applications
ELISA (1/10,000 -1/100,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184644 375.00
Check your price
  • Transferrin Polyclonal Antibody, Biotin conjugate for Western Blot, IHC, ELISA and IM
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA127251
  • Size 1mg
Applications
ELISA (1:16,000-1:68,000), Immunohistochemistry (Assay Dependent), Immunomicroscopy (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Siderophilin; Beta-1 metal-binding globulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127251 375.00
Check your price
  • Adenovirus Type 5 Hexon Polyclonal Antibody, FITC conjugate for IF
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173053
  • Size 1mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
HADV, AdV, Adeno Virus, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173053 375.00
Check your price