Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  110  120  130  140  150  160  170  180  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1200  1622 
6001 - 6030 of 48655 products displayed
  • SERCA1 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Amphibian, Dog, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3912
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:20), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Calcium pump 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3912 375.00
Check your price
  • SERCA2 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3919
  • Size 100 µL
Applications
Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (1:250), Immunofluorescence (1:100-1:200), Immunohistochemistry (1:100), Immunohistochemistry (Frozen) (1/100), Immunohistochemistry (Paraffin) (1 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; Calcium pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3919 375.00
Check your price
  • SERCA2 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3910
  • Size 100 µL
Applications
Flow Cytometry (1µg per 10^6 cells), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; Calcium pump 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3910 375.00
Check your price
  • SERPINB12 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123493
  • Size 100 µL
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1:1000)
Alternate names:
MGC119247, MGC119248, YUKOPIN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123493 375.00
Check your price
  • Sir2.1 Polyclonal Antibody for Western Blot, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species C. elegans
  • Conjugate Unconjugated
  • Catalog # PA116933
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1:3000)
Alternate names:
Regulatory protein SIR2 homolog 1; Protein sir-2.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116933 375.00
Check your price
  • SIRT2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3200
  • Size 100 µL
Applications
Immunocytochemistry (1:400), Immunofluorescence (1:400), Western Blot (1:1,000)
Alternate names:
SIR2-like protein 2; Regulatory protein SIR2 homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3200 375.00
Check your price
  • SLC7A11 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116893
  • Size 100 µL
Applications
Flow Cytometry (2-3 µg/ml), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1:1000)
Alternate names:
xCT; Solute carrier family 7 member 11; Calcium channel blocker resistance protein CCBR1; Amino acid transport system xc-
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116893 375.00
Check your price
  • SLIRP Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA146148
  • Size 100 µL
Applications
Immunocytochemistry (1:40), Immunofluorescence (1:40), Immunohistochemistry (Paraffin) (1:100), Western Blot (0.5-2.0 µg/ml)
Alternate names:
DC50, PD04872, SLIRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146148 375.00
Check your price
  • SMARCB1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Fish, Human, Mouse, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA125450
  • Size 100 µL
Applications
Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
SNF5 homolog; Integrase interactor 1 protein; BRG1-associated factor 47
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125450 375.00
Check your price
  • SNAP25 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14695
  • Size 100 µL
Applications
Western Blot (1/1000)
Alternate names:
Synaptosomal-associated 25 kDa protein; Super protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14695 375.00
Check your price
  • SNAP25 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Bovine, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA1740
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1 µg/ml)
Alternate names:
Synaptosome-associated protein 25.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1740 375.00
Check your price
  • SNTA1 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA125013
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:1000-1:2000), Western Blot (1:2,000)
Alternate names:
Syntrophin-1; 59-1 DAP; 59 kDa dystrophin-associated protein A1 acidic component 1; Pro-TGF-alpha cytoplasmic domain-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125013 375.00
Check your price
  • SOD1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118275
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
SOD1, ALS, ALS1, Cu/Zn superoxide dismutase, Homodimer, Indophenoloxidase A, IPOA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118275 375.00
Check your price
  • SOD1 Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA118276
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:100-1:1000), Western Blot (1:1000-1:4000)
Alternate names:
Superoxide dismutase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118276 375.00
Check your price
  • SOD1 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0013
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (1 µl), Western Blot (1:2000)
Alternate names:
Superoxide dismutase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0013 375.00
Check your price
  • SOD2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0021
  • Size 100 µL
Applications
Immunoprecipitation (1 µl), Western Blot (1:2000)
Alternate names:
SOD2, IPO B, Manganese SOD, Manganese superoxide dismutase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0021 375.00
Check your price
  • SOD3 Polyclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA118278
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:100), Western Blot (1:1,000)
Alternate names:
SOD3, ALS, ALS1, Cu/Zn superoxide dismutase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118278 375.00
Check your price
  • Sodium/Calcium Exchanger Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3926
  • Size 100 µL
Applications
Flow Cytometry (1/100), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (1:20-50), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/100), Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Solute carrier family 8 member 1; Na(+)/Ca(2+)-exchange protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3926 375.00
Check your price
  • SOX2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Sheep
  • Conjugate Unconjugated
  • Catalog # PA116968
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Immunohistochemistry (Paraffin) (1:125-1:250), Western Blot (0.5 µg/ml)
Alternate names:
ANOP3, MCOPS3, SRY-related HMG-box gene 2, transcription factor SOX-2, transcription factor SOX2, sex determining region Y box 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116968 375.00
Check your price
  • Spectrin beta-1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA3062
  • Size 100 µL
Applications
Flow Cytometry (0.5-2 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:100)
Alternate names:
Beta-I spectrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3062 375.00
Check your price
  • SR-BI Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Goat
  • Target Species Hamster, Human, Mink, Mouse, Rodent, Rat
  • Conjugate Unconjugated
  • Catalog # PA116803
  • Size 100 µL
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:500)
Alternate names:
SR-BI; Collagen type I receptor, thrombospondin receptor-like 1; CD36 antigen-like 1; CD36 and LIMPII analogous 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116803 375.00
Check your price
  • SSTR2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA316736
  • Size 100 µL
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:100-1:250), Immunofluorescence (1:100-1:250), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:100-1:1000)
Alternate names:
SRIF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316736 375.00
Check your price
  • SSTR3 Polyclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3110
  • Size 100 µL
Applications
Immunocytochemistry (1:5000), Immunohistochemistry (Paraffin) (1:2500), Western Blot (1:5000)
Alternate names:
SSR-28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3110 375.00
Check your price
  • SSTR4 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3111
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:2500), Western Blot (1:100-1:5000)
Alternate names:
SSTR4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3111 375.00
Check your price
  • SSTR5 Polyclonal Antibody for Western Blot, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3112
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:5000), Immunohistochemistry (Paraffin) (1:2500), Western Blot (1:4000-1:6000 )
Alternate names:
SSTR5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3112 375.00
Check your price
  • STIM1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146217
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:1000)
Alternate names:
D11S4896E, GOK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146217 375.00
Check your price
  • SUMO1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118267
  • Size 100 µL
Applications
Immunohistochemistry (1:1,000-1:2,000), Western Blot (1:2,000-1:4,000)
Alternate names:
Ubiquitin-like protein UBL1; Ubiquitin-like protein SMT3C; Ubiquitin-homology domain protein PIC1; Sentrin; SMT3 homolog 3; GAP-modifying protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118267 375.00
Check your price
  • Survivin Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Hamster, Human
  • Conjugate Unconjugated
  • Catalog # MA116854
  • Size 100 µL
Applications
ELISA (Assay-Dependent), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (20 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:1000)
Alternate names:
Apoptosis inhibitor survivin; Apoptosis inhibitor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116854 375.00
Check your price
  • Survivin Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P), Flow, IP, ChIP and ELISA
  • Host Rabbit
  • Target Species Dog, Cat, Guinea pig, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116836
  • Size 100 µL
Applications
ChIP assay (1:10-1:500), ELISA (Assay-Dependent), Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:50-1:250), Immunofluorescence (1:50-1:250), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:50-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1 µg/ml)
Alternate names:
Apoptosis inhibitor survivin; TIAP; Apoptosis inhibitor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116836 375.00
Check your price
  • Synaptotagmin 1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14699
  • Size 100 µL
Applications
Western Blot (1/1000)
Alternate names:
p65; Synaptotagmin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14699 375.00
Check your price