Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  110  120  130  140  150  160  170  180  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1200  1630 
6091 - 6120 of 48882 products displayed
  • Met-Enkephalin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA183140
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183140 375.00
Check your price
  • MMP1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112891
  • Size 200µg
Applications
Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Matrix metalloproteinase-1; Fibroblast collagenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112891 375.00
Check your price
  • MUSK Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11741
  • Size 200µg
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Immunohistochemistry (Paraffin) (1:20-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
Muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11741 375.00
Check your price
  • Mycobacterium avium Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183339
  • Size 200µg
Applications
ELISA (1:5000)
Alternate names:
M. avium
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183339 375.00
Check your price
  • Neisseria gonorrhoeae MOMP Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183358
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
N. gonorrhoeae, Gonococci
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183358 375.00
Check your price
  • Neutrophil elastase Monoclonal Antibody for Flow, ELISA and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183125
  • Size 200µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Functional Assay (Assay Dependent)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183125 375.00
Check your price
  • Osteocalcin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182973
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
Gamma-carboxyglutamic acid-containing protein; Bone Gla protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182973 375.00
Check your price
  • P-Selectin Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Bovine, Dog, Horse, Cat, Goat, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA181809
  • Size 200µg
Applications
Flow Cytometry (1/25-1/50), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Platelet activation dependent granule-external membrane protein; Leukocyte-endothelial cell adhesion molecule 3; Granule membrane protein 140; CD62 antigen-like family member P; CD62P antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181809 375.00
Check your price
  • PGC Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183392
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
Pepsinogen C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183392 375.00
Check your price
  • Plasmodium falciparum MSP1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA174365
  • Size 200µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
P. falciparum
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174365 375.00
Check your price
  • pro-Brain Natriuretic Peptide Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120584
  • Size 200µg
Applications
Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120584 375.00
Check your price
  • pro-Brain Natriuretic Peptide Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120626
  • Size 200µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Gamma-brain natriuretic peptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120626 375.00
Check your price
  • PSEN1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IHC (PFA), IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1751
  • Size 200µg
Applications
ELISA (4.5 ug/ml), Immunocytochemistry (Assay dependent), Immunofluorescence (0.75 ug/ml), Immunohistochemistry (Paraffin) (0.75 ug/ml), Immunohistochemistry (PFA fixed) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:100-1:1000)
Alternate names:
Protein S182
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1751 375.00
Check your price
  • Pseudomonas aeruginosa Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183430
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
P. aeruginosa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183430 375.00
Check your price
  • Salmonella Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA116686
  • Size 200µg
Applications
ELISA (1:100-1:2000)
Alternate names:
Salmonellae
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116686 375.00
Check your price
  • Salmonella typhimurium LPS Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183451
  • Size 200µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Salmonellae, S. typhimurium lipopolysaccharide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183451 375.00
Check your price
  • Shigella dysenteriae 1 Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183460
  • Size 200µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Shigella dysenteriae, shigella, shigellosis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183460 375.00
Check your price
  • Staphylococcus Enterotoxin pan Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183468
  • Size 200µg
Applications
ELISA (1/5,000-1/20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183468 375.00
Check your price
  • ABCG8 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116798
  • Size 200µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Western Blot (1:250-1:500)
Alternate names:
Sterolin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116798 375.00
Check your price
  • Acetylcholinesterase Monoclonal Antibody for IF, ICC, IHC (F), IP and ELISA
  • Host Mouse
  • Target Species Bovine, Cat, Guinea pig, Human, Mouse, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA3042
  • Size 200µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Frozen) (1:50), Immunoprecipitation (Assay dependent)
Alternate names:
AChE, EC 3.1.1.7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3042 375.00
Check your price
  • BMAL1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1523
  • Size 200µL
Applications
Western Blot (1:200)
Alternate names:
Brain and muscle ARNT-like 1; Arnt3; bHLH-PAS protein JAP3; PAS domain-containing protein 3; Member of PAS protein 3; Class E basic helix-loop-helix protein 5; Basic-helix-loop-helix-PAS protein MOP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1523 375.00
Check your price
  • Caspase 8 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA116841
  • Size 200µL
Applications
Western Blot (1:1000)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116841 375.00
Check your price
  • CDK9 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519670
  • Size 200µL
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1:250)
Alternate names:
Tat-associated kinase complex catalytic subunit; Serine/threonine-protein kinase PITALRE; Cell division protein kinase 9; Cell division cycle 2-like protein kinase 4; C-2K
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519670 375.00
Check your price
  • CEA Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125068
  • Size 200µL
Applications
Immunohistochemistry (Paraffin) (1:8000), Western Blot (1:8000)
Alternate names:
Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125068 375.00
Check your price
  • CLOCK Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1520
  • Size 200µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin, Frozen) (1:100-1:1000), Western Blot (1:200)
Alternate names:
Class E basic helix-loop-helix protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1520 375.00
Check your price
  • FIS1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141082
  • Size 200µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:500-1:1000)
Alternate names:
Tetratricopeptide repeat protein 11; FIS1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141082 375.00
Check your price
  • Glucocorticoid Receptor Polyclonal Antibody for Western Blot, IF, ICC, IHC, IP and GS
  • Host Rabbit
  • Target Species Human, Reptile, Rat
  • Conjugate Unconjugated
  • Catalog # PA1510A
  • Size 200µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:250), Immunoprecipitation (Assay dependent), Western Blot (1:100)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1510A 375.00
Check your price
  • Gram Negative Endotoxin Monoclonal Antibody for ELISA and IA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA191641
  • Size 200µL
Applications
ELISA (Assay-dependent), Inhibition Assays (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191641 375.00
Check your price
  • IL-6 Polyclonal Antibody for Western Blot, IHC (F), IHC (P), IP, ELISA and RIA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126811
  • Size 200µL
Applications
ELISA (1:1000-1:5000), Immunohistochemistry (Frozen) (1:400-1:800), Immunohistochemistry (Paraffin) (1:400-1:800), Immunoprecipitation (1:400-1:800), Neutralization (1:400), Radioimmune Assays (1:4000-1:8000), Western Blot (1:500-1:2000)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126811 375.00
Check your price
  • IL17RB Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141211
  • Size 200µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Interleukin-17B receptor; IL-17 receptor homolog 1; Cytokine receptor-like 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141211 375.00
Check your price