Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6361 - 6390 of 48881 products displayed
  • Chymotrypsin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110623
  • Size 500µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Caldecrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110623 375.00
Check your price
  • CXCL1 Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523707
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73); C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523707 375.00
Check your price
  • CXCL10 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523774
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine B10; Interferon-gamma induced protein CRG-2; C7; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523774 375.00
Check your price
  • CXCL10 Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523726
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.5-2.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine B10; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523726 375.00
Check your price
  • CXCL2 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523737
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Macrophage inflammatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523737 375.00
Check your price
  • CXCL8 Monoclonal Antibody for Western Blot, IF, ICC, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523697
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
T-cell chemotactic factor; Protein 3-10C; Neutrophil-activating protein 1; Monocyte-derived neutrophil-activating peptide; Monocyte-derived neutrophil chemotactic factor; Granulocyte chemotactic protein 1; Emoctakin; Chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523697 375.00
Check your price
  • Digoxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110811
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110811 375.00
Check your price
  • Digoxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110809
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110809 375.00
Check your price
  • EGF Monoclonal Antibody for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523704
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunohistochemistry (8-25 µg/ml)
Alternate names:
Epidermal Growth Factor, URG, Urogastrone, HOMG4, URG, beta-urogastrone, pro-epidermal growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523704 375.00
Check your price
  • FABP3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122721
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Muscle fatty acid-binding protein; Mammary-derived growth inhibitor; Heart-type fatty acid-binding protein; Fatty acid-binding protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122721 375.00
Check your price
  • FABP4 Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121216
  • Size 500µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Fatty acid-binding protein 4; Adipocyte-type fatty acid-binding protein; Adipocyte lipid-binding protein; Protein 422; P2 adipocyte protein; P15; Myelin P2 protein homolog; 3T3-L1 lipid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121216 375.00
Check your price
  • GnRH1 Monoclonal Antibody for IHC, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110817
  • Size 500µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
Progonadoliberin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110817 375.00
Check your price
  • Gram Negative Endotoxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110687
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110687 375.00
Check your price
  • Gram Negative Endotoxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110688
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110688 375.00
Check your price
  • Gram Negative Endotoxin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110684
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110684 375.00
Check your price
  • hCG beta Monoclonal Antibody for Western Blot, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182291
  • Size 500µg
Applications
ELISA (1/1000-1/80000), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182291 375.00
Check your price
  • Hemoglobin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110806
  • Size 500µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110806 375.00
Check your price
  • IFN gamma Monoclonal Antibody for IF, ICC and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523718
  • Size 500µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.02-0.06 µg/ml)
Alternate names:
Immune interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523718 375.00
Check your price
  • IGFBP1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523727
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Placental protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523727 375.00
Check your price
  • IL-1 beta Monoclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523691
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523691 375.00
Check your price
  • IL-1 beta Monoclonal Antibody for ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523721
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-5 µg/ml)
Alternate names:
IL-1 beta, catabolin, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta, IL1B, IL1 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523721 375.00
Check your price
  • IL-10 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523796
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.005-0.015 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523796 375.00
Check your price
  • IL-11 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523711
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Adipogenesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523711 375.00
Check your price
  • IL-12 p70 Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M122
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NK cell stimulatory factor chain 1; IL-12 subunit p35; Cytotoxic lymphocyte maturation factor 35 kDa subunit; NK cell stimulatory factor chain 2; IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
M122 375.00
Check your price
  • IL-17A Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523747
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Western Blot (1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523747 375.00
Check your price
  • IL-2 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523696
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523696 375.00
Check your price
  • IL-22 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523775
  • Size 500µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
IL-10-related T-cell-derived-inducible factor; Cytokine Zcyto18
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523775 375.00
Check your price
  • IL-6 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # M620
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
M620 375.00
Check your price
  • IL-6 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523698
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.05-0.15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interferon beta-2; Hybridoma growth factor; CTL differentiation factor; B-cell stimulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523698 375.00
Check your price
  • IL-6 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523800
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.005-0.025 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Interleukin HP-1; B-cell hybridoma growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523800 375.00
Check your price