Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6481 - 6510 of 48877 products displayed
  • Milk Fat Globule 1 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513553
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
SED1; Milk fat globule-EGF factor 8; MFGM; HMFG; Breast epithelial antigen BA46
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513553 375.00
Check your price
  • MMP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129178
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:2000), Western Blot (1:1000)
Alternate names:
Matrix metalloproteinase-1; Fibroblast collagenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129178 375.00
Check your price
  • MMP16 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129182
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:2000)
Alternate names:
Membrane-type-3 matrix metalloproteinase; Membrane-type matrix metalloproteinase 3; MMP-X2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129182 375.00
Check your price
  • MMP2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129183
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:2000), Western Blot (1:1000)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129183 375.00
Check your price
  • MMP9 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129187
  • Size 500µL
Applications
Immunoprecipitation (1:2000), Western Blot (1:1000)
Alternate names:
Gelatinase B; 92 kDa type IV collagenase; 92 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129187 375.00
Check your price
  • MUC5AC Monoclonal Antibody, Biotin conjugate for IHC (P)
  • Host Mouse
  • Target Species Chicken, Cat, Human, Mouse, Non-human primate, Pig, Rabbit, Rodent, Rat
  • Conjugate Biotin
  • Catalog # MA512175
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Tracheobronchial mucin; Mucin-5 subtype AC, tracheobronchial; Major airway glycoprotein; Lewis B blood group antigen; Gastric mucin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512175 375.00
Check your price
  • Mycobacterium tuberculosis Polyclonal Antibody, FITC conjugate for IF and ICC
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # PA128997
  • Size 500µL
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128997 375.00
Check your price
  • NOP1 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # MA110025
  • Size 500µL
Applications
Immunocytochemistry (1:2000-1:20,000), Immunofluorescence (1:2000-1:20,000), Immunoprecipitation (2-4 µl/lysate), Western Blot (1:10,000-1:25,000)
Alternate names:
U3 small nucleolar RNA-associated protein NOP1; Histone-glutamine methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110025 375.00
Check your price
  • NOTCH1 Monoclonal Antibody for IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA191405
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Translocation-associated notch protein TAN-1; p300; mT14; Motch A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191405 375.00
Check your price
  • NUP107 Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA110031
  • Size 500µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500)
Alternate names:
Nucleoporin Nup107; 107 kDa nucleoporin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110031 375.00
Check your price
  • OATP2/OATP8 Monoclonal Antibody for IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3935
  • Size 500µL
Applications
Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunohistochemistry (Frozen) (1:20), Immunoprecipitation (Assay dependent)
Alternate names:
Solute carrier family 21 member 8; Organic anion-transporting polypeptide 8; Organic anion transporter 8; Liver-specific organic anion transporter 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3935 375.00
Check your price
  • p53 Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA512554
  • Size 500µL
Applications
Flow Cytometry (0.5ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:100-1:200 ), Immunoprecipitation (1:50-1:100 ), Western Blot (1:50-1:200)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512554 375.00
Check your price
  • PAK2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532536
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
p58; p21-activated kinase 2; S6/H4 kinase; PAK65; Gamma-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532536 375.00
Check your price
  • PCNA Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA511355
  • Size 500µL
Applications
Flow Cytometry (1:200-1:1000), Immunocytochemistry (1:20-1:50), Immunofluorescence (1:20-1:50), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:1000)
Alternate names:
Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511355 375.00
Check your price
  • Perlecan Monoclonal Antibody for IF, ICC and IHC (F)
  • Host Rat
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA134860
  • Size 500µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (1:200)
Alternate names:
Perlecan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134860 375.00
Check your price
  • PGP9.5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121097
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Ubiquitin thioesterase L1; PGP 9.5; Neuron cytoplasmic protein 9.5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121097 375.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody, Biotin conjugate for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512691
  • Size 500µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (1:200)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512691 375.00
Check your price
  • PMP22 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191698
  • Size 500µL
Applications
Western Blot (Assay dependent)
Alternate names:
Growth arrest-specific protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191698 375.00
Check your price
  • POMC Polyclonal Antibody for IF, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA185109
  • Size 500µL
Applications
ELISA (1/500-1/1000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1/100-1/200)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185109 375.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for Western Blot, IF, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Chicken, Horse, Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA512642
  • Size 500µL
Applications
Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512642 375.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for IF and IP
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA512653
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512653 375.00
Check your price
  • Progesterone Receptor Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA512658
  • Size 500µL
Applications
Immunofluorescence (10 µg), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512658 375.00
Check your price
  • PTEN Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA512278
  • Size 500µL
Applications
Immunocytochemistry (1:1-1:100), Immunofluorescence (1:1-1:100), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:10-1:100)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512278 375.00
Check your price
  • Rb Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # MA511384
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
pp110; pRb; p105-Rb; pp105
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511384 375.00
Check your price
  • Rhodopsin Monoclonal Antibody, Biotin conjugate for Western Blot, IF and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Biotin
  • Catalog # MA511738
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:25-1:50), Western Blot (1-2 µg/ml)
Alternate names:
Opsin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511738 375.00
Check your price
  • S100 Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA126621
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50-1:100)
Alternate names:
S100 calcium-binding protein A1; S-100 protein subunit alpha; S-100 protein alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126621 375.00
Check your price
  • S100A4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Cat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516586
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
S100 calcium-binding protein A4; Protein Mts1; Placental calcium-binding protein; Metastasin; Calvasculin; P9K; Nerve growth factor-induced protein 42A; Placental calcium-binding protein homolog; Protein 18A2; PEL98; Metastatic cell protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516586 375.00
Check your price
  • S100A6 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Cat, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516590
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
S100 calcium-binding protein A6; Prolactin receptor-associated protein; Calcyclin; 5B10; MLN 4; Growth factor-inducible protein 2A9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516590 375.00
Check your price
  • SV40 Virus Large T Antigen Monoclonal Antibody for IF, ICC and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA190660
  • Size 500µL
Applications
Immunocytochemistry (1-5 ug/ml), Immunofluorescence (1-5 µg/ml), Immunoprecipitation (1-2 µg)
Alternate names:
Simian vacuolating virus 40, Simian virus 40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190660 375.00
Check your price
  • Tau Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA512808
  • Size 500µL
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (2µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512808 375.00
Check your price