Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  40  50  60  70  80  90  100  110  120  200  300  400  500  600  700  800  900  1000  1611 
631 - 660 of 48310 products displayed
  • Glucagon-Like Peptide 1 Antibody (Monoclonal, 8G9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1470602
  • Size 200 µl
Applications
ELISA (1:8000), Immunofluorescence (1:500), Immunohistochemistry (1:200)
Alternate names:
Glucagon-like peptide-1, GLP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1470602 600.00
Check your price
  • Influenza A NP Antibody (Monoclonal, 5D8)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1560102
  • Size 200 µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (Assay dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1560102 600.00
Check your price
  • Influenza B NP Antibody (Monoclonal, 1D8)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160102
  • Size 200 µl
Applications
ELISA (1:200), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (Assay dependent)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160102 600.00
Check your price
  • Influenza B NP Antibody (Monoclonal, 3E9)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160202
  • Size 200 µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160202 600.00
Check your price
  • MBL2 Antibody (Monoclonal, 11C9)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1311102
  • Size 200 µl
Applications
ELISA (1:10,000), Western Blot (Assay dependent)
Alternate names:
Mannose-binding lectin; Mannan-binding protein; MBP1; Collectin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1311102 600.00
Check your price
  • MBL2 Antibody (Monoclonal, 1E2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1311002
  • Size 200 µl
Applications
ELISA (1/40,000), Western Blot (Assay dependent)
Alternate names:
Mannose-binding lectin; Mannan-binding protein; MBP1; Collectin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1311002 600.00
Check your price
  • MBL2 Antibody (Monoclonal, 3B6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1310102
  • Size 200 µl
Applications
ELISA (1/20,000), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay Dependent)
Alternate names:
Mannose-binding lectin; Mannan-binding protein; MBP1; Collectin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1310102 600.00
Check your price
  • Mycobacterium tuberculosis ESAT-6 Antibody (Monoclonal, 11G4)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # HYB0760802
  • Size 200 µl
Applications
ELISA (1:3,200), Western Blot (Assay dependent)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0760802 600.00
Check your price
  • NGAL Antibody (Monoclonal, 29)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # ABS0432902
  • Size 200 µL
Applications
ELISA (1:4000), Immunofluorescence (1:1000), Western Blot (Assay dependent)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0432902 600.00
Check your price
  • PAEP Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0011302
  • Size 200 µl
Applications
ELISA (1/200), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Progesterone-associated endometrial protein; Progestagen-associated endometrial protein; Pregnancy-associated endometrial alpha-2 globulin; Placental protein 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0011302 600.00
Check your price
  • Procollagen I C-Peptide Antibody (Monoclonal, 18)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0021802
  • Size 200 µL
Applications
ELISA (1:3,000), Western Blot (Assay Dependent)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0021802 600.00
Check your price
  • Procollagen I C-Peptide Antibody (Monoclonal, 8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0020802
  • Size 200 µl
Applications
ELISA (1:6,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0020802 600.00
Check your price
  • Procollagen I C-Peptide Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0020402
  • Size 200 µl
Applications
ELISA (1/10,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0020402 600.00
Check your price
  • Procollagen IIA N-Peptide Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # BTE0030202
  • Size 200 µl
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
BTE0030202 600.00
Check your price
  • PYY Antibody (Monoclonal, 1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # ABS0290102
  • Size 200 µL
Applications
ELISA (1:16,000), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Peptide tyrosine tyrosine; PYY-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0290102 600.00
Check your price
  • Thrombospondin 1 Antibody (Monoclonal, A65M)
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # CSI0026502
  • Size 200 µl
Applications
ELISA (1:200), Immunohistochemistry (1:200), Immunoprecipitation (Assay dependent)
Alternate names:
THBS1, TSP, TSP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
CSI0026502 600.00
Check your price
  • Trypsin Antibody (Monoclonal, 2C4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0210802
  • Size 200 µl
Applications
ELISA (1:8000), Western Blot (Assay dependent)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0210802 600.00
Check your price
  • Trypsin Antibody (Monoclonal, 7C8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0210302
  • Size 200 µL
Applications
ELISA (1:8000), Western Blot (Assay dependent)
Alternate names:
Trypsin I; Serine protease 1; Cationic trypsinogen; Beta-trypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0210302 600.00
Check your price
  • HRH4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124553
  • Size 25 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent)
Alternate names:
SP9144; Pfi-013; GPRv53; G-protein coupled receptor 105; AXOR35
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124553 600.00
Check your price
  • CD42a Antibody (Monoclonal, FMC-25)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516684
  • Size 250 ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Glycoprotein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516684 600.00
Check your price
  • Mouse anti-Monkey IgA, Biotin
  • Host Mouse
  • Target Species Non-human primate
  • Conjugate Biotin
  • Catalog # MA516730
  • Size 250 ug
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516730 600.00
Check your price
  • LEF1 Antibody (Monoclonal, REMB1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA112417
  • Size 250 µg
Applications
Immunofluorescence (1:1000), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1-5 ug/ml)
Alternate names:
T cell-specific transcription factor 1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112417 600.00
Check your price
  • Chlamydia pneumoniae Antibody (Monoclonal, RR402)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA516307
  • Size 2mg
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
C. Pneumoniae
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516307 600.00
Check your price
  • CD40 Antibody, PE (Monoclonal, 3/23)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517856
  • Size 300 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
CD40L receptor; Bp50; B-cell surface antigen CD40
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517856 600.00
Check your price
  • CD44 Antibody, PE (Monoclonal, IM7.8.1)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517881
  • Size 300 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Lymphocyte antigen 24; Hyaluronate receptor; Hermes antigen; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517881 600.00
Check your price
  • CD8 beta Antibody, PE (Monoclonal, CT-CD8b)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517850
  • Size 300 ug
Applications
Flow Cytometry (0.1 ug/10^6 cells)
Alternate names:
T-cell surface glycoprotein Lyt-3; T-cell membrane glycoprotein Ly-3; Lymphocyte antigen 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517850 600.00
Check your price
  • Ly-6A/E Antibody, PE (Monoclonal, D7)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517891
  • Size 300 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells)
Alternate names:
T-cell-activating protein; Stem cell antigen 1; Thymic shared antigen 1; Stem cell antigen 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517891 600.00
Check your price
  • Transferrin Receptor Antibody, PE (Monoclonal, R17217.1.4)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517924
  • Size 300 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517924 600.00
Check your price
  • CCL3 Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA132966
  • Size 50 µg
Applications
ELISA (Assay-dependent), Functional Assay (Assay-dependent), Neutralization (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Small-inducible cytokine A3; Macrophage inflammatory protein 1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132966 600.00
Check your price
  • Collagen I Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185312
  • Size 50 µg
Applications
ELISA (1:500-1:3000), Immunohistochemistry (Paraffin, Frozen) (1:10-1:50)
Alternate names:
Alpha-2 type I collagen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185312 600.00
Check your price