Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  240  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1611 
6601 - 6630 of 48310 products displayed
  • ITGA2B/ITGA3 Monoclonal Antibody for Western Blot, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121188
  • Size 50 µL
Applications
ELISA (Assay dependent), Flow Cytometry (1/50-1/100), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (1/50-1/100), Western Blot (1/50-1/100)
Alternate names:
Platelet membrane glycoprotein IIIa; Platelet membrane glycoprotein IIb; GPalpha IIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121188 375.00
Check your price
  • Kallikrein 2 Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15810
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tissue kallikrein-2; Glandular kallikrein-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15810 375.00
Check your price
  • Leptin Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA1051
  • Size 50 µL
Applications
Immunocytochemistry (1:50-1:200), Immunohistochemistry (Frozen) (1:50-1:200), Western Blot (1:200-1:1000)
Alternate names:
Obesity factor; Obese protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1051 375.00
Check your price
  • MAD1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15830
  • Size 50 µL
Applications
Immunofluorescence (1:500-1:1000), Immunoprecipitation (5 µg / 0.5-1 ml), Western Blot (Assay Dependent)
Alternate names:
Tax-binding protein 181; Mitotic checkpoint MAD1 protein homolog; Mitotic arrest deficient 1-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15830 375.00
Check your price
  • MCM7 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126585
  • Size 50 µL
Applications
Western Blot (1:2000-1:10,000)
Alternate names:
P1.1-MCM3; CDC47 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126585 375.00
Check your price
  • MRE11 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, IP and ChIP
  • Host Rabbit
  • Target Species Chicken, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA316527
  • Size 50 µL
Applications
ChIP assay (Assay-Dependent), Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (3 µl), Western Blot (1:5000)
Alternate names:
Meiotic recombination 11 homolog A; Meiotic recombination 11 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316527 375.00
Check your price
  • NBS1 Polyclonal Antibody for IF, ICC, IHC, Flow, IP and ChIP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA316528
  • Size 50 µL
Applications
ChIP assay (Assay-Dependent), Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Assay-Dependent), Immunoprecipitation (3 µl), Western Blot (1:1000)
Alternate names:
Nijmegen breakage syndrome protein 1; Cell cycle regulatory protein p95; Nijmegen breakage syndrome protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316528 375.00
Check your price
  • NEFH Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Chicken
  • Target Species Bovine, Cat, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA316753
  • Size 50 µL
Applications
Immunocytochemistry (1:1000-1:10,000), Immunofluorescence (1:1000-1:10,000), Immunohistochemistry (Frozen) (1:1000-1:10,000), Immunohistochemistry (Paraffin) (1:1000-1:10,000), Western Blot (1:5000-1:10,000)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316753 375.00
Check your price
  • NEFH Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Bovine, Cat, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA316721
  • Size 50 µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:1000), Western Blot (1:10,000)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316721 375.00
Check your price
  • NMDAR2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA34664
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
N-methyl D-aspartate receptor subtype 2A; Glutamate [NMDA] receptor subunit epsilon-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA34664 375.00
Check your price
  • NMDAR2B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA34665
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
N-methyl-D-aspartate receptor subunit 3; N-methyl D-aspartate receptor subtype 2B; Glutamate [NMDA] receptor subunit epsilon-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA34665 375.00
Check your price
  • OPRM1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Bovine, Guinea pig, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA124628
  • Size 50 µL
Applications
Immunocytochemistry (1:2500-1:5000), Immunofluorescence (1:2500-1:5000), Immunohistochemistry (Frozen) (1:2500-1:5000), Western Blot (1:1000)
Alternate names:
Opioid receptor B; Mu opioid receptor; Mu opiate receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124628 375.00
Check your price
  • OVCA1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA15858
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Ovarian cancer-associated gene 1 protein; Diphthamide biosynthesis protein 2 homolog-like 1; DPH1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15858 375.00
Check your price
  • OVCA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA15860
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15860 375.00
Check your price
  • P2X2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Guinea pig, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA124624
  • Size 50 µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Frozen) (1:1000), Western Blot (1:1000)
Alternate names:
Purinergic receptor; ATP receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124624 375.00
Check your price
  • P2X2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Guinea Pig
  • Target Species Guinea pig, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA124623
  • Size 50 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (1:500), Western Blot (1:500)
Alternate names:
Purinergic receptor; ATP receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124623 375.00
Check your price
  • P2X3 Polyclonal Antibody for IF, IHC (F) and IM
  • Host Guinea Pig
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA127969
  • Size 50 µL
Applications
Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:1000), Immunomicroscopy (Assay-Dependent)
Alternate names:
Purinergic receptor; ATP receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127969 375.00
Check your price
  • Paraquat Polyclonal Antibody for RIA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA185762
  • Size 50 µL
Applications
Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185762 375.00
Check your price
  • PARP Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3950
  • Size 50 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:1000-1:5000)
Alternate names:
Poly[ADP-ribose] synthase 1; NAD(+) ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3950 375.00
Check your price
  • Phospho-beta Arrestin 1 (Ser412) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA126637
  • Size 50 µL
Applications
Western Blot (0.1-1 µg/ml)
Alternate names:
Arrestin beta-1; Arrestin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126637 375.00
Check your price
  • Phospho-beta Catenin (Ser33, Ser37) Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125084
  • Size 50 µL
Applications
ELISA (5-10 µg/ml), Western Blot (5-10 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125084 375.00
Check your price
  • Phospho-Btk (Ser180) Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191571
  • Size 50 µL
Applications
Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
Bruton tyrosine kinase; B-cell progenitor kinase; Agammaglobulinemia tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191571 375.00
Check your price
  • Phospho-ERK5 (Thr218) Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126728
  • Size 50 µL
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:1000)
Alternate names:
Extracellular signal-regulated kinase 5; Big MAP kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126728 375.00
Check your price
  • Phospho-HSP27 (Ser82) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125983
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125983 375.00
Check your price
  • Phospho-MARCKS (Ser152, Ser153) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129457
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Protein kinase C substrate, 80 kDa protein, light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129457 375.00
Check your price
  • Phospho-PAG3 (Tyr724) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126006
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Pyk2 C-terminus-associated protein; Paxillin-associated protein with ARF GAP activity 3; Development and differentiation-enhancing factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126006 375.00
Check your price
  • Phospho-POLR2A (Ser5) Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human, Yeast
  • Conjugate Unconjugated
  • Catalog # MA15875
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
RNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; DNA-directed RNA polymerase II subunit A; DNA polymerase II subunit A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15875 375.00
Check your price
  • Phospho-RSK1 (Ser221) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124769
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124769 375.00
Check your price
  • Phospho-S6 (Ser235, Ser236) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125544
  • Size 50 µL
Applications
Western Blot (1:1000)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125544 375.00
Check your price
  • Phosphothreonine Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA191728
  • Size 50 µL
Applications
ELISA (1:2000-1:10000), Western Blot (1:1000)
Alternate names:
pThr
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191728 375.00
Check your price