Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  240  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1630 
6661 - 6690 of 48882 products displayed
  • CENPF Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate DyLight® 488
  • Catalog # PA522933
  • Size 50µL
Applications
Immunocytochemistry (1:750), Immunofluorescence (1:750), Immunoprecipitation (1:50), Western Blot (1:1500)
Alternate names:
Mitosin; Kinetochore protein CENPF; AH antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522933 375.00
Check your price
  • CENPF Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate DyLight® 550
  • Catalog # PA522935
  • Size 50µL
Applications
Immunocytochemistry (1:750), Immunofluorescence (1:750), Immunoprecipitation (1:50), Western Blot (1:1500)
Alternate names:
Mitosin; Kinetochore protein CENPF; AH antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522935 375.00
Check your price
  • CENPF Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate DyLight® 650
  • Catalog # PA522932
  • Size 50µL
Applications
Immunocytochemistry (1:750), Immunofluorescence (1:750), Immunoprecipitation (1:50), Western Blot (1:1500)
Alternate names:
Mitosin; Kinetochore protein CENPF; AH antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522932 375.00
Check your price
  • CENPF Polyclonal Antibody, HRP conjugate for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate HRP
  • Catalog # PA522934
  • Size 50µL
Applications
Immunocytochemistry (1:750), Immunofluorescence (1:750), Immunoprecipitation (1:50), Western Blot (1:1500)
Alternate names:
Mitosin; Kinetochore protein CENPF; AH antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522934 375.00
Check your price
  • Coilin Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA125322
  • Size 50µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Western Blot (1-2 µg/ml)
Alternate names:
p80-coilin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125322 375.00
Check your price
  • Drebrin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA120222
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Developmentally-regulated brain protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120222 375.00
Check your price
  • EME1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15767
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
MMS4 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15767 375.00
Check your price
  • FANCD2 Polyclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Biotin
  • Catalog # PA522704
  • Size 50µL
Applications
Immunocytochemistry (1:200-1:500), Immunofluorescence (1:200-1:500), Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (1:10,000-1:20,000)
Alternate names:
FA-D2, FA4, FACD, FAD, FAD2, FANCD, Fanconi anemia group D2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522704 375.00
Check your price
  • FANCD2 Polyclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate DyLight® 488
  • Catalog # PA522706
  • Size 50µL
Applications
Immunocytochemistry (1:200-1:500), Immunofluorescence (1:200-1:500), Immunohistochemistry (2.5-5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (1:10,000-1:20,000)
Alternate names:
FA-D2, FA4, FACD, FAD, FAD2, FANCD, Fanconi anemia group D2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522706 375.00
Check your price
  • FANCD2 Polyclonal Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate DyLight® 550
  • Catalog # PA522708
  • Size 50µL
Applications
Immunocytochemistry (1:200-1:500), Immunofluorescence (1:200-1:500), Immunohistochemistry (2.5-5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (1:10,000-1:20,000)
Alternate names:
FA-D2, FA4, FACD, FAD, FAD2, FANCD, Fanconi anemia group D2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522708 375.00
Check your price
  • FANCD2 Polyclonal Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate DyLight® 650
  • Catalog # PA522705
  • Size 50µL
Applications
Immunocytochemistry (1:200-1:500), Immunofluorescence (1:200-1:500), Immunohistochemistry (2.5-5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (1:10,000-1:20,000)
Alternate names:
FA-D2, FA4, FACD, FAD, FAD2, FANCD, Fanconi anemia group D2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522705 375.00
Check your price
  • FANCD2 Polyclonal Antibody, HRP conjugate for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate HRP
  • Catalog # PA522707
  • Size 50µL
Applications
Immunocytochemistry (1:200-1:500), Immunofluorescence (1:200-1:500), Immunohistochemistry (Paraffin) (2.5-5 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (1:10,000-1:20,000)
Alternate names:
FA-D2, FA4, FACD, FAD, FAD2, FANCD, Fanconi anemia group D2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522707 375.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124764
  • Size 50µL
Applications
Immunohistochemistry (Paraffin) (1:1000), Immunoprecipitation (1:1,000), Western Blot (1:2,000)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124764 375.00
Check your price
  • GAP43 Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Rabbit
  • Target Species Cat, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA124970
  • Size 50µL
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:1000)
Alternate names:
pp46; Neural phosphoprotein B-50; Growth-associated protein 43; Axonal membrane protein GAP-43; Protein F1; Calmodulin-binding protein P-57
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124970 375.00
Check your price
  • GFAP Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Cat, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA316727
  • Size 50µL
Applications
Immunocytochemistry (1:1000-1:5000), Immunofluorescence (1:1000-1:5000), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:1000-1:5000), Western Blot (1:50,000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316727 375.00
Check your price
  • GFAP Polyclonal Antibody for Western Blot, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19565
  • Size 50µL
Applications
Immunocytochemistry (1:500-1:1000), Immunohistochemistry (1:500-1:1000), Western Blot (1:1000-1:2000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19565 375.00
Check your price
  • Glucocorticoid Receptor beta Polyclonal Antibody for Western Blot, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA3514
  • Size 50µL
Applications
Immunocytochemistry (1:1,500), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3514 375.00
Check your price
  • hnRNP A1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126736
  • Size 50µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
hnRNP core protein A1; Single-strand RNA-binding protein; Helix-destabilizing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126736 375.00
Check your price
  • HSC70 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Cat, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3014
  • Size 50µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Frozen) (1:20-1:200), Immunoprecipitation (2µl), Western Blot (1:1000-1:5000)
Alternate names:
Heat shock 70 kDa protein 8; Lipopolysaccharide-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3014 375.00
Check your price
  • HSL Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA116966
  • Size 50µL
Applications
Western Blot (Assay dependent)
Alternate names:
HSL antibody, hormone sensitive lipase antibody.
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116966 375.00
Check your price
  • HSP70 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and GS
  • Host Mouse
  • Target Species Amphibian, Arthropod, Avian, Fruit fly, Fish, Human, Mollusc, Mouse, Non-human primate, Plant, Pig, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA3006
  • Size 50µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:100), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (2 µl), Western Blot (1:1000-1:5000)
Alternate names:
Hsp68; Heat shock 70 kDa protein 3; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3006 375.00
Check your price
  • IFN beta Polyclonal Antibody for Neu, Ctrl and FN
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128044
  • Size 50µL
Applications
Control (Assay dependent), Functional Assay (Assay-dependent), Neutralization (Assay-dependent)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128044 375.00
Check your price
  • ITGA2B/ITGA3 Monoclonal Antibody for Western Blot, IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121188
  • Size 50µL
Applications
ELISA (Assay dependent), Flow Cytometry (1/50-1/100), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (1/50-1/100), Western Blot (1/50-1/100)
Alternate names:
Platelet membrane glycoprotein IIIa; Platelet membrane glycoprotein IIb; GPalpha IIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121188 375.00
Check your price
  • Kallikrein 2 Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15810
  • Size 50µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tissue kallikrein-2; Glandular kallikrein-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15810 375.00
Check your price
  • Leptin Polyclonal Antibody for Western Blot, ICC and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA1051
  • Size 50µL
Applications
Immunocytochemistry (1:50-1:200), Immunohistochemistry (Frozen) (1:50-1:200), Western Blot (1:200-1:1000)
Alternate names:
Obesity factor; Obese protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1051 375.00
Check your price
  • MAD1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA15830
  • Size 50µL
Applications
Immunofluorescence (1:500-1:1000), Immunoprecipitation (5 µg / 0.5-1 ml), Western Blot (Assay Dependent)
Alternate names:
Tax-binding protein 181; Mitotic checkpoint MAD1 protein homolog; Mitotic arrest deficient 1-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15830 375.00
Check your price
  • MCM7 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA126585
  • Size 50µL
Applications
Western Blot (1:2000-1:10,000)
Alternate names:
P1.1-MCM3; CDC47 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126585 375.00
Check your price
  • MRE11 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, IP and ChIP
  • Host Rabbit
  • Target Species Chicken, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA316527
  • Size 50µL
Applications
ChIP assay (Assay-Dependent), Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (3 µl), Western Blot (1:5000)
Alternate names:
Meiotic recombination 11 homolog A; Meiotic recombination 11 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316527 375.00
Check your price
  • NBS1 Polyclonal Antibody for IF, ICC, IHC, Flow, IP and ChIP
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA316528
  • Size 50µL
Applications
ChIP assay (Assay-Dependent), Flow Cytometry (Assay-Dependent), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Assay-Dependent), Immunoprecipitation (3 µl), Western Blot (1:1000)
Alternate names:
Nijmegen breakage syndrome protein 1; Cell cycle regulatory protein p95; Nijmegen breakage syndrome protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316528 375.00
Check your price
  • NEFH Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Chicken
  • Target Species Bovine, Cat, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA316753
  • Size 50µL
Applications
Immunocytochemistry (1:1000-1:10,000), Immunofluorescence (1:1000-1:10,000), Immunohistochemistry (Frozen) (1:1000-1:10,000), Immunohistochemistry (Paraffin) (1:1000-1:10,000), Western Blot (1:5000-1:10,000)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316753 375.00
Check your price