Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1605 
6811 - 6840 of 48138 products displayed
  • Maspin Antibody Monoclonal (EAW24) (MA5-12383)
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA512383
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:20-1:200)
Alternate names:
Peptidase inhibitor 5; Maspin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512383 359.00
Check your price
  • Prostate Stem Cell Antigen Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA120936
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:75)
Alternate names:
PRO232
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120936 359.00
Check your price
  • OXTR / Oxytocin Receptor Antibody Polyclonal (PA5-32962)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA532962
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (40 µg/ml)
Alternate names:
OT-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532962 359.00
Check your price
  • Glucose-6-Phosphate Dehydrogenase Antibody Polyclonal (PA1-84814)
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate
  • Catalog # PA184814
  • Size 50µg
Applications
Immunoprecipitation (2-10 µg/mg protein), Western Blot (1:1000-1:10000)
Alternate names:
G6PD1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184814 359.00
Check your price
  • MPM2 Antibody Monoclonal (0.T.181) (MA1-91855)
  • Host Mouse
  • Target Species Human, Yeast
  • Conjugate
  • Catalog # MA191855
  • Size 50µg
Applications
ELISA (Assay dependent), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 ug/ml), Western Blot (Assay Dependent)
Alternate names:
Winged-helix factor from INS-1 cells; Transcription factor Trident; MPM-2 reactive phosphoprotein 2; M-phase phosphoprotein 2; Hepatocyte nuclear factor 3 forkhead homolog 11; Forkhead-related protein FKHL16
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191855 359.00
Check your price
  • Phospho-Rb (Thr356) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # OPA103200
  • Size 50µg
Applications
Western Blot (0.5 ug/ml)
Alternate names:
pp110; pRb; p105-Rb
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA103200 359.00
Check your price
  • Histone H2B Antibody Polyclonal (PA1-22173)
  • Host Rabbit
  • Target Species Mammal
  • Conjugate
  • Catalog # PA122173
  • Size 50µL
Applications
ELISA (1:300-1:500), Immunocytochemistry (1:100-1:300), Immunofluorescence (1:100-1:300), Immunoprecipitation (Assay dependent), Western Blot (1:300-1:500)
Alternate names:
Histone H2B.f; Histone H2B.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122173 359.00
Check your price
  • N-epsilon acetyl Lysine Antibody Monoclonal (3C155) (MA1-91609)
  • Host Mouse
  • Target Species Chemical
  • Conjugate
  • Catalog # MA191609
  • Size 50µL
Applications
ELISA (1:5000-1:50,000), Immunohistochemistry (1:500-1:5000), Western Blot (1:500-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191609 359.00
Check your price
  • Phospho-PKA 2 beta pSer114 Antibody
  • Host Rabbit
  • Target Species Rat, Bovine, Human, Mouse, Rabbit
  • Conjugate
  • Catalog # PA126735
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126735 359.00
Check your price
  • Mel-CAM / CD146 Antibody Polyclonal (PA1-37381)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA137381
  • Size 7mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
S-endo 1 endothelial-associated antigen; Melanoma-associated antigen MUC18; Melanoma-associated antigen A32; Melanoma cell adhesion molecule; Cell surface glycoprotein P1H12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137381 359.00
Check your price
  • Melan A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA128212
  • Size 7mL
Applications
Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Protein Melan-A; Antigen SK29-AA; Antigen LB39-AA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128212 359.00
Check your price
  • NDUFA9 Antibody (Monoclonal, 20C11B11B11)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 459100
  • Size 100 µg
Applications
Western Blot (1.0 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 39 kDa subunit; Complex I-39kD; Sperm flagella protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
459100 358.00
Check your price
  • CD62L Antibody, Pacific Blue™ (Monoclonal, Dreg-56)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD62L28
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
gp90-MEL; TQ1; Lymph node homing receptor; Leukocyte-endothelial cell adhesion molecule 1; Leukocyte surface antigen Leu-8; Leukocyte adhesion molecule 1; CD62 antigen-like family member L
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD62L28 357.00
Check your price
  • CD63 Antibody, PE (Monoclonal, CLB-gran/12 (CLB-180))
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MHCD6304
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Tetraspanin-30; Ocular melanoma-associated antigen; OMA81H; Melanoma-associated antigen ME491; Lysosomal-associated membrane protein 3; Granulophysin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD6304 357.00
Check your price
  • SSEA1 Antibody, Pacific Blue™ (Monoclonal, VIMC6)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD1528
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
stage-specific embryonic antigen 1, SSEA1, MC-480, CD15, SSEA-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD1528 357.00
Check your price
  • Goat anti-Canine IgM, FITC
  • Host Goat
  • Target Species Dog
  • Conjugate FITC
  • Catalog # PA184637
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (1/200-1/2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184637 357.00
Check your price
  • c-Met Antibody (Monoclonal, 3D4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 187366
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Tyrosine-protein kinase Met; Scatter factor receptor; Proto-oncogene c-Met; HGF/SF receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
187366 357.00
Check your price
  • CD20 Antibody, Pacific Blue™ (Monoclonal, HI47)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD2028
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Membrane-spanning 4-domains subfamily A member 1; Leukocyte surface antigen Leu-16; Bp35; B-lymphocyte surface antigen B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD2028 356.00
Check your price
  • CD38 Antibody, APC-Cyanine5.5 (Monoclonal, HIT2)
  • Host Mouse
  • Target Species Human
  • Conjugate APC-Cyanine5.5
  • Catalog # MHCD3819
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T10; Cyclic ADP-ribose hydrolase 1; ADP-ribosyl cyclase 1; 2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD3819 356.00
Check your price
  • CD3d Antibody, Pacific Blue™ (Monoclonal, S4.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD0328
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor T3 delta chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0328 356.00
Check your price
  • CD4 Antibody, Alexa Fluor® 700 (Monoclonal, S3.5)
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 700
  • Catalog # MHCD0429
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0429 356.00
Check your price
  • CD8 Antibody, APC-Cyanine5.5 (Monoclonal, 3B5)
  • Host Mouse
  • Target Species Human
  • Conjugate APC-Cyanine5.5
  • Catalog # MHCD0819
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0819 356.00
Check your price
  • FAS Antibody, APC (Monoclonal, DX2)
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MHCD9505
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
FASLG receptor; Apoptosis-mediating surface antigen FAS; Apo-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD9505 356.00
Check your price
  • alpha Tubulin Antibody, Alexa Fluor® 488 (Monoclonal, B-5-1-2)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # 322588
  • Size 100 µg
Applications
Immunocytochemistry (2.5-5 µg/ml), Immunofluorescence (2.5-5 ug/ml)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
322588 356.00
Check your price
  • Complex IV Antibody (Monoclonal, DA5BC4)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # 459300
  • Size 100 µg
Applications
Western Blot (2-4 ug/ml)
Alternate names:
COX4, COX4-1, COXIV, COX IV-1, cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
459300 356.00
Check your price
  • FXN Antibody (Monoclonal, 18A5DB1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 456300
  • Size 100 µg
Applications
Immunocytochemistry (0.5 ug/ml), Immunofluorescence (Assay Dependent), Western Blot (3-5 ug/ml)
Alternate names:
Friedreich ataxia protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
456300 356.00
Check your price
  • UQCRC1 Antibody (Monoclonal, 16D10AD9AH5)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 459140
  • Size 100 µg
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Ubiquinol-cytochrome-c reductase complex core protein 1; Core protein I; Complex III subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
459140 356.00
Check your price
  • Rat IFN alpha 2 Recombinant Protein
  • Host E. Coli
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # 131001
  • Size 1x10^5 units
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon alpha-D
Catalog # Size Price (AUD) Your Price (AUD) Qty
131001 356.00
Check your price
  • CD8 Antibody, Pacific Blue™ (Monoclonal, 3B5)
  • Host Mouse
  • Target Species Human
  • Conjugate Pacific Blue™
  • Catalog # MHCD0828
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD0828 356.00
Check your price
  • TNFR2 Antibody, PE (Monoclonal, MR2-1)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # TNFR7504
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
p80 TNF-alpha receptor; p75; Tumor necrosis factor receptor type II; Tumor necrosis factor receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
TNFR7504 356.00
Check your price